Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54419235-54419775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTYH1
Entrez Gene ID
57348
Gene Name
tweety family member 1
Gene Aliases
-
UniGene
Hs.268728
Gene Chromosome Location
Chr.19: 54415431-54436719 on Build GRCh38
Transcripts
NM_001005367.2 NM_001201461.1 NM_020659.3 XM_017027025.1 XM_017027026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4956089 c.254C>T p.P85L Chr.19: 54419255-54419255 on Build GRCh38
4956088 c.254C>T p.P85L Chr.19: 54419255-54419255 on Build GRCh38
5091850 c.399G>A p.S133S Chr.19: 54419253-54419253 on Build GRCh38
4932430 c.256G>T p.G86W Chr.19: 54419257-54419257 on Build GRCh38
4932431 c.256G>T p.G86W Chr.19: 54419257-54419257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563381544 Chr.19: 54419333-54419333 on Build GRCh38
rs1044912181 Chr.19: 54419740-54419740 on Build GRCh38
rs201300105 Chr.19: 54419320-54419320 on Build GRCh38
rs764899506 Chr.19: 54419555-54419555 on Build GRCh38
rs547324487 Chr.19: 54419375-54419375 on Build GRCh38
rs753780211 Chr.19: 54419358-54419358 on Build GRCh38
rs558026127 Chr.19: 54419585-54419585 on Build GRCh38
rs903936206 Chr.19: 54419535-54419535 on Build GRCh38
rs3745432 Chr.19: 54419298-54419298 on Build GRCh38
rs62621451 Chr.19: 54419450-54419450 on Build GRCh38
rs3745431 Chr.19: 54419353-54419353 on Build GRCh38
rs369700865 Chr.19: 54419254-54419254 on Build GRCh38
rs564798025 Chr.19: 54419255-54419255 on Build GRCh38
rs929601976 Chr.19: 54419741-54419741 on Build GRCh38
rs983228136 Chr.19: 54419377-54419377 on Build GRCh38
rs747176218 Chr.19: 54419552-54419552 on Build GRCh38
rs974196148 Chr.19: 54419707-54419707 on Build GRCh38
rs772328872 Chr.19: 54419325-54419325 on Build GRCh38
rs11556553 Chr.19: 54419258-54419258 on Build GRCh38
rs766807087 Chr.19: 54419332-54419332 on Build GRCh38
rs532464025 Chr.19: 54419739-54419739 on Build GRCh38
rs544098978 Chr.19: 54419686-54419686 on Build GRCh38
rs980918968 Chr.19: 54419666-54419666 on Build GRCh38
rs1000971067 Chr.19: 54419291-54419291 on Build GRCh38
rs150993074 Chr.19: 54419269-54419269 on Build GRCh38
rs761325643 Chr.19: 54419259-54419259 on Build GRCh38
rs546407473 Chr.19: 54419462-54419462 on Build GRCh38
rs780000024 Chr.19: 54419301-54419301 on Build GRCh38
rs569687507 Chr.19: 54419426-54419426 on Build GRCh38
rs1013608066 Chr.19: 54419593-54419593 on Build GRCh38
rs779767219 Chr.19: 54419355-54419355 on Build GRCh38
rs774427522 Chr.19: 54419316-54419316 on Build GRCh38
rs918172877 Chr.19: 54419336-54419336 on Build GRCh38
rs1031116154 Chr.19: 54419494-54419494 on Build GRCh38
rs3745433 Chr.19: 54419257-54419257 on Build GRCh38
rs142605148 Chr.19: 54419256-54419256 on Build GRCh38
rs1056187723 Chr.19: 54419388-54419388 on Build GRCh38
rs535211427 Chr.19: 54419521-54419521 on Build GRCh38
rs760057923 Chr.19: 54419339-54419339 on Build GRCh38
rs1038036418 Chr.19: 54419401-54419401 on Build GRCh38
rs779322857 Chr.19: 54419313-54419313 on Build GRCh38
rs118137187 Chr.19: 54419421-54419421 on Build GRCh38
rs768683157 Chr.19: 54419253-54419253 on Build GRCh38
rs763517110 Chr.19: 54419342-54419342 on Build GRCh38
rs373670702 Chr.19: 54419302-54419302 on Build GRCh38
rs191550695 Chr.19: 54419385-54419385 on Build GRCh38
rs778169809 Chr.19: 54419517-54419517 on Build GRCh38
rs74693637 Chr.19: 54419687-54419687 on Build GRCh38
rs1039536308 Chr.19: 54419533-54419533 on Build GRCh38
rs756867029 Chr.19: 54419273-54419273 on Build GRCh38
rs758009799 Chr.19: 54419264-54419264 on Build GRCh38
rs956883481 Chr.19: 54419570-54419570 on Build GRCh38
rs1027068873 Chr.19: 54419580-54419580 on Build GRCh38
rs1025200995 Chr.19: 54419610-54419610 on Build GRCh38
rs374613293 Chr.19: 54419329-54419329 on Build GRCh38
rs752292747 Chr.19: 54419262-54419262 on Build GRCh38
rs73606750 Chr.19: 54419743-54419743 on Build GRCh38
rs1011954911 Chr.19: 54419287-54419287 on Build GRCh38
rs1031095510 Chr.19: 54419569-54419569 on Build GRCh38
rs771432079 Chr.19: 54419553-54419553 on Build GRCh38
rs770916672 Chr.19: 54419688-54419688 on Build GRCh38
rs140451581 Chr.19: 54419461-54419461 on Build GRCh38
rs539307506 Chr.19: 54419394-54419394 on Build GRCh38
rs931317136 Chr.19: 54419356-54419356 on Build GRCh38
rs116554903 Chr.19: 54419709-54419709 on Build GRCh38
rs1019800552 Chr.19: 54419324-54419324 on Build GRCh38
rs779232053 Chr.19: 54419542-54419542 on Build GRCh38
rs773057009 Chr.19: 54419673-54419673 on Build GRCh38
rs754593165 Chr.19: 54419348-54419348 on Build GRCh38
rs758945236 Chr.19: 54419386-54419386 on Build GRCh38
rs572493433 Chr.19: 54419472-54419472 on Build GRCh38
rs376518513 Chr.19: 54419261-54419261 on Build GRCh38
rs940589969 Chr.19: 54419686-54419687 on Build GRCh38
rs947779185 Chr.19: 54419427-54419427 on Build GRCh38
rs1008485352 Chr.19: 54419520-54419520 on Build GRCh38
rs767174570 Chr.19: 54419346-54419346 on Build GRCh38
rs370969915 Chr.19: 54419327-54419327 on Build GRCh38
rs750040497 Chr.19: 54419350-54419350 on Build GRCh38
rs756917154 Chr.19: 54419344-54419344 on Build GRCh38
rs35134302 Chr.19: 54419648-54419648 on Build GRCh38
rs1019769599 Chr.19: 54419549-54419549 on Build GRCh38
rs768736373 Chr.19: 54419306-54419306 on Build GRCh38
rs751075560 Chr.19: 54419343-54419343 on Build GRCh38
rs747781504 Chr.19: 54419567-54419567 on Build GRCh38
rs763606814 Chr.19: 54419268-54419268 on Build GRCh38
rs142265106 Chr.19: 54419289-54419289 on Build GRCh38

View Full Product Details