Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55186351-55186760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRH
Entrez Gene ID
5794
Gene Name
protein tyrosine phosphatase, receptor type H
Gene Aliases
R-PTP-H, SAP1
UniGene
Hs.179770
Gene Chromosome Location
Chr.19: 55181247-55209506 on Build GRCh38
Transcripts
NM_001161440.2 NM_002842.4 XM_011527183.1 XM_011527188.1 XM_017027056.1 XM_017027057.1 XM_017027058.1 XM_017027059.1 XM_017027060.1 XM_017027061.1 XM_017027062.1 XM_017027063.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1001416 c.2647C>A p.L883I Chr.19: 55186356-55186356 on Build GRCh38
3757239 c.2586C>T p.P862P Chr.19: 55186521-55186521 on Build GRCh38
374325 c.2643+1G>T p.? Chr.19: 55186463-55186463 on Build GRCh38
3423180 c.2578C>T p.R860W Chr.19: 55186529-55186529 on Build GRCh38
4413996 c.2584C>T p.P862S Chr.19: 55186523-55186523 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74901631 Chr.19: 55186463-55186463 on Build GRCh38
rs751998411 Chr.19: 55186515-55186515 on Build GRCh38
rs775720660 Chr.19: 55186589-55186589 on Build GRCh38
rs570342111 Chr.19: 55186643-55186643 on Build GRCh38
rs1025710334 Chr.19: 55186648-55186648 on Build GRCh38
rs918681846 Chr.19: 55186474-55186474 on Build GRCh38
rs77296713 Chr.19: 55186398-55186398 on Build GRCh38
rs1032765339 Chr.19: 55186392-55186392 on Build GRCh38
rs747421432 Chr.19: 55186670-55186670 on Build GRCh38
rs1026185310 Chr.19: 55186680-55186680 on Build GRCh38
rs891551751 Chr.19: 55186557-55186557 on Build GRCh38
rs1024404706 Chr.19: 55186564-55186564 on Build GRCh38
rs369001173 Chr.19: 55186455-55186455 on Build GRCh38
rs765753137 Chr.19: 55186584-55186603 on Build GRCh38
rs771175100 Chr.19: 55186480-55186480 on Build GRCh38
rs752984266 Chr.19: 55186432-55186432 on Build GRCh38
rs779280991 Chr.19: 55186464-55186464 on Build GRCh38
rs760631178 Chr.19: 55186543-55186545 on Build GRCh38
rs776967300 Chr.19: 55186561-55186561 on Build GRCh38
rs199808001 Chr.19: 55186466-55186466 on Build GRCh38
rs768657129 Chr.19: 55186388-55186388 on Build GRCh38
rs367805725 Chr.19: 55186528-55186528 on Build GRCh38
rs60494799 Chr.19: 55186640-55186641 on Build GRCh38
rs537635937 Chr.19: 55186669-55186669 on Build GRCh38
rs761575656 Chr.19: 55186384-55186384 on Build GRCh38
rs146444215 Chr.19: 55186623-55186623 on Build GRCh38
rs200391066 Chr.19: 55186406-55186406 on Build GRCh38
rs372031071 Chr.19: 55186401-55186401 on Build GRCh38
rs902962324 Chr.19: 55186391-55186391 on Build GRCh38
rs201283120 Chr.19: 55186442-55186442 on Build GRCh38
rs571966760 Chr.19: 55186531-55186531 on Build GRCh38
rs997311380 Chr.19: 55186628-55186628 on Build GRCh38
rs755791091 Chr.19: 55186518-55186518 on Build GRCh38
rs1007248924 Chr.19: 55186660-55186660 on Build GRCh38
rs745362309 Chr.19: 55186377-55186377 on Build GRCh38
rs778712446 Chr.19: 55186529-55186529 on Build GRCh38
rs768444908 Chr.19: 55186458-55186458 on Build GRCh38
rs374901166 Chr.19: 55186504-55186504 on Build GRCh38
rs746845784 Chr.19: 55186545-55186545 on Build GRCh38
rs1016193700 Chr.19: 55186428-55186428 on Build GRCh38
rs748858726 Chr.19: 55186457-55186457 on Build GRCh38
rs758354543 Chr.19: 55186538-55186538 on Build GRCh38
rs769841540 Chr.19: 55186577-55186577 on Build GRCh38
rs750323454 Chr.19: 55186508-55186513 on Build GRCh38
rs944606209 Chr.19: 55186519-55186519 on Build GRCh38
rs1009884305 Chr.19: 55186558-55186558 on Build GRCh38
rs755837998 Chr.19: 55186456-55186456 on Build GRCh38
rs556029435 Chr.19: 55186736-55186736 on Build GRCh38
rs34037862 Chr.19: 55186565-55186566 on Build GRCh38
rs777503023 Chr.19: 55186541-55186541 on Build GRCh38
rs975060054 Chr.19: 55186715-55186715 on Build GRCh38
rs893403410 Chr.19: 55186533-55186533 on Build GRCh38
rs764491294 Chr.19: 55186506-55186506 on Build GRCh38
rs758894659 Chr.19: 55186434-55186434 on Build GRCh38
rs753317700 Chr.19: 55186503-55186503 on Build GRCh38
rs781043561 Chr.19: 55186375-55186375 on Build GRCh38
rs760739546 Chr.19: 55186407-55186407 on Build GRCh38
rs929922703 Chr.19: 55186493-55186493 on Build GRCh38
rs902928996 Chr.19: 55186627-55186627 on Build GRCh38
rs368363856 Chr.19: 55186426-55186426 on Build GRCh38
rs766388277 Chr.19: 55186416-55186416 on Build GRCh38
rs148069644 Chr.19: 55186385-55186385 on Build GRCh38
rs951453549 Chr.19: 55186652-55186652 on Build GRCh38
rs753268061 Chr.19: 55186524-55186524 on Build GRCh38
rs987223874 Chr.19: 55186494-55186494 on Build GRCh38
rs962836385 Chr.19: 55186636-55186636 on Build GRCh38
rs776544618 Chr.19: 55186484-55186484 on Build GRCh38
rs760839481 Chr.19: 55186468-55186468 on Build GRCh38
rs1018947274 Chr.19: 55186382-55186382 on Build GRCh38
rs777193532 Chr.19: 55186390-55186390 on Build GRCh38
rs372087400 Chr.19: 55186606-55186606 on Build GRCh38
rs181157531 Chr.19: 55186372-55186372 on Build GRCh38
rs947529729 Chr.19: 55186552-55186552 on Build GRCh38
rs1034648089 Chr.19: 55186630-55186630 on Build GRCh38
rs1136578 Chr.19: 55186521-55186521 on Build GRCh38
rs202052377 Chr.19: 55186378-55186378 on Build GRCh38
rs140285575 Chr.19: 55186499-55186499 on Build GRCh38
rs543736826 Chr.19: 55186710-55186710 on Build GRCh38
rs986930518 Chr.19: 55186664-55186664 on Build GRCh38
rs111326663 Chr.19: 55186427-55186427 on Build GRCh38
rs749118407 Chr.19: 55186379-55186379 on Build GRCh38
rs185713498 Chr.19: 55186556-55186556 on Build GRCh38
rs745927408 Chr.19: 55186572-55186572 on Build GRCh38

View Full Product Details