Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53210291-53210800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPT2
Entrez Gene ID
1376
Gene Name
carnitine palmitoyltransferase 2
Gene Aliases
CPT1, CPTASE, IIAE4
UniGene
Hs.713535
Gene Chromosome Location
Chr.1: 53196429-53214197 on Build GRCh38
Transcripts
NM_000098.2 XM_005270484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4674502 c.686C>A p.P229H Chr.1: 53210360-53210360 on Build GRCh38
2193281 c.850C>T p.P284S Chr.1: 53210524-53210524 on Build GRCh38
7244633 c.744A>G p.K248K Chr.1: 53210418-53210418 on Build GRCh38
7127479 c.718G>T p.A240S Chr.1: 53210392-53210392 on Build GRCh38
6009286 c.1074C>G p.L358L Chr.1: 53210748-53210748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769931396 Chr.1: 53210446-53210446 on Build GRCh38
rs985374504 Chr.1: 53210715-53210715 on Build GRCh38
rs753270905 Chr.1: 53210351-53210351 on Build GRCh38
rs1057517729 Chr.1: 53210526-53210526 on Build GRCh38
rs763788839 Chr.1: 53210593-53210593 on Build GRCh38
rs138575554 Chr.1: 53210526-53210526 on Build GRCh38
rs138855128 Chr.1: 53210520-53210520 on Build GRCh38
rs144805101 Chr.1: 53210372-53210372 on Build GRCh38
rs773546947 Chr.1: 53210625-53210625 on Build GRCh38
rs765964579 Chr.1: 53210483-53210483 on Build GRCh38
rs750191719 Chr.1: 53210659-53210659 on Build GRCh38
rs776833476 Chr.1: 53210341-53210341 on Build GRCh38
rs760989930 Chr.1: 53210626-53210626 on Build GRCh38
rs373918498 Chr.1: 53210441-53210441 on Build GRCh38
rs515726175 Chr.1: 53210657-53210657 on Build GRCh38
rs727503887 Chr.1: 53210560-53210560 on Build GRCh38
rs758064377 Chr.1: 53210495-53210495 on Build GRCh38
rs781398541 Chr.1: 53210766-53210766 on Build GRCh38
rs886046409 Chr.1: 53210357-53210357 on Build GRCh38
rs796051944 Chr.1: 53210393-53210393 on Build GRCh38
rs373638740 Chr.1: 53210365-53210365 on Build GRCh38
rs1799821 Chr.1: 53210776-53210776 on Build GRCh38
rs2229291 Chr.1: 53210729-53210729 on Build GRCh38
rs762366252 Chr.1: 53210343-53210343 on Build GRCh38
rs760978599 Chr.1: 53210339-53210339 on Build GRCh38
rs920941550 Chr.1: 53210330-53210330 on Build GRCh38
rs757247552 Chr.1: 53210649-53210649 on Build GRCh38
rs727503888 Chr.1: 53210627-53210627 on Build GRCh38
rs763703786 Chr.1: 53210633-53210633 on Build GRCh38
rs778477892 Chr.1: 53210500-53210500 on Build GRCh38
rs753303896 Chr.1: 53210388-53210388 on Build GRCh38
rs938668766 Chr.1: 53210774-53210774 on Build GRCh38
rs767601469 Chr.1: 53210651-53210651 on Build GRCh38
rs780168485 Chr.1: 53210601-53210601 on Build GRCh38
rs748646501 Chr.1: 53210614-53210614 on Build GRCh38
rs515726174 Chr.1: 53210315-53210315 on Build GRCh38
rs771250137 Chr.1: 53210437-53210437 on Build GRCh38
rs775030648 Chr.1: 53210629-53210629 on Build GRCh38
rs200252755 Chr.1: 53210395-53210395 on Build GRCh38
rs775144404 Chr.1: 53210714-53210714 on Build GRCh38
rs766004699 Chr.1: 53210569-53210570 on Build GRCh38
rs771537809 Chr.1: 53210763-53210763 on Build GRCh38
rs758337938 Chr.1: 53210507-53210507 on Build GRCh38
rs754375091 Chr.1: 53210493-53210493 on Build GRCh38
rs762105213 Chr.1: 53210553-53210553 on Build GRCh38
rs986941002 Chr.1: 53210640-53210640 on Build GRCh38
rs199673903 Chr.1: 53210438-53210438 on Build GRCh38
rs757881397 Chr.1: 53210570-53210570 on Build GRCh38
rs755882353 Chr.1: 53210662-53210662 on Build GRCh38
rs773236568 Chr.1: 53210331-53210331 on Build GRCh38
rs752139090 Chr.1: 53210421-53210421 on Build GRCh38
rs774859940 Chr.1: 53210552-53210552 on Build GRCh38
rs771966581 Chr.1: 53210425-53210425 on Build GRCh38
rs767119836 Chr.1: 53210695-53210695 on Build GRCh38
rs752211265 Chr.1: 53210505-53210505 on Build GRCh38
rs151003641 Chr.1: 53210722-53210722 on Build GRCh38
rs759733220 Chr.1: 53210347-53210347 on Build GRCh38
rs866307911 Chr.1: 53210694-53210694 on Build GRCh38
rs530706050 Chr.1: 53210367-53210367 on Build GRCh38
rs755500312 Chr.1: 53210755-53210755 on Build GRCh38
rs756328565 Chr.1: 53210598-53210598 on Build GRCh38
rs748117051 Chr.1: 53210407-53210407 on Build GRCh38
rs375109382 Chr.1: 53210690-53210690 on Build GRCh38
rs746284903 Chr.1: 53210771-53210771 on Build GRCh38
rs74315298 Chr.1: 53210354-53210354 on Build GRCh38
rs554776730 Chr.1: 53210679-53210679 on Build GRCh38
rs772541350 Chr.1: 53210698-53210698 on Build GRCh38
rs145237292 Chr.1: 53210551-53210551 on Build GRCh38
rs750820184 Chr.1: 53210610-53210610 on Build GRCh38
rs759086398 Chr.1: 53210667-53210668 on Build GRCh38
rs751090469 Chr.1: 53210595-53210597 on Build GRCh38
rs111896041 Chr.1: 53210399-53210399 on Build GRCh38
rs367691827 Chr.1: 53210329-53210329 on Build GRCh38
rs763502641 Chr.1: 53210468-53210468 on Build GRCh38
rs750339299 Chr.1: 53210728-53210728 on Build GRCh38
rs764849762 Chr.1: 53210561-53210561 on Build GRCh38
rs753720446 Chr.1: 53210742-53210742 on Build GRCh38
rs773628654 Chr.1: 53210559-53210559 on Build GRCh38
rs764076288 Chr.1: 53210577-53210577 on Build GRCh38
rs144658100 Chr.1: 53210699-53210699 on Build GRCh38
rs751558734 Chr.1: 53210582-53210582 on Build GRCh38
rs200906458 Chr.1: 53210527-53210527 on Build GRCh38
rs759423137 Chr.1: 53210391-53210391 on Build GRCh38
rs764557610 Chr.1: 53210472-53210472 on Build GRCh38
rs775793760 Chr.1: 53210462-53210462 on Build GRCh38
rs778655162 Chr.1: 53210398-53210398 on Build GRCh38
rs961059441 Chr.1: 53210687-53210687 on Build GRCh38
rs569273347 Chr.1: 53210634-53210634 on Build GRCh38
rs794727616 Chr.1: 53210348-53210348 on Build GRCh38
rs761438840 Chr.1: 53210727-53210727 on Build GRCh38
rs770874001 Chr.1: 53210521-53210521 on Build GRCh38
rs749833236 Chr.1: 53210754-53210754 on Build GRCh38
rs1024639640 Chr.1: 53210685-53210685 on Build GRCh38
rs190459228 Chr.1: 53210517-53210517 on Build GRCh38
rs201165795 Chr.1: 53210661-53210661 on Build GRCh38
rs766080131 Chr.1: 53210737-53210737 on Build GRCh38
rs934539178 Chr.1: 53210368-53210368 on Build GRCh38
rs147889730 Chr.1: 53210350-53210350 on Build GRCh38
rs199894581 Chr.1: 53210746-53210746 on Build GRCh38
rs751888992 Chr.1: 53210474-53210474 on Build GRCh38
rs766770153 Chr.1: 53210340-53210340 on Build GRCh38
rs771214714 Chr.1: 53210707-53210707 on Build GRCh38
rs369369333 Chr.1: 53210366-53210366 on Build GRCh38
rs868159960 Chr.1: 53210666-53210667 on Build GRCh38
rs148512862 Chr.1: 53210721-53210721 on Build GRCh38
rs778946896 Chr.1: 53210696-53210696 on Build GRCh38
rs202145705 Chr.1: 53210544-53210544 on Build GRCh38
rs141553491 Chr.1: 53210586-53210586 on Build GRCh38
rs1057517515 Chr.1: 53210720-53210721 on Build GRCh38
rs767870817 Chr.1: 53210556-53210556 on Build GRCh38
rs752222424 Chr.1: 53210565-53210565 on Build GRCh38
rs561154406 Chr.1: 53210487-53210487 on Build GRCh38
rs773966429 Chr.1: 53210723-53210723 on Build GRCh38
rs745698305 Chr.1: 53210595-53210595 on Build GRCh38
rs777614326 Chr.1: 53210428-53210428 on Build GRCh38
rs752737849 Chr.1: 53210753-53210753 on Build GRCh38
rs762468710 Chr.1: 53210630-53210630 on Build GRCh38
rs914465235 Chr.1: 53210431-53210431 on Build GRCh38
rs780263768 Chr.1: 53210677-53210677 on Build GRCh38
rs762195160 Chr.1: 53210475-53210475 on Build GRCh38
rs541736685 Chr.1: 53210607-53210607 on Build GRCh38
rs747352901 Chr.1: 53210706-53210706 on Build GRCh38
rs759640597 Chr.1: 53210440-53210440 on Build GRCh38
rs142790440 Chr.1: 53210606-53210606 on Build GRCh38
rs772621468 Chr.1: 53210617-53210617 on Build GRCh38
rs746853000 Chr.1: 53210433-53210433 on Build GRCh38
rs201163382 Chr.1: 53210524-53210524 on Build GRCh38
rs371971257 Chr.1: 53210604-53210604 on Build GRCh38

View Full Product Details