Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 106804396-106804897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST6GAL2
Entrez Gene ID
84620
Gene Name
ST6 beta-galactoside alpha-2,6-sialyltransferase 2
Gene Aliases
SIAT2, ST6GalII
UniGene
Hs.98265
Gene Chromosome Location
Chr.2: 106801600-106887410 on Build GRCh38
Transcripts
NM_001142351.1 NM_001322362.1 NM_032528.2 NR_136313.1 XM_006712802.1 XM_011511999.2

SNP Detail

SNP ID Chromosome Location
rs992294698 Chr.2: 106804870-106804870 on Build GRCh38
rs1021410871 Chr.2: 106804788-106804788 on Build GRCh38
rs115513984 Chr.2: 106804579-106804579 on Build GRCh38
rs192009466 Chr.2: 106804651-106804651 on Build GRCh38
rs988735170 Chr.2: 106804782-106804782 on Build GRCh38
rs1039741565 Chr.2: 106804797-106804797 on Build GRCh38
rs938165127 Chr.2: 106804864-106804864 on Build GRCh38
rs966478909 Chr.2: 106804580-106804580 on Build GRCh38
rs976000041 Chr.2: 106804529-106804529 on Build GRCh38
rs552748323 Chr.2: 106804855-106804855 on Build GRCh38
rs994290664 Chr.2: 106804491-106804491 on Build GRCh38
rs1005962968 Chr.2: 106804508-106804508 on Build GRCh38
rs70956211 Chr.2: 106804859-106804860 on Build GRCh38
rs570782307 Chr.2: 106804820-106804820 on Build GRCh38
rs575253861 Chr.2: 106804469-106804469 on Build GRCh38
rs372637582 Chr.2: 106804765-106804765 on Build GRCh38
rs977820444 Chr.2: 106804607-106804607 on Build GRCh38
rs1006406829 Chr.2: 106804661-106804661 on Build GRCh38
rs1051139070 Chr.2: 106804507-106804507 on Build GRCh38
rs1051194464 Chr.2: 106804773-106804774 on Build GRCh38
rs960672450 Chr.2: 106804637-106804637 on Build GRCh38
rs887449681 Chr.2: 106804657-106804657 on Build GRCh38
rs533591729 Chr.2: 106804678-106804678 on Build GRCh38
rs1017894744 Chr.2: 106804511-106804511 on Build GRCh38
rs779486051 Chr.2: 106804590-106804590 on Build GRCh38
rs896702563 Chr.2: 106804628-106804628 on Build GRCh38
rs369946025 Chr.2: 106804806-106804806 on Build GRCh38
rs964496492 Chr.2: 106804527-106804527 on Build GRCh38
rs6543444 Chr.2: 106804710-106804710 on Build GRCh38
rs1026067512 Chr.2: 106804430-106804430 on Build GRCh38
rs184244118 Chr.2: 106804796-106804796 on Build GRCh38
rs563759390 Chr.2: 106804778-106804778 on Build GRCh38
rs938134083 Chr.2: 106804595-106804595 on Build GRCh38
rs1056752699 Chr.2: 106804608-106804608 on Build GRCh38
rs10650737 Chr.2: 106804841-106804842 on Build GRCh38
rs866469012 Chr.2: 106804821-106804821 on Build GRCh38
rs371700636 Chr.2: 106804865-106804865 on Build GRCh38
rs893125483 Chr.2: 106804856-106804857 on Build GRCh38
rs550797123 Chr.2: 106804777-106804777 on Build GRCh38
rs928965389 Chr.2: 106804638-106804638 on Build GRCh38
rs561855105 Chr.2: 106804701-106804701 on Build GRCh38
rs556593568 Chr.2: 106804437-106804437 on Build GRCh38
rs918760390 Chr.2: 106804693-106804693 on Build GRCh38
rs923162412 Chr.2: 106804538-106804538 on Build GRCh38
rs1006863399 Chr.2: 106804547-106804550 on Build GRCh38
rs545750973 Chr.2: 106804564-106804564 on Build GRCh38
rs955392143 Chr.2: 106804561-106804561 on Build GRCh38
rs946823131 Chr.2: 106804567-106804567 on Build GRCh38
rs897318352 Chr.2: 106804434-106804434 on Build GRCh38
rs77630969 Chr.2: 106804834-106804834 on Build GRCh38
rs117213857 Chr.2: 106804860-106804860 on Build GRCh38
rs951852818 Chr.2: 106804451-106804451 on Build GRCh38
rs913995736 Chr.2: 106804566-106804566 on Build GRCh38
rs1010828622 Chr.2: 106804868-106804870 on Build GRCh38
rs979940330 Chr.2: 106804854-106804854 on Build GRCh38

View Full Product Details