Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 108253176-108253701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SULT1C3
Entrez Gene ID
442038
Gene Name
sulfotransferase family 1C member 3
Gene Aliases
ST1C3
UniGene
Gene Chromosome Location
Chr.2: 108239992-108265608 on Build GRCh38
Transcripts
NM_001008743.2 NM_001320878.1 XM_017004153.1 XM_017004154.1 XM_017004155.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5394091 c.306G>A p.L102L Chr.2: 108253349-108253349 on Build GRCh38
6899609 c.393C>T p.N131N Chr.2: 108253436-108253436 on Build GRCh38
3565051 c.312C>T p.F104F Chr.2: 108253355-108253355 on Build GRCh38
5579663 c.339G>A p.L113L Chr.2: 108253382-108253382 on Build GRCh38
5394092 c.306G>A p.L102L Chr.2: 108253349-108253349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776945370 Chr.2: 108253371-108253371 on Build GRCh38
rs1051544686 Chr.2: 108253657-108253657 on Build GRCh38
rs937232188 Chr.2: 108253256-108253256 on Build GRCh38
rs963568718 Chr.2: 108253648-108253648 on Build GRCh38
rs779684598 Chr.2: 108253345-108253345 on Build GRCh38
rs919534637 Chr.2: 108253404-108253404 on Build GRCh38
rs200479401 Chr.2: 108253664-108253664 on Build GRCh38
rs537737380 Chr.2: 108253499-108253499 on Build GRCh38
rs76202513 Chr.2: 108253483-108253483 on Build GRCh38
rs146458854 Chr.2: 108253368-108253368 on Build GRCh38
rs1036105351 Chr.2: 108253320-108253320 on Build GRCh38
rs1043412589 Chr.2: 108253286-108253286 on Build GRCh38
rs1007601212 Chr.2: 108253635-108253635 on Build GRCh38
rs888770953 Chr.2: 108253660-108253660 on Build GRCh38
rs746168728 Chr.2: 108253380-108253380 on Build GRCh38
rs753395625 Chr.2: 108253491-108253491 on Build GRCh38
rs762872514 Chr.2: 108253425-108253425 on Build GRCh38
rs191990502 Chr.2: 108253553-108253553 on Build GRCh38
rs933041733 Chr.2: 108253647-108253647 on Build GRCh38
rs1011511933 Chr.2: 108253269-108253269 on Build GRCh38
rs111976908 Chr.2: 108253466-108253466 on Build GRCh38
rs764160260 Chr.2: 108253427-108253427 on Build GRCh38
rs140329992 Chr.2: 108253349-108253349 on Build GRCh38
rs758959924 Chr.2: 108253352-108253352 on Build GRCh38
rs374294288 Chr.2: 108253343-108253343 on Build GRCh38
rs747393318 Chr.2: 108253356-108253356 on Build GRCh38
rs1007129495 Chr.2: 108253367-108253367 on Build GRCh38
rs867534693 Chr.2: 108253254-108253254 on Build GRCh38
rs973687148 Chr.2: 108253395-108253395 on Build GRCh38
rs941899960 Chr.2: 108253302-108253302 on Build GRCh38
rs1011789065 Chr.2: 108253381-108253381 on Build GRCh38
rs769893228 Chr.2: 108253394-108253394 on Build GRCh38
rs755868968 Chr.2: 108253268-108253268 on Build GRCh38
rs778210913 Chr.2: 108253355-108253355 on Build GRCh38
rs915751760 Chr.2: 108253527-108253527 on Build GRCh38
rs767663057 Chr.2: 108253315-108253318 on Build GRCh38
rs928304818 Chr.2: 108253675-108253675 on Build GRCh38
rs1017692197 Chr.2: 108253644-108253644 on Build GRCh38
rs903192949 Chr.2: 108253323-108253323 on Build GRCh38
rs555709504 Chr.2: 108253266-108253266 on Build GRCh38
rs769055137 Chr.2: 108253277-108253277 on Build GRCh38
rs774383331 Chr.2: 108253436-108253436 on Build GRCh38
rs903137942 Chr.2: 108253574-108253574 on Build GRCh38
rs113960100 Chr.2: 108253587-108253587 on Build GRCh38
rs775651945 Chr.2: 108253423-108253423 on Build GRCh38
rs1026551535 Chr.2: 108253663-108253663 on Build GRCh38
rs919105691 Chr.2: 108253643-108253643 on Build GRCh38
rs199728317 Chr.2: 108253437-108253437 on Build GRCh38
rs763878476 Chr.2: 108253304-108253304 on Build GRCh38
rs555930916 Chr.2: 108253634-108253634 on Build GRCh38
rs767300866 Chr.2: 108253325-108253325 on Build GRCh38
rs957516177 Chr.2: 108253373-108253373 on Build GRCh38
rs370377573 Chr.2: 108253490-108253490 on Build GRCh38
rs1051728287 Chr.2: 108253621-108253621 on Build GRCh38
rs751405800 Chr.2: 108253319-108253319 on Build GRCh38
rs200351879 Chr.2: 108253324-108253324 on Build GRCh38
rs760215064 Chr.2: 108253555-108253555 on Build GRCh38
rs982417926 Chr.2: 108253669-108253669 on Build GRCh38
rs184794678 Chr.2: 108253339-108253339 on Build GRCh38
rs767460393 Chr.2: 108253462-108253462 on Build GRCh38
rs951065394 Chr.2: 108253666-108253666 on Build GRCh38
rs995450590 Chr.2: 108253649-108253649 on Build GRCh38
rs200769198 Chr.2: 108253351-108253351 on Build GRCh38
rs545030165 Chr.2: 108253326-108253326 on Build GRCh38
rs941865750 Chr.2: 108253566-108253566 on Build GRCh38
rs928659059 Chr.2: 108253213-108253213 on Build GRCh38
rs998853757 Chr.2: 108253605-108253605 on Build GRCh38
rs754393488 Chr.2: 108253493-108253493 on Build GRCh38
rs753666259 Chr.2: 108253400-108253400 on Build GRCh38
rs1020452996 Chr.2: 108253392-108253392 on Build GRCh38

View Full Product Details