Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 120351227-120351752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INHBB
Entrez Gene ID
3625
Gene Name
inhibin beta B subunit
Gene Aliases
-
UniGene
Hs.1735
Gene Chromosome Location
Chr.2: 120346143-120351807 on Build GRCh38
Transcripts
NM_002193.2

SNP Detail

SNP ID Chromosome Location
rs1005963182 Chr.2: 120351605-120351605 on Build GRCh38
rs554651758 Chr.2: 120351257-120351257 on Build GRCh38
rs1024444419 Chr.2: 120351672-120351672 on Build GRCh38
rs878894026 Chr.2: 120351399-120351400 on Build GRCh38
rs887139290 Chr.2: 120351604-120351604 on Build GRCh38
rs958953832 Chr.2: 120351646-120351646 on Build GRCh38
rs541181608 Chr.2: 120351489-120351489 on Build GRCh38
rs934975223 Chr.2: 120351346-120351346 on Build GRCh38
rs778327864 Chr.2: 120351610-120351611 on Build GRCh38
rs986314877 Chr.2: 120351586-120351586 on Build GRCh38
rs769732473 Chr.2: 120351261-120351261 on Build GRCh38
rs923662234 Chr.2: 120351329-120351329 on Build GRCh38
rs1056074505 Chr.2: 120351392-120351392 on Build GRCh38
rs909361575 Chr.2: 120351633-120351633 on Build GRCh38
rs1023395538 Chr.2: 120351462-120351462 on Build GRCh38
rs1036387513 Chr.2: 120351313-120351313 on Build GRCh38
rs773182985 Chr.2: 120351315-120351315 on Build GRCh38
rs958902278 Chr.2: 120351253-120351253 on Build GRCh38
rs754472153 Chr.2: 120351375-120351376 on Build GRCh38
rs1053750398 Chr.2: 120351443-120351443 on Build GRCh38
rs949139990 Chr.2: 120351265-120351265 on Build GRCh38
rs548844896 Chr.2: 120351656-120351656 on Build GRCh38
rs919289452 Chr.2: 120351707-120351707 on Build GRCh38
rs1031114535 Chr.2: 120351514-120351514 on Build GRCh38
rs1050548429 Chr.2: 120351271-120351271 on Build GRCh38
rs138000535 Chr.2: 120351585-120351585 on Build GRCh38
rs142599342 Chr.2: 120351644-120351644 on Build GRCh38
rs71706683 Chr.2: 120351399-120351399 on Build GRCh38
rs967789861 Chr.2: 120351471-120351471 on Build GRCh38
rs909307497 Chr.2: 120351282-120351286 on Build GRCh38
rs80136509 Chr.2: 120351515-120351515 on Build GRCh38
rs555822834 Chr.2: 120351258-120351258 on Build GRCh38
rs994524490 Chr.2: 120351410-120351410 on Build GRCh38
rs770832312 Chr.2: 120351616-120351616 on Build GRCh38
rs533966708 Chr.2: 120351600-120351600 on Build GRCh38
rs1056381491 Chr.2: 120351416-120351416 on Build GRCh38
rs971940888 Chr.2: 120351694-120351694 on Build GRCh38
rs183806955 Chr.2: 120351647-120351647 on Build GRCh38
rs923554181 Chr.2: 120351561-120351561 on Build GRCh38
rs900983436 Chr.2: 120351545-120351545 on Build GRCh38
rs560960797 Chr.2: 120351710-120351710 on Build GRCh38
rs879173390 Chr.2: 120351398-120351399 on Build GRCh38
rs150764799 Chr.2: 120351577-120351577 on Build GRCh38
rs141367116 Chr.2: 120351487-120351487 on Build GRCh38
rs1017585421 Chr.2: 120351250-120351250 on Build GRCh38
rs942561057 Chr.2: 120351488-120351488 on Build GRCh38
rs942067697 Chr.2: 120351302-120351303 on Build GRCh38
rs76068660 Chr.2: 120351415-120351415 on Build GRCh38
rs1012044592 Chr.2: 120351447-120351447 on Build GRCh38

View Full Product Details