Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19177835-19178322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A1
Entrez Gene ID
6576
Gene Name
solute carrier family 25 member 1
Gene Aliases
CTP, D2L2AD, SEA, SLC20A3
UniGene
Hs.111024
Gene Chromosome Location
Chr.22: 19175575-19178863 on Build GRCh38
Transcripts
NM_001256534.1 NM_001287387.1 NM_005984.4 NR_046298.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3992017 c.179A>C p.H60P Chr.22: 19178156-19178156 on Build GRCh38
5892481 c.203G>A p.G68E Chr.22: 19178041-19178041 on Build GRCh38
1740767 c.156G>T p.Q52H Chr.22: 19178179-19178179 on Build GRCh38
5661595 c.324C>G p.L108L Chr.22: 19177844-19177844 on Build GRCh38
4388242 c.174C>T p.R58R Chr.22: 19178161-19178161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781828760 Chr.22: 19177871-19177871 on Build GRCh38
rs782424329 Chr.22: 19178257-19178257 on Build GRCh38
rs1055425347 Chr.22: 19177864-19177864 on Build GRCh38
rs1055622295 Chr.22: 19178127-19178127 on Build GRCh38
rs782771350 Chr.22: 19178111-19178111 on Build GRCh38
rs953111140 Chr.22: 19177977-19177977 on Build GRCh38
rs782460590 Chr.22: 19178141-19178141 on Build GRCh38
rs568890818 Chr.22: 19178008-19178008 on Build GRCh38
rs2070255 Chr.22: 19177965-19177965 on Build GRCh38
rs781936654 Chr.22: 19178273-19178273 on Build GRCh38
rs782748338 Chr.22: 19177933-19177933 on Build GRCh38
rs782414775 Chr.22: 19177896-19177896 on Build GRCh38
rs782120232 Chr.22: 19178030-19178030 on Build GRCh38
rs369464760 Chr.22: 19178292-19178292 on Build GRCh38
rs566540393 Chr.22: 19178298-19178298 on Build GRCh38
rs782346548 Chr.22: 19178026-19178026 on Build GRCh38
rs782281296 Chr.22: 19178253-19178253 on Build GRCh38
rs782189539 Chr.22: 19178130-19178130 on Build GRCh38
rs782085953 Chr.22: 19177923-19177923 on Build GRCh38
rs949741445 Chr.22: 19178162-19178162 on Build GRCh38
rs782492046 Chr.22: 19177872-19177872 on Build GRCh38
rs782188868 Chr.22: 19177902-19177902 on Build GRCh38
rs782224788 Chr.22: 19177986-19177986 on Build GRCh38
rs781981461 Chr.22: 19178300-19178300 on Build GRCh38
rs782232451 Chr.22: 19178169-19178169 on Build GRCh38
rs371619113 Chr.22: 19177992-19177992 on Build GRCh38
rs781818709 Chr.22: 19178072-19178072 on Build GRCh38
rs781924664 Chr.22: 19178017-19178017 on Build GRCh38
rs782638209 Chr.22: 19177873-19177873 on Build GRCh38
rs782598183 Chr.22: 19178155-19178155 on Build GRCh38
rs897579947 Chr.22: 19178274-19178274 on Build GRCh38
rs782272401 Chr.22: 19178007-19178007 on Build GRCh38
rs984371316 Chr.22: 19177984-19177984 on Build GRCh38
rs782677335 Chr.22: 19178118-19178118 on Build GRCh38
rs781808930 Chr.22: 19177922-19177922 on Build GRCh38
rs782008540 Chr.22: 19177909-19177909 on Build GRCh38
rs367788882 Chr.22: 19177971-19177971 on Build GRCh38
rs782112811 Chr.22: 19177908-19177908 on Build GRCh38
rs782590548 Chr.22: 19177860-19177860 on Build GRCh38
rs782477102 Chr.22: 19178104-19178104 on Build GRCh38
rs1013989679 Chr.22: 19177904-19177904 on Build GRCh38
rs782579935 Chr.22: 19177898-19177898 on Build GRCh38
rs1005836255 Chr.22: 19178299-19178299 on Build GRCh38
rs781859466 Chr.22: 19177950-19177950 on Build GRCh38
rs782258869 Chr.22: 19177890-19177890 on Build GRCh38
rs782021001 Chr.22: 19178051-19178051 on Build GRCh38
rs967454635 Chr.22: 19178108-19178108 on Build GRCh38
rs782209746 Chr.22: 19177907-19177907 on Build GRCh38
rs782457823 Chr.22: 19177856-19177856 on Build GRCh38
rs782004863 Chr.22: 19178252-19178252 on Build GRCh38
rs925803238 Chr.22: 19178114-19178114 on Build GRCh38
rs994748256 Chr.22: 19177928-19177928 on Build GRCh38
rs782589426 Chr.22: 19177970-19177970 on Build GRCh38
rs888598092 Chr.22: 19178297-19178297 on Build GRCh38
rs782627397 Chr.22: 19177906-19177907 on Build GRCh38
rs56386322 Chr.22: 19177967-19177967 on Build GRCh38
rs782465506 Chr.22: 19177969-19177969 on Build GRCh38
rs534355939 Chr.22: 19178285-19178285 on Build GRCh38
rs782371152 Chr.22: 19178178-19178178 on Build GRCh38
rs387907477 Chr.22: 19178197-19178197 on Build GRCh38
rs782646725 Chr.22: 19178005-19178005 on Build GRCh38
rs387907476 Chr.22: 19177958-19177958 on Build GRCh38
rs57827877 Chr.22: 19178116-19178116 on Build GRCh38
rs371134887 Chr.22: 19178022-19178022 on Build GRCh38
rs782167390 Chr.22: 19177911-19177911 on Build GRCh38
rs922680720 Chr.22: 19178100-19178100 on Build GRCh38
rs781789414 Chr.22: 19177968-19177968 on Build GRCh38
rs782781234 Chr.22: 19178050-19178050 on Build GRCh38

View Full Product Details