Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19359245-19359746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIRA
Entrez Gene ID
7290
Gene Name
histone cell cycle regulator
Gene Aliases
DGCR1, TUP1, TUPLE1
UniGene
Hs.474206
Gene Chromosome Location
Chr.22: 19330701-19431696 on Build GRCh38
Transcripts
NM_003325.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4690381 c.2144delG p.G715fs*2 Chr.22: 19359426-19359426 on Build GRCh38
3552164 c.2214C>T p.I738I Chr.22: 19359356-19359356 on Build GRCh38
4569870 c.2111T>C p.I704T Chr.22: 19359459-19359459 on Build GRCh38
3552165 c.2157C>T p.S719S Chr.22: 19359413-19359413 on Build GRCh38
1743496 c.2210G>A p.R737Q Chr.22: 19359360-19359360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752712842 Chr.22: 19359294-19359294 on Build GRCh38
rs9605996 Chr.22: 19359312-19359312 on Build GRCh38
rs143152290 Chr.22: 19359361-19359361 on Build GRCh38
rs910859106 Chr.22: 19359475-19359475 on Build GRCh38
rs147482921 Chr.22: 19359411-19359411 on Build GRCh38
rs922863316 Chr.22: 19359616-19359616 on Build GRCh38
rs139903076 Chr.22: 19359424-19359424 on Build GRCh38
rs559082146 Chr.22: 19359525-19359525 on Build GRCh38
rs781144969 Chr.22: 19359536-19359536 on Build GRCh38
rs201411183 Chr.22: 19359412-19359412 on Build GRCh38
rs758362224 Chr.22: 19359365-19359365 on Build GRCh38
rs765623617 Chr.22: 19359525-19359525 on Build GRCh38
rs932832957 Chr.22: 19359390-19359390 on Build GRCh38
rs1032860733 Chr.22: 19359565-19359565 on Build GRCh38
rs1039246628 Chr.22: 19359506-19359506 on Build GRCh38
rs767900239 Chr.22: 19359353-19359353 on Build GRCh38
rs182607148 Chr.22: 19359510-19359510 on Build GRCh38
rs373602444 Chr.22: 19359338-19359338 on Build GRCh38
rs774976171 Chr.22: 19359414-19359414 on Build GRCh38
rs902589204 Chr.22: 19359547-19359547 on Build GRCh38
rs530466334 Chr.22: 19359330-19359330 on Build GRCh38
rs143821957 Chr.22: 19359431-19359431 on Build GRCh38
rs760188458 Chr.22: 19359496-19359496 on Build GRCh38
rs760967846 Chr.22: 19359426-19359426 on Build GRCh38
rs545334184 Chr.22: 19359274-19359274 on Build GRCh38
rs764073070 Chr.22: 19359465-19359465 on Build GRCh38
rs749834610 Chr.22: 19359482-19359482 on Build GRCh38
rs751469729 Chr.22: 19359466-19359466 on Build GRCh38
rs754989808 Chr.22: 19359515-19359515 on Build GRCh38
rs755858451 Chr.22: 19359496-19359496 on Build GRCh38
rs1024840531 Chr.22: 19359632-19359632 on Build GRCh38
rs755363675 Chr.22: 19359505-19359505 on Build GRCh38
rs571273724 Chr.22: 19359613-19359613 on Build GRCh38
rs376780591 Chr.22: 19359358-19359358 on Build GRCh38
rs976726906 Chr.22: 19359607-19359607 on Build GRCh38
rs373162024 Chr.22: 19359530-19359530 on Build GRCh38
rs748686457 Chr.22: 19359521-19359521 on Build GRCh38
rs774636516 Chr.22: 19359345-19359345 on Build GRCh38
rs367959199 Chr.22: 19359402-19359402 on Build GRCh38
rs150237834 Chr.22: 19359341-19359341 on Build GRCh38
rs993413150 Chr.22: 19359307-19359307 on Build GRCh38
rs773794107 Chr.22: 19359503-19359503 on Build GRCh38
rs754278280 Chr.22: 19359504-19359504 on Build GRCh38
rs531005255 Chr.22: 19359281-19359281 on Build GRCh38
rs773534968 Chr.22: 19359356-19359356 on Build GRCh38
rs780865485 Chr.22: 19359396-19359396 on Build GRCh38
rs762646917 Chr.22: 19359452-19359452 on Build GRCh38
rs993552326 Chr.22: 19359290-19359290 on Build GRCh38
rs373189208 Chr.22: 19359524-19359524 on Build GRCh38
rs777212260 Chr.22: 19359425-19359426 on Build GRCh38
rs747947367 Chr.22: 19359310-19359310 on Build GRCh38
rs1012670280 Chr.22: 19359617-19359617 on Build GRCh38
rs762479563 Chr.22: 19359540-19359540 on Build GRCh38
rs756740115 Chr.22: 19359375-19359375 on Build GRCh38
rs567129767 Chr.22: 19359397-19359397 on Build GRCh38
rs746744878 Chr.22: 19359395-19359395 on Build GRCh38
rs773379388 Chr.22: 19359425-19359425 on Build GRCh38
rs761702236 Chr.22: 19359470-19359470 on Build GRCh38
rs761969691 Chr.22: 19359346-19359346 on Build GRCh38
rs370595702 Chr.22: 19359325-19359325 on Build GRCh38
rs372342926 Chr.22: 19359648-19359648 on Build GRCh38
rs941514826 Chr.22: 19359670-19359670 on Build GRCh38
rs752377093 Chr.22: 19359362-19359362 on Build GRCh38
rs376450821 Chr.22: 19359298-19359298 on Build GRCh38
rs114909614 Chr.22: 19359374-19359374 on Build GRCh38
rs1021016684 Chr.22: 19359555-19359555 on Build GRCh38
rs764018146 Chr.22: 19359367-19359367 on Build GRCh38
rs765125977 Chr.22: 19359289-19359289 on Build GRCh38
rs1046299989 Chr.22: 19359438-19359438 on Build GRCh38
rs970658512 Chr.22: 19359642-19359642 on Build GRCh38
rs148683985 Chr.22: 19359476-19359476 on Build GRCh38
rs1002459398 Chr.22: 19359471-19359471 on Build GRCh38
rs1000930633 Chr.22: 19359549-19359549 on Build GRCh38
rs972521262 Chr.22: 19359646-19359646 on Build GRCh38
rs201236759 Chr.22: 19359311-19359311 on Build GRCh38
rs766444068 Chr.22: 19359429-19359429 on Build GRCh38
rs369195565 Chr.22: 19359490-19359490 on Build GRCh38
rs777391169 Chr.22: 19359376-19359376 on Build GRCh38
rs138814028 Chr.22: 19359344-19359344 on Build GRCh38
rs746283651 Chr.22: 19359326-19359326 on Build GRCh38

View Full Product Details