Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 3148139-3148642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRBN
Entrez Gene ID
51185
Gene Name
cereblon
Gene Aliases
MRT2, MRT2A
UniGene
Hs.18925
Gene Chromosome Location
Chr.3: 3148490-3179717 on Build GRCh38
Transcripts
XM_011533791.2 NM_001302946.1 NM_182916.2 XM_011533776.2 XM_011533777.2 XM_011533778.2

Gene Symbol
TRNT1
Entrez Gene ID
51095
Gene Name
tRNA nucleotidyl transferase 1
Gene Aliases
CCA1, CGI-47, MtCCA, RPEM, SIFD
UniGene
Hs.732725
Gene Chromosome Location
Chr.3: 3126916-3153435 on Build GRCh38
Transcripts
XM_011533791.2 NM_001302946.1 NM_182916.2 XM_011533776.2 XM_011533777.2 XM_011533778.2

SNP Detail

SNP ID Chromosome Location
rs910961277 Chr.3: 3148545-3148545 on Build GRCh38
rs1042070445 Chr.3: 3148253-3148253 on Build GRCh38
rs768444581 Chr.3: 3148608-3148609 on Build GRCh38
rs1025255317 Chr.3: 3148425-3148425 on Build GRCh38
rs1050689364 Chr.3: 3148266-3148266 on Build GRCh38
rs185263036 Chr.3: 3148257-3148257 on Build GRCh38
rs903762642 Chr.3: 3148424-3148424 on Build GRCh38
rs933707003 Chr.3: 3148303-3148303 on Build GRCh38
rs889347316 Chr.3: 3148505-3148505 on Build GRCh38
rs754465194 Chr.3: 3148179-3148179 on Build GRCh38
rs758588985 Chr.3: 3148473-3148473 on Build GRCh38
rs900877980 Chr.3: 3148317-3148317 on Build GRCh38
rs567907193 Chr.3: 3148569-3148571 on Build GRCh38
rs983985871 Chr.3: 3148434-3148434 on Build GRCh38
rs571155174 Chr.3: 3148283-3148283 on Build GRCh38
rs567413351 Chr.3: 3148365-3148365 on Build GRCh38
rs757175462 Chr.3: 3148575-3148575 on Build GRCh38
rs867277541 Chr.3: 3148519-3148519 on Build GRCh38
rs777531278 Chr.3: 3148201-3148201 on Build GRCh38
rs534795311 Chr.3: 3148214-3148214 on Build GRCh38
rs925924682 Chr.3: 3148475-3148475 on Build GRCh38
rs762327777 Chr.3: 3148175-3148175 on Build GRCh38
rs1019127613 Chr.3: 3148315-3148315 on Build GRCh38
rs558685323 Chr.3: 3148463-3148463 on Build GRCh38
rs984034956 Chr.3: 3148174-3148174 on Build GRCh38
rs184877192 Chr.3: 3148503-3148503 on Build GRCh38
rs889273579 Chr.3: 3148274-3148274 on Build GRCh38
rs767061019 Chr.3: 3148180-3148180 on Build GRCh38
rs1053055402 Chr.3: 3148580-3148580 on Build GRCh38
rs140435051 Chr.3: 3148255-3148255 on Build GRCh38
rs946171529 Chr.3: 3148219-3148219 on Build GRCh38
rs749293620 Chr.3: 3148567-3148567 on Build GRCh38
rs963267502 Chr.3: 3148500-3148500 on Build GRCh38
rs937200900 Chr.3: 3148205-3148205 on Build GRCh38
rs139714355 Chr.3: 3148564-3148564 on Build GRCh38
rs867162998 Chr.3: 3148301-3148301 on Build GRCh38
rs192546200 Chr.3: 3148450-3148450 on Build GRCh38
rs760280702 Chr.3: 3148190-3148190 on Build GRCh38
rs1050930938 Chr.3: 3148314-3148315 on Build GRCh38
rs370590725 Chr.3: 3148238-3148238 on Build GRCh38
rs888101193 Chr.3: 3148566-3148566 on Build GRCh38
rs1014258032 Chr.3: 3148440-3148440 on Build GRCh38
rs762404826 Chr.3: 3148260-3148260 on Build GRCh38
rs77554992 Chr.3: 3148403-3148403 on Build GRCh38
rs78363013 Chr.3: 3148522-3148522 on Build GRCh38
rs755830386 Chr.3: 3148188-3148188 on Build GRCh38
rs911025605 Chr.3: 3148328-3148328 on Build GRCh38
rs1033997062 Chr.3: 3148568-3148568 on Build GRCh38
rs371830913 Chr.3: 3148187-3148187 on Build GRCh38
rs750776479 Chr.3: 3148350-3148350 on Build GRCh38
rs115688744 Chr.3: 3148206-3148206 on Build GRCh38
rs1043937525 Chr.3: 3148561-3148562 on Build GRCh38
rs79659648 Chr.3: 3148594-3148594 on Build GRCh38
rs765753900 Chr.3: 3148268-3148268 on Build GRCh38
rs946090931 Chr.3: 3148469-3148469 on Build GRCh38
rs147416562 Chr.3: 3148488-3148488 on Build GRCh38
rs532366787 Chr.3: 3148561-3148561 on Build GRCh38
rs776921520 Chr.3: 3148166-3148166 on Build GRCh38
rs1024429722 Chr.3: 3148443-3148444 on Build GRCh38
rs544076317 Chr.3: 3148534-3148534 on Build GRCh38
rs552220409 Chr.3: 3148264-3148264 on Build GRCh38
rs538956352 Chr.3: 3148406-3148407 on Build GRCh38
rs369264199 Chr.3: 3148441-3148441 on Build GRCh38
rs533692358 Chr.3: 3148300-3148300 on Build GRCh38
rs187353748 Chr.3: 3148439-3148439 on Build GRCh38
rs952422393 Chr.3: 3148433-3148433 on Build GRCh38
rs1000230104 Chr.3: 3148434-3148434 on Build GRCh38
rs538780074 Chr.3: 3148455-3148455 on Build GRCh38
rs12107469 Chr.3: 3148556-3148556 on Build GRCh38
rs751043046 Chr.3: 3148177-3148177 on Build GRCh38
rs561645433 Chr.3: 3148507-3148507 on Build GRCh38
rs745867248 Chr.3: 3148495-3148496 on Build GRCh38
rs958631220 Chr.3: 3148454-3148454 on Build GRCh38
rs767699629 Chr.3: 3148208-3148208 on Build GRCh38
rs547678203 Chr.3: 3148364-3148364 on Build GRCh38
rs531240749 Chr.3: 3148272-3148274 on Build GRCh38
rs780588029 Chr.3: 3148169-3148169 on Build GRCh38

View Full Product Details