Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196734156-196734667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIGX
Entrez Gene ID
54965
Gene Name
phosphatidylinositol glycan anchor biosynthesis class X
Gene Aliases
PIG-X
UniGene
Hs.223296
Gene Chromosome Location
Chr.3: 196712374-196736007 on Build GRCh38
Transcripts
NM_001166304.1 NM_017861.3

SNP Detail

SNP ID Chromosome Location
rs1039848671 Chr.3: 196734522-196734522 on Build GRCh38
rs1033208907 Chr.3: 196734494-196734494 on Build GRCh38
rs114586754 Chr.3: 196734422-196734422 on Build GRCh38
rs899159373 Chr.3: 196734204-196734204 on Build GRCh38
rs1051296028 Chr.3: 196734297-196734297 on Build GRCh38
rs999942555 Chr.3: 196734491-196734491 on Build GRCh38
rs542165519 Chr.3: 196734268-196734268 on Build GRCh38
rs971337533 Chr.3: 196734361-196734361 on Build GRCh38
rs533145290 Chr.3: 196734529-196734529 on Build GRCh38
rs6805472 Chr.3: 196734298-196734298 on Build GRCh38
rs985368498 Chr.3: 196734549-196734549 on Build GRCh38
rs929640166 Chr.3: 196734477-196734477 on Build GRCh38
rs962470527 Chr.3: 196734408-196734408 on Build GRCh38
rs918227809 Chr.3: 196734445-196734445 on Build GRCh38
rs3850646 Chr.3: 196734531-196734531 on Build GRCh38
rs892477862 Chr.3: 196734338-196734338 on Build GRCh38
rs1010118823 Chr.3: 196734340-196734340 on Build GRCh38
rs761738924 Chr.3: 196734306-196734306 on Build GRCh38
rs1047113 Chr.3: 196734603-196734603 on Build GRCh38
rs750374402 Chr.3: 196734518-196734518 on Build GRCh38
rs386670222 Chr.3: 196734529-196734531 on Build GRCh38
rs912800783 Chr.3: 196734502-196734502 on Build GRCh38
rs1030971707 Chr.3: 196734336-196734336 on Build GRCh38
rs369020423 Chr.3: 196734542-196734542 on Build GRCh38
rs1047120 Chr.3: 196734310-196734310 on Build GRCh38
rs547270954 Chr.3: 196734469-196734469 on Build GRCh38
rs3177527 Chr.3: 196734363-196734363 on Build GRCh38
rs907102570 Chr.3: 196734554-196734554 on Build GRCh38
rs369227756 Chr.3: 196734645-196734645 on Build GRCh38
rs986461071 Chr.3: 196734495-196734495 on Build GRCh38
rs4916542 Chr.3: 196734519-196734519 on Build GRCh38
rs751758649 Chr.3: 196734541-196734541 on Build GRCh38
rs1023123943 Chr.3: 196734368-196734368 on Build GRCh38
rs929423025 Chr.3: 196734629-196734629 on Build GRCh38
rs376527693 Chr.3: 196734620-196734620 on Build GRCh38
rs369302951 Chr.3: 196734359-196734359 on Build GRCh38
rs1034622746 Chr.3: 196734386-196734386 on Build GRCh38
rs112053558 Chr.3: 196734557-196734557 on Build GRCh38
rs372124725 Chr.3: 196734284-196734284 on Build GRCh38
rs115030651 Chr.3: 196734596-196734596 on Build GRCh38
rs1052951180 Chr.3: 196734532-196734532 on Build GRCh38
rs988843779 Chr.3: 196734520-196734520 on Build GRCh38
rs1001376340 Chr.3: 196734307-196734307 on Build GRCh38
rs765221478 Chr.3: 196734346-196734346 on Build GRCh38
rs1014525767 Chr.3: 196734367-196734367 on Build GRCh38
rs1040302252 Chr.3: 196734303-196734303 on Build GRCh38
rs1025009831 Chr.3: 196734550-196734550 on Build GRCh38
rs942862776 Chr.3: 196734300-196734300 on Build GRCh38

View Full Product Details