Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73074661-73075203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD17
Entrez Gene ID
26057
Gene Name
ankyrin repeat domain 17
Gene Aliases
GTAR, MASK2, NY-BR-16
UniGene
Hs.601206
Gene Chromosome Location
Chr.4: 73073966-73259142 on Build GRCh38
Transcripts
NM_001286771.2 NM_015574.1 NM_032217.4 NM_198889.2 XM_005265671.3 XM_017008011.1 XM_017008012.1 XM_017008013.1

SNP Detail

SNP ID Chromosome Location
rs935024062 Chr.4: 73074994-73074994 on Build GRCh38
rs1027028254 Chr.4: 73075169-73075169 on Build GRCh38
rs989079707 Chr.4: 73074949-73074949 on Build GRCh38
rs965804492 Chr.4: 73074839-73074839 on Build GRCh38
rs533176931 Chr.4: 73074685-73074685 on Build GRCh38
rs774304951 Chr.4: 73074916-73074916 on Build GRCh38
rs970214001 Chr.4: 73074816-73074816 on Build GRCh38
rs900297137 Chr.4: 73074692-73074692 on Build GRCh38
rs189596553 Chr.4: 73075011-73075011 on Build GRCh38
rs887002853 Chr.4: 73075168-73075168 on Build GRCh38
rs573690401 Chr.4: 73074834-73074834 on Build GRCh38
rs184726926 Chr.4: 73074796-73074796 on Build GRCh38
rs143788294 Chr.4: 73074760-73074761 on Build GRCh38
rs577746791 Chr.4: 73074727-73074728 on Build GRCh38
rs995575981 Chr.4: 73074790-73074790 on Build GRCh38
rs549195822 Chr.4: 73074831-73074831 on Build GRCh38
rs897121009 Chr.4: 73075090-73075090 on Build GRCh38
rs78397762 Chr.4: 73074761-73074761 on Build GRCh38
rs553398049 Chr.4: 73074983-73074983 on Build GRCh38
rs1015531096 Chr.4: 73074761-73074761 on Build GRCh38
rs559413818 Chr.4: 73075058-73075058 on Build GRCh38
rs762936674 Chr.4: 73074869-73074869 on Build GRCh38
rs573084338 Chr.4: 73075133-73075133 on Build GRCh38
rs995328134 Chr.4: 73075056-73075056 on Build GRCh38
rs890312049 Chr.4: 73075038-73075038 on Build GRCh38
rs567545122 Chr.4: 73074882-73074882 on Build GRCh38
rs551850436 Chr.4: 73074708-73074708 on Build GRCh38
rs571173502 Chr.4: 73074985-73074985 on Build GRCh38
rs575443948 Chr.4: 73075044-73075044 on Build GRCh38
rs949079396 Chr.4: 73075025-73075025 on Build GRCh38
rs951443386 Chr.4: 73074751-73074751 on Build GRCh38
rs1052505242 Chr.4: 73075008-73075008 on Build GRCh38
rs557657399 Chr.4: 73074715-73074718 on Build GRCh38
rs11727957 Chr.4: 73074748-73074748 on Build GRCh38
rs542821142 Chr.4: 73075083-73075083 on Build GRCh38
rs995917878 Chr.4: 73075165-73075165 on Build GRCh38
rs775857243 Chr.4: 73075030-73075030 on Build GRCh38
rs983161214 Chr.4: 73074769-73074769 on Build GRCh38
rs961695575 Chr.4: 73074775-73074775 on Build GRCh38
rs1019715138 Chr.4: 73074772-73074772 on Build GRCh38
rs57949038 Chr.4: 73074769-73074770 on Build GRCh38
rs112403152 Chr.4: 73074918-73074918 on Build GRCh38
rs755872757 Chr.4: 73074833-73074833 on Build GRCh38
rs10092 Chr.4: 73075026-73075026 on Build GRCh38
rs554776318 Chr.4: 73075126-73075126 on Build GRCh38
rs921674594 Chr.4: 73075019-73075019 on Build GRCh38
rs980266234 Chr.4: 73074864-73074864 on Build GRCh38

View Full Product Details