Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76023267-76023790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ART3
Entrez Gene ID
419
Gene Name
ADP-ribosyltransferase 3
Gene Aliases
ARTC3
UniGene
Hs.24976
Gene Chromosome Location
Chr.4: 76011180-76112802 on Build GRCh38
Transcripts
NM_001130017.2 NM_001565.3

Gene Symbol
CXCL10
Entrez Gene ID
3627
Gene Name
C-X-C motif chemokine ligand 10
Gene Aliases
C7, IFI10, INP10, IP-10, SCYB10, crg-2, gIP-10, mob-1
UniGene
Hs.632586
Gene Chromosome Location
Chr.4: 76021116-76023536 on Build GRCh38
Transcripts
NM_001130017.2 NM_001565.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7130634 c.19C>G p.L7V Chr.4: 76023413-76023413 on Build GRCh38
4554396 c.61G>A p.G21R Chr.4: 76023371-76023371 on Build GRCh38
1057619 c.22A>G p.I8V Chr.4: 76023410-76023410 on Build GRCh38
6477357 c.51T>C p.S17S Chr.4: 76023381-76023381 on Build GRCh38
4616907 c.23T>C p.I8T Chr.4: 76023409-76023409 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904019947 Chr.4: 76023619-76023619 on Build GRCh38
rs375688092 Chr.4: 76023447-76023447 on Build GRCh38
rs767339215 Chr.4: 76023463-76023463 on Build GRCh38
rs868338912 Chr.4: 76023521-76023521 on Build GRCh38
rs577363299 Chr.4: 76023636-76023636 on Build GRCh38
rs1008653464 Chr.4: 76023684-76023685 on Build GRCh38
rs759245646 Chr.4: 76023321-76023321 on Build GRCh38
rs544624430 Chr.4: 76023646-76023646 on Build GRCh38
rs945672609 Chr.4: 76023315-76023315 on Build GRCh38
rs918169810 Chr.4: 76023558-76023558 on Build GRCh38
rs761474224 Chr.4: 76023346-76023346 on Build GRCh38
rs997887779 Chr.4: 76023607-76023607 on Build GRCh38
rs377480085 Chr.4: 76023451-76023451 on Build GRCh38
rs1126856 Chr.4: 76023417-76023417 on Build GRCh38
rs556589797 Chr.4: 76023653-76023653 on Build GRCh38
rs761786572 Chr.4: 76023462-76023462 on Build GRCh38
rs534096405 Chr.4: 76023504-76023504 on Build GRCh38
rs1000929937 Chr.4: 76023629-76023629 on Build GRCh38
rs769473524 Chr.4: 76023324-76023324 on Build GRCh38
rs1028356344 Chr.4: 76023302-76023302 on Build GRCh38
rs926332248 Chr.4: 76023483-76023483 on Build GRCh38
rs143493954 Chr.4: 76023379-76023379 on Build GRCh38
rs370870249 Chr.4: 76023445-76023445 on Build GRCh38
rs971005020 Chr.4: 76023709-76023709 on Build GRCh38
rs201821887 Chr.4: 76023387-76023387 on Build GRCh38
rs549320464 Chr.4: 76023334-76023334 on Build GRCh38
rs887638042 Chr.4: 76023402-76023402 on Build GRCh38
rs373483732 Chr.4: 76023335-76023335 on Build GRCh38
rs981508462 Chr.4: 76023724-76023724 on Build GRCh38
rs781668820 Chr.4: 76023423-76023423 on Build GRCh38
rs951840690 Chr.4: 76023735-76023735 on Build GRCh38
rs569490353 Chr.4: 76023534-76023534 on Build GRCh38
rs146711269 Chr.4: 76023753-76023753 on Build GRCh38
rs779028513 Chr.4: 76023384-76023384 on Build GRCh38
rs958167140 Chr.4: 76023690-76023690 on Build GRCh38
rs776021765 Chr.4: 76023761-76023761 on Build GRCh38
rs1006029121 Chr.4: 76023505-76023505 on Build GRCh38
rs914120425 Chr.4: 76023703-76023703 on Build GRCh38
rs766644940 Chr.4: 76023352-76023352 on Build GRCh38
rs745586920 Chr.4: 76023430-76023430 on Build GRCh38
rs574834526 Chr.4: 76023696-76023696 on Build GRCh38
rs769557687 Chr.4: 76023431-76023431 on Build GRCh38
rs568275340 Chr.4: 76023306-76023306 on Build GRCh38
rs377744455 Chr.4: 76023396-76023396 on Build GRCh38
rs897755462 Chr.4: 76023571-76023571 on Build GRCh38
rs752756202 Chr.4: 76023386-76023386 on Build GRCh38
rs771253146 Chr.4: 76023419-76023419 on Build GRCh38
rs4241578 Chr.4: 76023338-76023338 on Build GRCh38
rs56061981 Chr.4: 76023632-76023632 on Build GRCh38
rs765381166 Chr.4: 76023478-76023478 on Build GRCh38
rs748852335 Chr.4: 76023322-76023322 on Build GRCh38
rs1032493588 Chr.4: 76023665-76023665 on Build GRCh38
rs1026180652 Chr.4: 76023718-76023718 on Build GRCh38
rs755160508 Chr.4: 76023383-76023383 on Build GRCh38
rs1042570404 Chr.4: 76023594-76023594 on Build GRCh38
rs554892214 Chr.4: 76023466-76023466 on Build GRCh38
rs751375839 Chr.4: 76023341-76023341 on Build GRCh38
rs759807549 Chr.4: 76023476-76023476 on Build GRCh38
rs770530840 Chr.4: 76023343-76023343 on Build GRCh38
rs567039545 Chr.4: 76023480-76023480 on Build GRCh38
rs770640096 Chr.4: 76023446-76023446 on Build GRCh38
rs1019573925 Chr.4: 76023685-76023685 on Build GRCh38
rs778072445 Chr.4: 76023392-76023392 on Build GRCh38

View Full Product Details