Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 77056168-77056590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNI
Entrez Gene ID
10983
Gene Name
cyclin I
Gene Aliases
CCNI1, CYC1, CYI
UniGene
Hs.518827
Gene Chromosome Location
Chr.4: 77047155-77076013 on Build GRCh38
Transcripts
NM_006835.2 XM_011531555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5770832 c.276G>A p.E92E Chr.4: 77056249-77056249 on Build GRCh38
6168025 c.202-2A>T p.? Chr.4: 77056325-77056325 on Build GRCh38
5717200 c.256T>A p.Y86N Chr.4: 77056311-77056311 on Build GRCh38
6856803 c.245_246insT p.L83fs*7 Chr.4: 77056279-77056280 on Build GRCh38
5826287 c.203C>T p.A68V Chr.4: 77056322-77056322 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748405465 Chr.4: 77056508-77056508 on Build GRCh38
rs772332215 Chr.4: 77056328-77056328 on Build GRCh38
rs748705549 Chr.4: 77056226-77056228 on Build GRCh38
rs773413143 Chr.4: 77056355-77056356 on Build GRCh38
rs4252896 Chr.4: 77056373-77056373 on Build GRCh38
rs770920292 Chr.4: 77056346-77056346 on Build GRCh38
rs555748209 Chr.4: 77056548-77056548 on Build GRCh38
rs371031927 Chr.4: 77056315-77056315 on Build GRCh38
rs887020942 Chr.4: 77056440-77056440 on Build GRCh38
rs771315602 Chr.4: 77056357-77056384 on Build GRCh38
rs747216076 Chr.4: 77056345-77056345 on Build GRCh38
rs1031179029 Chr.4: 77056552-77056552 on Build GRCh38
rs749053668 Chr.4: 77056327-77056327 on Build GRCh38
rs982274384 Chr.4: 77056539-77056539 on Build GRCh38
rs138861603 Chr.4: 77056557-77056557 on Build GRCh38
rs745802676 Chr.4: 77056309-77056309 on Build GRCh38
rs904527552 Chr.4: 77056338-77056338 on Build GRCh38
rs145127291 Chr.4: 77056358-77056358 on Build GRCh38
rs4252895 Chr.4: 77056409-77056409 on Build GRCh38
rs770137157 Chr.4: 77056324-77056350 on Build GRCh38
rs75657784 Chr.4: 77056414-77056414 on Build GRCh38
rs777994116 Chr.4: 77056337-77056337 on Build GRCh38
rs965126387 Chr.4: 77056564-77056564 on Build GRCh38
rs4252892 Chr.4: 77056459-77056459 on Build GRCh38
rs758162996 Chr.4: 77056223-77056223 on Build GRCh38
rs1012647533 Chr.4: 77056503-77056503 on Build GRCh38
rs779890239 Chr.4: 77056380-77056380 on Build GRCh38
rs921417642 Chr.4: 77056207-77056207 on Build GRCh38
rs570241925 Chr.4: 77056512-77056512 on Build GRCh38
rs1024843228 Chr.4: 77056411-77056411 on Build GRCh38
rs878880002 Chr.4: 77056403-77056403 on Build GRCh38
rs1024042261 Chr.4: 77056517-77056517 on Build GRCh38
rs4252897 Chr.4: 77056238-77056238 on Build GRCh38
rs778943762 Chr.4: 77056423-77056423 on Build GRCh38
rs971170314 Chr.4: 77056539-77056543 on Build GRCh38
rs780682687 Chr.4: 77056303-77056303 on Build GRCh38
rs776045357 Chr.4: 77056374-77056374 on Build GRCh38
rs1005468778 Chr.4: 77056453-77056454 on Build GRCh38
rs781383785 Chr.4: 77056497-77056497 on Build GRCh38
rs191810699 Chr.4: 77056540-77056540 on Build GRCh38
rs866638318 Chr.4: 77056310-77056310 on Build GRCh38
rs915255319 Chr.4: 77056382-77056382 on Build GRCh38
rs779895732 Chr.4: 77056326-77056326 on Build GRCh38
rs751205219 Chr.4: 77056234-77056234 on Build GRCh38
rs4252894 Chr.4: 77056418-77056418 on Build GRCh38
rs201092460 Chr.4: 77056216-77056216 on Build GRCh38
rs765576898 Chr.4: 77056217-77056217 on Build GRCh38
rs4252893 Chr.4: 77056426-77056426 on Build GRCh38
rs749301452 Chr.4: 77056356-77056362 on Build GRCh38
rs753106390 Chr.4: 77056218-77056218 on Build GRCh38
rs4252898 Chr.4: 77056203-77056203 on Build GRCh38
rs942752092 Chr.4: 77056406-77056406 on Build GRCh38
rs760027831 Chr.4: 77056359-77056359 on Build GRCh38
rs895881979 Chr.4: 77056401-77056401 on Build GRCh38
rs373289448 Chr.4: 77056227-77056227 on Build GRCh38

View Full Product Details