Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 170718639-170719040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNIP1
Entrez Gene ID
30820
Gene Name
potassium voltage-gated channel interacting protein 1
Gene Aliases
KCHIP1, VABP
UniGene
Hs.484111
Gene Chromosome Location
Chr.5: 170353055-170736632 on Build GRCh38
Transcripts
NM_001034837.2 NM_001034838.2 NM_001278339.1 NM_001278340.1 NM_014592.3 XM_017009407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5546386 c.132G>A p.R44R Chr.5: 170718801-170718801 on Build GRCh38
108667 c.177C>T p.F59F Chr.5: 170718840-170718840 on Build GRCh38
120929 c.184A>T p.R62W Chr.5: 170718847-170718847 on Build GRCh38
3715171 c.195T>G p.L65L Chr.5: 170718864-170718864 on Build GRCh38
3827679 c.130C>T p.R44W Chr.5: 170718799-170718799 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34377595 Chr.5: 170718917-170718917 on Build GRCh38
rs897056042 Chr.5: 170718721-170718721 on Build GRCh38
rs754218719 Chr.5: 170718720-170718720 on Build GRCh38
rs562177002 Chr.5: 170718975-170718975 on Build GRCh38
rs781715532 Chr.5: 170718787-170718787 on Build GRCh38
rs1008260063 Chr.5: 170718857-170718857 on Build GRCh38
rs750538048 Chr.5: 170718869-170718869 on Build GRCh38
rs532478634 Chr.5: 170718911-170718911 on Build GRCh38
rs775378266 Chr.5: 170718912-170718912 on Build GRCh38
rs754395821 Chr.5: 170718931-170718931 on Build GRCh38
rs778764319 Chr.5: 170718903-170718903 on Build GRCh38
rs759073985 Chr.5: 170718917-170718918 on Build GRCh38
rs763967497 Chr.5: 170719016-170719016 on Build GRCh38
rs920138189 Chr.5: 170718978-170718978 on Build GRCh38
rs950075007 Chr.5: 170718918-170718918 on Build GRCh38
rs746476533 Chr.5: 170718976-170718976 on Build GRCh38
rs957207798 Chr.5: 170718911-170718911 on Build GRCh38
rs767941388 Chr.5: 170718889-170718889 on Build GRCh38
rs941171366 Chr.5: 170718706-170718706 on Build GRCh38
rs777345443 Chr.5: 170718804-170718804 on Build GRCh38
rs759525940 Chr.5: 170718822-170718822 on Build GRCh38
rs767583636 Chr.5: 170718924-170718924 on Build GRCh38
rs116648788 Chr.5: 170718716-170718716 on Build GRCh38
rs752941135 Chr.5: 170718891-170718891 on Build GRCh38
rs752673681 Chr.5: 170718741-170718741 on Build GRCh38
rs768679350 Chr.5: 170718914-170718914 on Build GRCh38
rs145062462 Chr.5: 170718872-170718872 on Build GRCh38
rs748677083 Chr.5: 170718788-170718788 on Build GRCh38
rs759378796 Chr.5: 170718717-170718717 on Build GRCh38
rs763897144 Chr.5: 170718868-170718868 on Build GRCh38
rs560631743 Chr.5: 170719013-170719013 on Build GRCh38
rs767599302 Chr.5: 170718740-170718740 on Build GRCh38
rs375016690 Chr.5: 170718892-170718892 on Build GRCh38
rs79599238 Chr.5: 170718968-170718968 on Build GRCh38
rs533670866 Chr.5: 170718830-170718830 on Build GRCh38
rs529944333 Chr.5: 170718911-170718912 on Build GRCh38
rs941488256 Chr.5: 170718967-170718967 on Build GRCh38
rs149882468 Chr.5: 170718785-170718785 on Build GRCh38
rs750402454 Chr.5: 170718762-170718762 on Build GRCh38
rs748710812 Chr.5: 170718814-170718814 on Build GRCh38
rs774611923 Chr.5: 170718797-170718797 on Build GRCh38
rs868827395 Chr.5: 170718769-170718769 on Build GRCh38
rs770645158 Chr.5: 170718714-170718714 on Build GRCh38
rs370873026 Chr.5: 170718823-170718823 on Build GRCh38
rs568805596 Chr.5: 170718682-170718682 on Build GRCh38
rs200294438 Chr.5: 170718916-170718916 on Build GRCh38
rs758678065 Chr.5: 170718764-170718764 on Build GRCh38
rs553548344 Chr.5: 170718843-170718843 on Build GRCh38
rs994135191 Chr.5: 170718744-170718745 on Build GRCh38
rs760664501 Chr.5: 170718848-170718848 on Build GRCh38
rs141241101 Chr.5: 170718885-170718885 on Build GRCh38
rs745523251 Chr.5: 170718909-170718909 on Build GRCh38
rs756820477 Chr.5: 170718898-170718898 on Build GRCh38
rs771977715 Chr.5: 170718831-170718831 on Build GRCh38
rs765369969 Chr.5: 170718755-170718755 on Build GRCh38
rs150751695 Chr.5: 170718926-170718926 on Build GRCh38
rs771794095 Chr.5: 170718910-170718910 on Build GRCh38
rs780201363 Chr.5: 170718771-170718771 on Build GRCh38
rs751776849 Chr.5: 170718882-170718882 on Build GRCh38
rs369653987 Chr.5: 170718913-170718913 on Build GRCh38
rs770145353 Chr.5: 170718791-170718791 on Build GRCh38
rs577358260 Chr.5: 170718786-170718786 on Build GRCh38
rs930166462 Chr.5: 170719015-170719015 on Build GRCh38
rs529516562 Chr.5: 170719017-170719017 on Build GRCh38
rs753133567 Chr.5: 170718925-170718925 on Build GRCh38
rs150181862 Chr.5: 170718840-170718840 on Build GRCh38
rs369778254 Chr.5: 170718709-170718709 on Build GRCh38
rs10039251 Chr.5: 170718745-170718745 on Build GRCh38
rs762998289 Chr.5: 170718871-170718871 on Build GRCh38
rs181255804 Chr.5: 170718897-170718897 on Build GRCh38
rs770503987 Chr.5: 170718910-170718911 on Build GRCh38
rs926972414 Chr.5: 170718945-170718945 on Build GRCh38

View Full Product Details