Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176595448-176595954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDHR2
Entrez Gene ID
54825
Gene Name
cadherin related family member 2
Gene Aliases
PCDH24, PCLKC
UniGene
Hs.4205
Gene Chromosome Location
Chr.5: 176542511-176595768 on Build GRCh38
Transcripts
NM_001171976.1 NM_017675.4 NM_052899.2

Gene Symbol
GPRIN1
Entrez Gene ID
114787
Gene Name
G protein regulated inducer of neurite outgrowth 1
Gene Aliases
GRIN1
UniGene
Hs.150549
Gene Chromosome Location
Chr.5: 176595802-176610130 on Build GRCh38
Transcripts
NM_001171976.1 NM_017675.4 NM_052899.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
233051 c.3890G>A p.G1297E Chr.5: 176595629-176595629 on Build GRCh38
7116880 c.3808C>T p.H1270Y Chr.5: 176595547-176595547 on Build GRCh38
6170575 c.3932G>A p.*1311* Chr.5: 176595671-176595671 on Build GRCh38
3614359 c.3877G>A p.G1293R Chr.5: 176595616-176595616 on Build GRCh38
1620079 c.3840G>C p.L1280L Chr.5: 176595579-176595579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138301358 Chr.5: 176595857-176595857 on Build GRCh38
rs754839509 Chr.5: 176595607-176595607 on Build GRCh38
rs937835946 Chr.5: 176595765-176595765 on Build GRCh38
rs749470354 Chr.5: 176595579-176595579 on Build GRCh38
rs953721067 Chr.5: 176595779-176595779 on Build GRCh38
rs773544974 Chr.5: 176595482-176595482 on Build GRCh38
rs1043625708 Chr.5: 176595718-176595718 on Build GRCh38
rs896978489 Chr.5: 176595911-176595911 on Build GRCh38
rs778169607 Chr.5: 176595574-176595574 on Build GRCh38
rs750621897 Chr.5: 176595694-176595694 on Build GRCh38
rs906577104 Chr.5: 176595876-176595876 on Build GRCh38
rs772890402 Chr.5: 176595662-176595662 on Build GRCh38
rs762150531 Chr.5: 176595648-176595648 on Build GRCh38
rs766972868 Chr.5: 176595678-176595679 on Build GRCh38
rs954135031 Chr.5: 176595930-176595930 on Build GRCh38
rs943789741 Chr.5: 176595788-176595788 on Build GRCh38
rs566189556 Chr.5: 176595537-176595537 on Build GRCh38
rs971235249 Chr.5: 176595491-176595491 on Build GRCh38
rs764809131 Chr.5: 176595595-176595595 on Build GRCh38
rs1030261328 Chr.5: 176595870-176595870 on Build GRCh38
rs779185964 Chr.5: 176595625-176595625 on Build GRCh38
rs774942320 Chr.5: 176595489-176595489 on Build GRCh38
rs772165277 Chr.5: 176595633-176595633 on Build GRCh38
rs561252582 Chr.5: 176595587-176595587 on Build GRCh38
rs763787314 Chr.5: 176595517-176595517 on Build GRCh38
rs985571144 Chr.5: 176595782-176595782 on Build GRCh38
rs768002048 Chr.5: 176595922-176595922 on Build GRCh38
rs750387948 Chr.5: 176595527-176595527 on Build GRCh38
rs924123633 Chr.5: 176595802-176595802 on Build GRCh38
rs759135912 Chr.5: 176595655-176595655 on Build GRCh38
rs1004989497 Chr.5: 176595879-176595879 on Build GRCh38
rs576350560 Chr.5: 176595717-176595717 on Build GRCh38
rs34418000 Chr.5: 176595583-176595583 on Build GRCh38
rs761070780 Chr.5: 176595592-176595592 on Build GRCh38
rs373675182 Chr.5: 176595598-176595598 on Build GRCh38
rs562666087 Chr.5: 176595789-176595789 on Build GRCh38
rs764403091 Chr.5: 176595593-176595593 on Build GRCh38
rs771051706 Chr.5: 176595580-176595580 on Build GRCh38
rs1001088684 Chr.5: 176595615-176595615 on Build GRCh38
rs547866475 Chr.5: 176595533-176595533 on Build GRCh38
rs143952500 Chr.5: 176595603-176595603 on Build GRCh38
rs961292469 Chr.5: 176595530-176595530 on Build GRCh38
rs545459748 Chr.5: 176595790-176595790 on Build GRCh38
rs757583504 Chr.5: 176595477-176595477 on Build GRCh38
rs767925256 Chr.5: 176595500-176595500 on Build GRCh38
rs1042539684 Chr.5: 176595599-176595599 on Build GRCh38
rs776290108 Chr.5: 176595594-176595594 on Build GRCh38
rs368114066 Chr.5: 176595628-176595628 on Build GRCh38
rs895884390 Chr.5: 176595781-176595781 on Build GRCh38
rs867294244 Chr.5: 176595559-176595559 on Build GRCh38
rs559804698 Chr.5: 176595673-176595673 on Build GRCh38
rs779047877 Chr.5: 176595690-176595690 on Build GRCh38
rs564142639 Chr.5: 176595831-176595831 on Build GRCh38
rs771092678 Chr.5: 176595486-176595486 on Build GRCh38
rs974000172 Chr.5: 176595724-176595724 on Build GRCh38
rs116373545 Chr.5: 176595915-176595915 on Build GRCh38
rs772517444 Chr.5: 176595585-176595585 on Build GRCh38
rs375445534 Chr.5: 176595511-176595511 on Build GRCh38
rs114504389 Chr.5: 176595645-176595645 on Build GRCh38
rs778309422 Chr.5: 176595914-176595914 on Build GRCh38
rs1005769330 Chr.5: 176595828-176595828 on Build GRCh38
rs747522280 Chr.5: 176595707-176595707 on Build GRCh38
rs758431718 Chr.5: 176595695-176595695 on Build GRCh38
rs776916587 Chr.5: 176595647-176595647 on Build GRCh38
rs770154431 Chr.5: 176595719-176595719 on Build GRCh38
rs752206361 Chr.5: 176595758-176595758 on Build GRCh38
rs756793079 Chr.5: 176595522-176595522 on Build GRCh38
rs370492729 Chr.5: 176595597-176595597 on Build GRCh38
rs754894579 Chr.5: 176595675-176595675 on Build GRCh38
rs764845336 Chr.5: 176595661-176595661 on Build GRCh38
rs541631603 Chr.5: 176595663-176595663 on Build GRCh38
rs759898872 Chr.5: 176595495-176595495 on Build GRCh38
rs753073978 Chr.5: 176595503-176595503 on Build GRCh38
rs746847146 Chr.5: 176595540-176595540 on Build GRCh38
rs749523542 Chr.5: 176595610-176595610 on Build GRCh38
rs948071449 Chr.5: 176595837-176595837 on Build GRCh38
rs778240094 Chr.5: 176595525-176595525 on Build GRCh38
rs113238915 Chr.5: 176595665-176595665 on Build GRCh38
rs752601181 Chr.5: 176595679-176595679 on Build GRCh38
rs774224954 Chr.5: 176595582-176595582 on Build GRCh38
rs973695587 Chr.5: 176595562-176595562 on Build GRCh38
rs912307465 Chr.5: 176595784-176595784 on Build GRCh38
rs140266714 Chr.5: 176595609-176595609 on Build GRCh38
rs995772151 Chr.5: 176595912-176595912 on Build GRCh38
rs140473174 Chr.5: 176595520-176595520 on Build GRCh38
rs967683397 Chr.5: 176595924-176595925 on Build GRCh38
rs754788217 Chr.5: 176595548-176595548 on Build GRCh38
rs767556039 Chr.5: 176595678-176595678 on Build GRCh38
rs753092368 Chr.5: 176595512-176595513 on Build GRCh38
rs763492255 Chr.5: 176595654-176595655 on Build GRCh38
rs1054021715 Chr.5: 176595817-176595817 on Build GRCh38
rs748499977 Chr.5: 176595710-176595710 on Build GRCh38
rs200234481 Chr.5: 176595641-176595641 on Build GRCh38
rs550468153 Chr.5: 176595905-176595905 on Build GRCh38
rs1015408059 Chr.5: 176595893-176595893 on Build GRCh38
rs111390019 Chr.5: 176595855-176595855 on Build GRCh38
rs764684691 Chr.5: 176595668-176595668 on Build GRCh38
rs867462520 Chr.5: 176595654-176595654 on Build GRCh38
rs375673297 Chr.5: 176595687-176595687 on Build GRCh38
rs936869187 Chr.5: 176595807-176595807 on Build GRCh38
rs757484467 Chr.5: 176595616-176595616 on Build GRCh38
rs368523225 Chr.5: 176595708-176595708 on Build GRCh38
rs1027347051 Chr.5: 176595744-176595744 on Build GRCh38
rs763164712 Chr.5: 176595484-176595484 on Build GRCh38
rs769022271 Chr.5: 176595646-176595646 on Build GRCh38
rs780051272 Chr.5: 176595696-176595696 on Build GRCh38

View Full Product Details