Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30172118-30172619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM15
Entrez Gene ID
89870
Gene Name
tripartite motif containing 15
Gene Aliases
RNF93, ZNF178, ZNFB7
UniGene
Hs.591789
Gene Chromosome Location
Chr.6: 30163206-30172696 on Build GRCh38
Transcripts
NM_033229.2 XM_011514987.1 XM_011514988.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4526323 c.1389G>A p.L463L Chr.6: 30172340-30172340 on Build GRCh38
6499868 c.1384_1385insAA p.T462fs*3 Chr.6: 30172335-30172336 on Build GRCh38
6807872 c.1238G>A p.R413H Chr.6: 30172189-30172189 on Build GRCh38
6807873 c.1385C>T p.T462M Chr.6: 30172336-30172336 on Build GRCh38
1442983 c.1213G>A p.D405N Chr.6: 30172164-30172164 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1063278 Chr.6: 30172284-30172284 on Build GRCh38
rs776924380 Chr.6: 30172221-30172221 on Build GRCh38
rs761622621 Chr.6: 30172376-30172376 on Build GRCh38
rs758845185 Chr.6: 30172556-30172556 on Build GRCh38
rs748506643 Chr.6: 30172191-30172191 on Build GRCh38
rs570900960 Chr.6: 30172589-30172589 on Build GRCh38
rs371041335 Chr.6: 30172503-30172506 on Build GRCh38
rs748837389 Chr.6: 30172173-30172173 on Build GRCh38
rs746634759 Chr.6: 30172396-30172396 on Build GRCh38
rs369877929 Chr.6: 30172395-30172395 on Build GRCh38
rs982092575 Chr.6: 30172491-30172491 on Build GRCh38
rs1063276 Chr.6: 30172264-30172264 on Build GRCh38
rs1063280 Chr.6: 30172565-30172565 on Build GRCh38
rs541125510 Chr.6: 30172269-30172269 on Build GRCh38
rs750368271 Chr.6: 30172387-30172387 on Build GRCh38
rs867489743 Chr.6: 30172288-30172288 on Build GRCh38
rs915024713 Chr.6: 30172582-30172582 on Build GRCh38
rs1018675161 Chr.6: 30172369-30172369 on Build GRCh38
rs757242994 Chr.6: 30172157-30172157 on Build GRCh38
rs781702580 Chr.6: 30172596-30172596 on Build GRCh38
rs969338906 Chr.6: 30172439-30172439 on Build GRCh38
rs778460871 Chr.6: 30172541-30172541 on Build GRCh38
rs748823955 Chr.6: 30172301-30172301 on Build GRCh38
rs754476518 Chr.6: 30172528-30172528 on Build GRCh38
rs779780664 Chr.6: 30172171-30172171 on Build GRCh38
rs575112359 Chr.6: 30172592-30172592 on Build GRCh38
rs765419095 Chr.6: 30172362-30172362 on Build GRCh38
rs115649931 Chr.6: 30172194-30172194 on Build GRCh38
rs927997917 Chr.6: 30172498-30172498 on Build GRCh38
rs1063279 Chr.6: 30172292-30172292 on Build GRCh38
rs138979251 Chr.6: 30172588-30172588 on Build GRCh38
rs769066003 Chr.6: 30172189-30172189 on Build GRCh38
rs774642498 Chr.6: 30172190-30172190 on Build GRCh38
rs370143829 Chr.6: 30172405-30172405 on Build GRCh38
rs779367161 Chr.6: 30172286-30172286 on Build GRCh38
rs773878262 Chr.6: 30172336-30172336 on Build GRCh38
rs960523625 Chr.6: 30172543-30172543 on Build GRCh38
rs542411839 Chr.6: 30172419-30172419 on Build GRCh38
rs777114069 Chr.6: 30172467-30172467 on Build GRCh38
rs759672374 Chr.6: 30172280-30172281 on Build GRCh38
rs768943796 Chr.6: 30172446-30172446 on Build GRCh38
rs753965939 Chr.6: 30172148-30172148 on Build GRCh38
rs751365999 Chr.6: 30172156-30172156 on Build GRCh38
rs142991113 Chr.6: 30172238-30172238 on Build GRCh38
rs532059328 Chr.6: 30172417-30172417 on Build GRCh38
rs759745176 Chr.6: 30172226-30172226 on Build GRCh38
rs980499668 Chr.6: 30172591-30172591 on Build GRCh38
rs767477887 Chr.6: 30172263-30172263 on Build GRCh38
rs759511241 Chr.6: 30172579-30172579 on Build GRCh38
rs747753929 Chr.6: 30172548-30172548 on Build GRCh38
rs753244485 Chr.6: 30172290-30172291 on Build GRCh38
rs773823321 Chr.6: 30172211-30172211 on Build GRCh38
rs916843368 Chr.6: 30172433-30172433 on Build GRCh38
rs1007572028 Chr.6: 30172406-30172406 on Build GRCh38
rs577557690 Chr.6: 30172361-30172361 on Build GRCh38
rs747458380 Chr.6: 30172347-30172347 on Build GRCh38
rs766319104 Chr.6: 30172216-30172216 on Build GRCh38
rs754642700 Chr.6: 30172317-30172317 on Build GRCh38
rs777371306 Chr.6: 30172154-30172154 on Build GRCh38
rs760869221 Chr.6: 30172215-30172215 on Build GRCh38
rs1011232786 Chr.6: 30172262-30172262 on Build GRCh38
rs948065928 Chr.6: 30172192-30172192 on Build GRCh38
rs751404304 Chr.6: 30172253-30172253 on Build GRCh38
rs918789279 Chr.6: 30172157-30172157 on Build GRCh38
rs745378049 Chr.6: 30172167-30172167 on Build GRCh38
rs935138344 Chr.6: 30172228-30172228 on Build GRCh38
rs936596592 Chr.6: 30172252-30172252 on Build GRCh38
rs906646318 Chr.6: 30172219-30172219 on Build GRCh38
rs1028917682 Chr.6: 30172275-30172275 on Build GRCh38
rs765710708 Chr.6: 30172284-30172284 on Build GRCh38
rs150264994 Chr.6: 30172390-30172390 on Build GRCh38
rs779598508 Chr.6: 30172394-30172394 on Build GRCh38
rs993738975 Chr.6: 30172449-30172449 on Build GRCh38
rs771045627 Chr.6: 30172348-30172348 on Build GRCh38
rs758233481 Chr.6: 30172153-30172153 on Build GRCh38
rs1063277 Chr.6: 30172280-30172280 on Build GRCh38
rs776634571 Chr.6: 30172354-30172354 on Build GRCh38
rs1052339120 Chr.6: 30172279-30172279 on Build GRCh38
rs753866886 Chr.6: 30172248-30172248 on Build GRCh38
rs1008846702 Chr.6: 30172595-30172595 on Build GRCh38
rs775653581 Chr.6: 30172375-30172375 on Build GRCh38
rs557429382 Chr.6: 30172324-30172324 on Build GRCh38
rs757015864 Chr.6: 30172260-30172260 on Build GRCh38
rs778602332 Chr.6: 30172320-30172320 on Build GRCh38
rs935400611 Chr.6: 30172564-30172564 on Build GRCh38
rs375289482 Chr.6: 30172139-30172139 on Build GRCh38
rs562268693 Chr.6: 30172421-30172421 on Build GRCh38

View Full Product Details