Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 107744712-107745213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCML4
Entrez Gene ID
256380
Gene Name
sex comb on midleg-like 4 (Drosophila)
Gene Aliases
dJ47M23.1
UniGene
Hs.486109
Gene Chromosome Location
Chr.6: 107697297-107845959 on Build GRCh38
Transcripts
NM_001286408.1 NM_198081.4 XM_011535703.2 XM_011535704.2 XM_011535706.1 XM_011535707.2 XM_011535710.2 XM_017010677.1 XM_017010678.1 XM_017010679.1 XM_017010680.1 XM_017010681.1 XM_017010682.1 XM_017010683.1 XM_017010684.1 XM_017010685.1 XM_017010686.1 XM_017010687.1 XM_017010688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6772786 c.507C>T p.C169C Chr.6: 107745037-107745037 on Build GRCh38
5716012 c.535C>T p.P179S Chr.6: 107745009-107745009 on Build GRCh38
6772776 c.472C>T p.R158C Chr.6: 107745072-107745072 on Build GRCh38
5592985 c.477C>T p.F159F Chr.6: 107745067-107745067 on Build GRCh38
4155942 c.581G>T p.G194V Chr.6: 107744963-107744963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778617727 Chr.6: 107745008-107745008 on Build GRCh38
rs367626585 Chr.6: 107744896-107744896 on Build GRCh38
rs879617196 Chr.6: 107744841-107744841 on Build GRCh38
rs547328477 Chr.6: 107744855-107744855 on Build GRCh38
rs771489633 Chr.6: 107744951-107744951 on Build GRCh38
rs538326075 Chr.6: 107745037-107745037 on Build GRCh38
rs760034859 Chr.6: 107744961-107744961 on Build GRCh38
rs778138871 Chr.6: 107745032-107745032 on Build GRCh38
rs749454362 Chr.6: 107745036-107745036 on Build GRCh38
rs112875714 Chr.6: 107745121-107745121 on Build GRCh38
rs562131062 Chr.6: 107745178-107745178 on Build GRCh38
rs761352662 Chr.6: 107745185-107745185 on Build GRCh38
rs773618973 Chr.6: 107744945-107744945 on Build GRCh38
rs752162500 Chr.6: 107744898-107744898 on Build GRCh38
rs78465925 Chr.6: 107744737-107744737 on Build GRCh38
rs113433888 Chr.6: 107744890-107744890 on Build GRCh38
rs749405198 Chr.6: 107745119-107745119 on Build GRCh38
rs763719878 Chr.6: 107745001-107745001 on Build GRCh38
rs934710621 Chr.6: 107745025-107745025 on Build GRCh38
rs767935814 Chr.6: 107744963-107744963 on Build GRCh38
rs772602597 Chr.6: 107745071-107745071 on Build GRCh38
rs572164034 Chr.6: 107745161-107745161 on Build GRCh38
rs991405683 Chr.6: 107744875-107744875 on Build GRCh38
rs555376188 Chr.6: 107745050-107745050 on Build GRCh38
rs374292351 Chr.6: 107745175-107745175 on Build GRCh38
rs764899782 Chr.6: 107745099-107745099 on Build GRCh38
rs535969370 Chr.6: 107745193-107745193 on Build GRCh38
rs753355646 Chr.6: 107745085-107745085 on Build GRCh38
rs774861358 Chr.6: 107744955-107744955 on Build GRCh38
rs958632012 Chr.6: 107744811-107744811 on Build GRCh38
rs140445627 Chr.6: 107745084-107745084 on Build GRCh38
rs79314211 Chr.6: 107744872-107744872 on Build GRCh38
rs748239424 Chr.6: 107745028-107745028 on Build GRCh38
rs753551974 Chr.6: 107745003-107745003 on Build GRCh38
rs1024232045 Chr.6: 107745169-107745169 on Build GRCh38
rs761405518 Chr.6: 107745087-107745087 on Build GRCh38
rs376727961 Chr.6: 107745103-107745103 on Build GRCh38
rs149397511 Chr.6: 107745110-107745110 on Build GRCh38
rs1043412707 Chr.6: 107744930-107744930 on Build GRCh38
rs775924065 Chr.6: 107745171-107745171 on Build GRCh38
rs540821140 Chr.6: 107745115-107745115 on Build GRCh38
rs148576733 Chr.6: 107745127-107745127 on Build GRCh38
rs750206499 Chr.6: 107745010-107745010 on Build GRCh38
rs771913383 Chr.6: 107744926-107744926 on Build GRCh38
rs770058612 Chr.6: 107745030-107745030 on Build GRCh38
rs760171261 Chr.6: 107745080-107745080 on Build GRCh38
rs752214060 Chr.6: 107744968-107744968 on Build GRCh38
rs750199850 Chr.6: 107745102-107745102 on Build GRCh38
rs746161888 Chr.6: 107745006-107745007 on Build GRCh38
rs775916413 Chr.6: 107745072-107745072 on Build GRCh38
rs1027038325 Chr.6: 107745120-107745120 on Build GRCh38
rs754971320 Chr.6: 107745114-107745114 on Build GRCh38
rs758315215 Chr.6: 107745019-107745019 on Build GRCh38
rs770107756 Chr.6: 107744943-107744943 on Build GRCh38
rs371508343 Chr.6: 107744946-107744946 on Build GRCh38
rs6922304 Chr.6: 107744940-107744940 on Build GRCh38
rs917248435 Chr.6: 107744829-107744829 on Build GRCh38
rs779895279 Chr.6: 107745024-107745024 on Build GRCh38
rs995184696 Chr.6: 107745107-107745107 on Build GRCh38
rs117229252 Chr.6: 107744778-107744778 on Build GRCh38
rs368339985 Chr.6: 107744777-107744777 on Build GRCh38
rs746224053 Chr.6: 107745139-107745139 on Build GRCh38
rs753605657 Chr.6: 107744900-107744900 on Build GRCh38
rs991436038 Chr.6: 107744815-107744815 on Build GRCh38
rs147163297 Chr.6: 107745064-107745064 on Build GRCh38
rs371356387 Chr.6: 107745167-107745167 on Build GRCh38
rs951253549 Chr.6: 107744747-107744747 on Build GRCh38
rs755744849 Chr.6: 107744899-107744899 on Build GRCh38
rs951164424 Chr.6: 107744800-107744800 on Build GRCh38
rs200851875 Chr.6: 107744913-107744913 on Build GRCh38
rs1005396768 Chr.6: 107744767-107744767 on Build GRCh38
rs928500588 Chr.6: 107744860-107744860 on Build GRCh38
rs971427414 Chr.6: 107744866-107744866 on Build GRCh38
rs189161625 Chr.6: 107745176-107745176 on Build GRCh38
rs779699996 Chr.6: 107745111-107745111 on Build GRCh38
rs757099860 Chr.6: 107744902-107744902 on Build GRCh38
rs756903575 Chr.6: 107745006-107745006 on Build GRCh38
rs772382808 Chr.6: 107745157-107745157 on Build GRCh38

View Full Product Details