Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 107749544-107750043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCML4
Entrez Gene ID
256380
Gene Name
sex comb on midleg-like 4 (Drosophila)
Gene Aliases
dJ47M23.1
UniGene
Hs.486109
Gene Chromosome Location
Chr.6: 107697297-107845959 on Build GRCh38
Transcripts
NM_001286408.1 NM_198081.4 XM_011535703.2 XM_011535704.2 XM_011535706.1 XM_011535707.2 XM_017010677.1 XM_017010678.1 XM_017010679.1 XM_017010680.1 XM_017010681.1 XM_017010682.1 XM_017010683.1 XM_017010684.1 XM_017010685.1 XM_017010686.1 XM_017010687.1 XM_017010688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4479887 c.149C>T p.P50L Chr.6: 107749734-107749734 on Build GRCh38
5716631 c.148C>T p.P50S Chr.6: 107749735-107749735 on Build GRCh38
3723482 c.116G>A p.R39Q Chr.6: 107749767-107749767 on Build GRCh38
287133 c.79C>T p.R27W Chr.6: 107749804-107749804 on Build GRCh38
4839749 c.113C>T p.P38L Chr.6: 107749770-107749770 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377664747 Chr.6: 107749650-107749650 on Build GRCh38
rs551886211 Chr.6: 107749824-107749824 on Build GRCh38
rs764498398 Chr.6: 107749703-107749703 on Build GRCh38
rs991990393 Chr.6: 107749969-107749969 on Build GRCh38
rs896051145 Chr.6: 107749755-107749755 on Build GRCh38
rs140517190 Chr.6: 107749718-107749718 on Build GRCh38
rs754599652 Chr.6: 107749774-107749774 on Build GRCh38
rs533889253 Chr.6: 107749956-107749956 on Build GRCh38
rs746604149 Chr.6: 107749771-107749771 on Build GRCh38
rs1006698072 Chr.6: 107749783-107749783 on Build GRCh38
rs191381751 Chr.6: 107750017-107750017 on Build GRCh38
rs1056034624 Chr.6: 107749742-107749742 on Build GRCh38
rs752976156 Chr.6: 107749686-107749686 on Build GRCh38
rs75193274 Chr.6: 107749628-107749628 on Build GRCh38
rs118096825 Chr.6: 107750016-107750016 on Build GRCh38
rs928650607 Chr.6: 107749944-107749944 on Build GRCh38
rs200167364 Chr.6: 107749768-107749768 on Build GRCh38
rs1021877359 Chr.6: 107749673-107749673 on Build GRCh38
rs774429199 Chr.6: 107749675-107749675 on Build GRCh38
rs745938993 Chr.6: 107749796-107749796 on Build GRCh38
rs1053446369 Chr.6: 107750015-107750015 on Build GRCh38
rs991642674 Chr.6: 107749933-107749933 on Build GRCh38
rs142613803 Chr.6: 107749948-107749948 on Build GRCh38
rs75107588 Chr.6: 107749570-107749570 on Build GRCh38
rs568675664 Chr.6: 107749828-107749828 on Build GRCh38
rs200973219 Chr.6: 107749678-107749678 on Build GRCh38
rs754621277 Chr.6: 107749861-107749861 on Build GRCh38
rs561631407 Chr.6: 107749725-107749725 on Build GRCh38
rs375625250 Chr.6: 107749767-107749767 on Build GRCh38
rs762313147 Chr.6: 107749811-107749811 on Build GRCh38
rs376604668 Chr.6: 107749704-107749704 on Build GRCh38
rs757850264 Chr.6: 107749710-107749710 on Build GRCh38
rs748183780 Chr.6: 107749804-107749804 on Build GRCh38
rs180709464 Chr.6: 107749930-107749930 on Build GRCh38
rs200114918 Chr.6: 107749748-107749748 on Build GRCh38
rs902143563 Chr.6: 107749987-107749987 on Build GRCh38
rs111981256 Chr.6: 107749617-107749617 on Build GRCh38
rs963911515 Chr.6: 107749715-107749715 on Build GRCh38
rs200389996 Chr.6: 107749648-107749648 on Build GRCh38
rs370516097 Chr.6: 107749831-107749831 on Build GRCh38
rs375147910 Chr.6: 107749837-107749837 on Build GRCh38
rs769748312 Chr.6: 107749728-107749728 on Build GRCh38
rs374418850 Chr.6: 107750020-107750020 on Build GRCh38
rs150723162 Chr.6: 107750000-107750001 on Build GRCh38
rs766109526 Chr.6: 107749726-107749726 on Build GRCh38
rs774375860 Chr.6: 107749729-107749729 on Build GRCh38
rs769687908 Chr.6: 107749789-107749789 on Build GRCh38
rs775440011 Chr.6: 107749802-107749802 on Build GRCh38
rs113350523 Chr.6: 107749871-107749871 on Build GRCh38
rs915957188 Chr.6: 107749970-107749970 on Build GRCh38
rs781322087 Chr.6: 107749803-107749803 on Build GRCh38
rs955270068 Chr.6: 107749698-107749698 on Build GRCh38
rs148455559 Chr.6: 107749676-107749676 on Build GRCh38
rs146259790 Chr.6: 107749647-107749647 on Build GRCh38
rs764455186 Chr.6: 107749754-107749754 on Build GRCh38
rs762907235 Chr.6: 107749756-107749757 on Build GRCh38
rs112676572 Chr.6: 107749770-107749770 on Build GRCh38
rs113130210 Chr.6: 107750001-107750002 on Build GRCh38
rs745819716 Chr.6: 107749881-107749881 on Build GRCh38
rs772209399 Chr.6: 107749801-107749801 on Build GRCh38
rs185488092 Chr.6: 107749983-107749983 on Build GRCh38
rs201967131 Chr.6: 107749696-107749696 on Build GRCh38
rs775844019 Chr.6: 107749750-107749750 on Build GRCh38
rs371372189 Chr.6: 107749727-107749727 on Build GRCh38
rs143609542 Chr.6: 107749812-107749812 on Build GRCh38
rs150131727 Chr.6: 107749724-107749724 on Build GRCh38
rs759653925 Chr.6: 107749741-107749741 on Build GRCh38
rs780993385 Chr.6: 107749781-107749781 on Build GRCh38
rs745399101 Chr.6: 107749711-107749711 on Build GRCh38
rs758089532 Chr.6: 107749717-107749717 on Build GRCh38
rs760946291 Chr.6: 107749751-107749751 on Build GRCh38
rs201863497 Chr.6: 107749661-107749661 on Build GRCh38
rs148891280 Chr.6: 107749769-107749769 on Build GRCh38
rs773186443 Chr.6: 107749790-107749790 on Build GRCh38
rs145577601 Chr.6: 107749761-107749761 on Build GRCh38
rs768455029 Chr.6: 107749634-107749634 on Build GRCh38
rs774387521 Chr.6: 107749841-107749841 on Build GRCh38
rs747970249 Chr.6: 107749787-107749787 on Build GRCh38
rs79393866 Chr.6: 107749630-107749630 on Build GRCh38

View Full Product Details