Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 108043285-108043821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSTM1
Entrez Gene ID
28962
Gene Name
osteopetrosis associated transmembrane protein 1
Gene Aliases
GIPN, GL, HSPC019, OPTB5
UniGene
Hs.226780
Gene Chromosome Location
Chr.6: 108041409-108074737 on Build GRCh38
Transcripts
NM_014028.3

SNP Detail

SNP ID Chromosome Location
rs9374008 Chr.6: 108043451-108043451 on Build GRCh38
rs577908204 Chr.6: 108043631-108043631 on Build GRCh38
rs577413432 Chr.6: 108043679-108043679 on Build GRCh38
rs920677135 Chr.6: 108043625-108043625 on Build GRCh38
rs920103241 Chr.6: 108043738-108043738 on Build GRCh38
rs148730821 Chr.6: 108043703-108043703 on Build GRCh38
rs907830667 Chr.6: 108043676-108043676 on Build GRCh38
rs948182939 Chr.6: 108043743-108043743 on Build GRCh38
rs528241865 Chr.6: 108043330-108043330 on Build GRCh38
rs1009328205 Chr.6: 108043324-108043324 on Build GRCh38
rs937698436 Chr.6: 108043794-108043794 on Build GRCh38
rs116796449 Chr.6: 108043358-108043358 on Build GRCh38
rs867928731 Chr.6: 108043606-108043606 on Build GRCh38
rs572109702 Chr.6: 108043468-108043468 on Build GRCh38
rs955902671 Chr.6: 108043571-108043571 on Build GRCh38
rs566197276 Chr.6: 108043587-108043587 on Build GRCh38
rs879856212 Chr.6: 108043326-108043326 on Build GRCh38
rs17069228 Chr.6: 108043624-108043624 on Build GRCh38
rs9372179 Chr.6: 108043433-108043433 on Build GRCh38
rs140320992 Chr.6: 108043344-108043344 on Build GRCh38
rs368663751 Chr.6: 108043782-108043782 on Build GRCh38
rs974770728 Chr.6: 108043328-108043328 on Build GRCh38
rs145514255 Chr.6: 108043580-108043580 on Build GRCh38
rs145259466 Chr.6: 108043687-108043687 on Build GRCh38
rs939584757 Chr.6: 108043394-108043394 on Build GRCh38
rs886060967 Chr.6: 108043762-108043762 on Build GRCh38
rs573274541 Chr.6: 108043714-108043714 on Build GRCh38
rs1064346 Chr.6: 108043747-108043747 on Build GRCh38
rs1031303504 Chr.6: 108043537-108043537 on Build GRCh38
rs1021311058 Chr.6: 108043341-108043341 on Build GRCh38
rs886060965 Chr.6: 108043316-108043316 on Build GRCh38
rs891434231 Chr.6: 108043781-108043781 on Build GRCh38
rs12567 Chr.6: 108043378-108043378 on Build GRCh38
rs1000785037 Chr.6: 108043627-108043627 on Build GRCh38
rs927521800 Chr.6: 108043787-108043787 on Build GRCh38
rs73504439 Chr.6: 108043418-108043418 on Build GRCh38
rs937645914 Chr.6: 108043511-108043511 on Build GRCh38
rs904263669 Chr.6: 108043751-108043751 on Build GRCh38
rs926312576 Chr.6: 108043494-108043494 on Build GRCh38
rs1056156496 Chr.6: 108043524-108043524 on Build GRCh38
rs961435170 Chr.6: 108043701-108043701 on Build GRCh38
rs1057119821 Chr.6: 108043407-108043407 on Build GRCh38
rs954033801 Chr.6: 108043643-108043643 on Build GRCh38
rs972872350 Chr.6: 108043426-108043426 on Build GRCh38
rs1053299538 Chr.6: 108043778-108043778 on Build GRCh38
rs767937896 Chr.6: 108043607-108043608 on Build GRCh38
rs954147508 Chr.6: 108043386-108043386 on Build GRCh38
rs192232328 Chr.6: 108043630-108043630 on Build GRCh38
rs999912483 Chr.6: 108043762-108043762 on Build GRCh38
rs920709621 Chr.6: 108043334-108043334 on Build GRCh38
rs987384439 Chr.6: 108043577-108043577 on Build GRCh38
rs886060966 Chr.6: 108043386-108043386 on Build GRCh38
rs943226650 Chr.6: 108043314-108043314 on Build GRCh38
rs886682312 Chr.6: 108043389-108043389 on Build GRCh38
rs557667717 Chr.6: 108043619-108043619 on Build GRCh38
rs1009010049 Chr.6: 108043783-108043783 on Build GRCh38
rs543280303 Chr.6: 108043700-108043700 on Build GRCh38
rs550900778 Chr.6: 108043425-108043425 on Build GRCh38

View Full Product Details