Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 109377808-109378305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD164
Entrez Gene ID
8763
Gene Name
CD164 molecule
Gene Aliases
DFNA66, MGC-24, MUC-24, endolyn
UniGene
Hs.520313
Gene Chromosome Location
Chr.6: 109366514-109382559 on Build GRCh38
Transcripts
NM_001142401.1 NM_001142402.1 NM_001142403.1 NM_001142404.1 NM_006016.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4670375 c.330C>T p.S110S Chr.6: 109377901-109377901 on Build GRCh38
4670374 c.330C>T p.S110S Chr.6: 109377901-109377901 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772397811 Chr.6: 109377867-109377867 on Build GRCh38
rs1033167040 Chr.6: 109378053-109378053 on Build GRCh38
rs1407620 Chr.6: 109378156-109378156 on Build GRCh38
rs750585578 Chr.6: 109377995-109377995 on Build GRCh38
rs937850340 Chr.6: 109378153-109378153 on Build GRCh38
rs989668698 Chr.6: 109378142-109378142 on Build GRCh38
rs750068380 Chr.6: 109377965-109377965 on Build GRCh38
rs550218226 Chr.6: 109377850-109377850 on Build GRCh38
rs889252306 Chr.6: 109378194-109378194 on Build GRCh38
rs772751932 Chr.6: 109377971-109377971 on Build GRCh38
rs867957005 Chr.6: 109378171-109378171 on Build GRCh38
rs761186778 Chr.6: 109378096-109378096 on Build GRCh38
rs765986775 Chr.6: 109377911-109377911 on Build GRCh38
rs1005758664 Chr.6: 109378260-109378260 on Build GRCh38
rs76182593 Chr.6: 109378172-109378172 on Build GRCh38
rs952680211 Chr.6: 109378039-109378039 on Build GRCh38
rs768219295 Chr.6: 109377931-109377940 on Build GRCh38
rs936876950 Chr.6: 109378139-109378139 on Build GRCh38
rs377505131 Chr.6: 109377889-109377889 on Build GRCh38
rs892130916 Chr.6: 109377953-109377953 on Build GRCh38
rs772863146 Chr.6: 109377989-109377990 on Build GRCh38
rs747219785 Chr.6: 109378023-109378023 on Build GRCh38
rs373118837 Chr.6: 109377912-109377912 on Build GRCh38
rs761093679 Chr.6: 109377888-109377888 on Build GRCh38
rs966301141 Chr.6: 109378275-109378276 on Build GRCh38
rs570090603 Chr.6: 109378152-109378152 on Build GRCh38
rs369971930 Chr.6: 109378102-109378102 on Build GRCh38
rs370093477 Chr.6: 109377975-109377975 on Build GRCh38
rs558024264 Chr.6: 109377985-109377985 on Build GRCh38
rs137918186 Chr.6: 109378105-109378105 on Build GRCh38
rs752639259 Chr.6: 109377928-109377928 on Build GRCh38
rs918941528 Chr.6: 109378127-109378127 on Build GRCh38
rs1056751209 Chr.6: 109378156-109378156 on Build GRCh38
rs199887608 Chr.6: 109377901-109377901 on Build GRCh38
rs539893370 Chr.6: 109377849-109377849 on Build GRCh38
rs757764538 Chr.6: 109377979-109377979 on Build GRCh38
rs767293168 Chr.6: 109377981-109377981 on Build GRCh38
rs761656185 Chr.6: 109377983-109377983 on Build GRCh38
rs926436109 Chr.6: 109378149-109378149 on Build GRCh38
rs142043630 Chr.6: 109377900-109377900 on Build GRCh38
rs945327888 Chr.6: 109378157-109378157 on Build GRCh38
rs1000253734 Chr.6: 109378182-109378182 on Build GRCh38
rs745909701 Chr.6: 109377945-109377945 on Build GRCh38
rs1049974731 Chr.6: 109378220-109378220 on Build GRCh38
rs1033096652 Chr.6: 109378193-109378193 on Build GRCh38
rs772874526 Chr.6: 109377902-109377902 on Build GRCh38
rs779173634 Chr.6: 109378026-109378026 on Build GRCh38
rs35452836 Chr.6: 109378243-109378243 on Build GRCh38
rs144462229 Chr.6: 109378020-109378020 on Build GRCh38
rs773985578 Chr.6: 109377978-109377978 on Build GRCh38
rs767360953 Chr.6: 109377925-109377925 on Build GRCh38
rs900899916 Chr.6: 109378017-109378017 on Build GRCh38
rs550279584 Chr.6: 109378134-109378134 on Build GRCh38
rs1007721823 Chr.6: 109378209-109378211 on Build GRCh38
rs184774511 Chr.6: 109378080-109378080 on Build GRCh38
rs370521912 Chr.6: 109378029-109378029 on Build GRCh38
rs115860166 Chr.6: 109378072-109378072 on Build GRCh38
rs972754525 Chr.6: 109378275-109378275 on Build GRCh38
rs953542788 Chr.6: 109378069-109378069 on Build GRCh38
rs779363125 Chr.6: 109378166-109378166 on Build GRCh38
rs542601850 Chr.6: 109378062-109378062 on Build GRCh38
rs945647563 Chr.6: 109378163-109378163 on Build GRCh38
rs926805027 Chr.6: 109378109-109378109 on Build GRCh38
rs192734268 Chr.6: 109377962-109377962 on Build GRCh38
rs779533936 Chr.6: 109378094-109378094 on Build GRCh38
rs369727634 Chr.6: 109377866-109377866 on Build GRCh38
rs551556265 Chr.6: 109378082-109378082 on Build GRCh38
rs556025572 Chr.6: 109377955-109377955 on Build GRCh38
rs1014266553 Chr.6: 109377993-109377993 on Build GRCh38
rs759997740 Chr.6: 109378115-109378117 on Build GRCh38
rs757430967 Chr.6: 109377936-109377936 on Build GRCh38
rs376261079 Chr.6: 109377968-109377968 on Build GRCh38
rs1048861219 Chr.6: 109377956-109377956 on Build GRCh38
rs1015443926 Chr.6: 109378276-109378276 on Build GRCh38
rs151149269 Chr.6: 109377954-109377954 on Build GRCh38
rs748825585 Chr.6: 109377883-109377892 on Build GRCh38
rs375609614 Chr.6: 109378013-109378013 on Build GRCh38
rs926149812 Chr.6: 109377859-109377859 on Build GRCh38
rs779179297 Chr.6: 109377943-109377943 on Build GRCh38
rs754742080 Chr.6: 109377916-109377916 on Build GRCh38
rs577340749 Chr.6: 109377986-109377986 on Build GRCh38
rs867717851 Chr.6: 109377890-109377890 on Build GRCh38
rs562978507 Chr.6: 109378045-109378045 on Build GRCh38
rs767405702 Chr.6: 109377987-109377988 on Build GRCh38
rs758757315 Chr.6: 109377952-109377952 on Build GRCh38
rs114995468 Chr.6: 109377842-109377842 on Build GRCh38
rs762644274 Chr.6: 109377910-109377910 on Build GRCh38
rs754180518 Chr.6: 109377976-109377977 on Build GRCh38
rs573280599 Chr.6: 109378050-109378050 on Build GRCh38
rs201362068 Chr.6: 109377865-109377865 on Build GRCh38
rs566677642 Chr.6: 109378098-109378098 on Build GRCh38
rs374139168 Chr.6: 109377887-109377887 on Build GRCh38
rs752409583 Chr.6: 109377984-109377984 on Build GRCh38
rs978917538 Chr.6: 109377857-109377857 on Build GRCh38

View Full Product Details