Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 111206625-111207167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC16A10
Entrez Gene ID
117247
Gene Name
solute carrier family 16 member 10
Gene Aliases
MCT10, PRO0813, TAT1
UniGene
Hs.591327
Gene Chromosome Location
Chr.6: 111087503-111227125 on Build GRCh38
Transcripts
NM_018593.4 XM_017010237.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3857530 c.1009G>A p.V337I Chr.6: 111206658-111206658 on Build GRCh38
1329354 c.1066G>T p.V356L Chr.6: 111206715-111206715 on Build GRCh38
4760135 c.1046T>A p.I349N Chr.6: 111206695-111206695 on Build GRCh38
5941275 c.1047T>G p.I349M Chr.6: 111206696-111206696 on Build GRCh38
1744377 c.1076T>A p.V359D Chr.6: 111206725-111206725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761548040 Chr.6: 111207082-111207082 on Build GRCh38
rs935226490 Chr.6: 111206851-111206851 on Build GRCh38
rs933065412 Chr.6: 111206884-111206884 on Build GRCh38
rs779796347 Chr.6: 111206713-111206713 on Build GRCh38
rs768489003 Chr.6: 111206747-111206747 on Build GRCh38
rs769583773 Chr.6: 111206757-111206757 on Build GRCh38
rs769339670 Chr.6: 111206717-111206719 on Build GRCh38
rs910398123 Chr.6: 111206921-111206921 on Build GRCh38
rs34277174 Chr.6: 111206756-111206756 on Build GRCh38
rs749263323 Chr.6: 111206770-111206771 on Build GRCh38
rs529244901 Chr.6: 111206877-111206877 on Build GRCh38
rs1003619492 Chr.6: 111207021-111207021 on Build GRCh38
rs1004428650 Chr.6: 111206975-111206975 on Build GRCh38
rs780553509 Chr.6: 111206751-111206751 on Build GRCh38
rs931389245 Chr.6: 111207137-111207137 on Build GRCh38
rs12191266 Chr.6: 111206838-111206838 on Build GRCh38
rs758857738 Chr.6: 111206704-111206704 on Build GRCh38
rs762604005 Chr.6: 111206773-111206773 on Build GRCh38
rs200813678 Chr.6: 111206691-111206691 on Build GRCh38
rs550373736 Chr.6: 111207023-111207023 on Build GRCh38
rs375867887 Chr.6: 111206777-111206777 on Build GRCh38
rs757687123 Chr.6: 111206686-111206686 on Build GRCh38
rs899045618 Chr.6: 111206974-111206974 on Build GRCh38
rs402940 Chr.6: 111206785-111206785 on Build GRCh38
rs925079134 Chr.6: 111207041-111207041 on Build GRCh38
rs771755146 Chr.6: 111206891-111206891 on Build GRCh38
rs750713853 Chr.6: 111206692-111206692 on Build GRCh38
rs12197525 Chr.6: 111207018-111207018 on Build GRCh38
rs548787695 Chr.6: 111207096-111207096 on Build GRCh38
rs975770954 Chr.6: 111206994-111206994 on Build GRCh38
rs766560553 Chr.6: 111207101-111207101 on Build GRCh38
rs433569 Chr.6: 111206905-111206905 on Build GRCh38
rs528316312 Chr.6: 111206973-111206973 on Build GRCh38
rs959551612 Chr.6: 111207062-111207062 on Build GRCh38
rs202085593 Chr.6: 111206676-111206676 on Build GRCh38
rs760596829 Chr.6: 111206923-111206923 on Build GRCh38
rs1027123861 Chr.6: 111206955-111206955 on Build GRCh38
rs1027671028 Chr.6: 111206718-111206718 on Build GRCh38
rs551164167 Chr.6: 111206977-111206977 on Build GRCh38
rs564620274 Chr.6: 111206803-111206803 on Build GRCh38
rs949441058 Chr.6: 111207079-111207079 on Build GRCh38
rs1014573182 Chr.6: 111206750-111206750 on Build GRCh38
rs113965591 Chr.6: 111206684-111206684 on Build GRCh38
rs139780581 Chr.6: 111207007-111207007 on Build GRCh38
rs559377717 Chr.6: 111206969-111206969 on Build GRCh38
rs770719551 Chr.6: 111206775-111206775 on Build GRCh38
rs754012515 Chr.6: 111206677-111206677 on Build GRCh38
rs966925922 Chr.6: 111207138-111207138 on Build GRCh38
rs773230693 Chr.6: 111206766-111206766 on Build GRCh38
rs965736148 Chr.6: 111206985-111206985 on Build GRCh38
rs760684404 Chr.6: 111206658-111206658 on Build GRCh38
rs1034153546 Chr.6: 111207056-111207056 on Build GRCh38
rs372191084 Chr.6: 111206738-111206738 on Build GRCh38
rs749623985 Chr.6: 111206689-111206689 on Build GRCh38
rs567280441 Chr.6: 111207139-111207139 on Build GRCh38
rs935259063 Chr.6: 111207059-111207059 on Build GRCh38
rs905259879 Chr.6: 111206922-111206922 on Build GRCh38
rs181746758 Chr.6: 111206657-111206657 on Build GRCh38
rs186296060 Chr.6: 111206870-111206870 on Build GRCh38
rs775341828 Chr.6: 111206750-111206751 on Build GRCh38
rs1010991614 Chr.6: 111207076-111207076 on Build GRCh38
rs543512600 Chr.6: 111206859-111206859 on Build GRCh38
rs979276469 Chr.6: 111206819-111206820 on Build GRCh38
rs868682380 Chr.6: 111206778-111206778 on Build GRCh38
rs996104852 Chr.6: 111206953-111206953 on Build GRCh38
rs943159252 Chr.6: 111206965-111206965 on Build GRCh38

View Full Product Details