Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 4682204-4682698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXK1
Entrez Gene ID
221937
Gene Name
forkhead box K1
Gene Aliases
FOXK1L
UniGene
Hs.487393
Gene Chromosome Location
Chr.7: 4644691-4771443 on Build GRCh38
Transcripts
NM_001037165.1 XM_011515191.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4985370 c.202G>A p.A68T Chr.7: 4682510-4682510 on Build GRCh38
4999024 c.384C>G p.N128K Chr.7: 4682692-4682692 on Build GRCh38
1622894 c.39G>A p.L13L Chr.7: 4682347-4682347 on Build GRCh38
1622895 c.301A>T p.S101C Chr.7: 4682609-4682609 on Build GRCh38
4164361 c.103C>G p.P35A Chr.7: 4682411-4682411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041146684 Chr.7: 4682613-4682613 on Build GRCh38
rs761489465 Chr.7: 4682669-4682669 on Build GRCh38
rs1014605010 Chr.7: 4682488-4682488 on Build GRCh38
rs951837792 Chr.7: 4682284-4682284 on Build GRCh38
rs991326472 Chr.7: 4682536-4682536 on Build GRCh38
rs1027159877 Chr.7: 4682460-4682461 on Build GRCh38
rs940469438 Chr.7: 4682313-4682313 on Build GRCh38
rs914431746 Chr.7: 4682540-4682540 on Build GRCh38
rs113685723 Chr.7: 4682367-4682367 on Build GRCh38
rs903273207 Chr.7: 4682254-4682254 on Build GRCh38
rs1037482648 Chr.7: 4682246-4682246 on Build GRCh38
rs937253839 Chr.7: 4682235-4682235 on Build GRCh38
rs940209733 Chr.7: 4682243-4682243 on Build GRCh38
rs543100102 Chr.7: 4682537-4682537 on Build GRCh38
rs1041719660 Chr.7: 4682423-4682423 on Build GRCh38
rs893132537 Chr.7: 4682386-4682387 on Build GRCh38
rs1019756747 Chr.7: 4682273-4682273 on Build GRCh38
rs762878185 Chr.7: 4682619-4682619 on Build GRCh38
rs907577369 Chr.7: 4682292-4682292 on Build GRCh38
rs981210413 Chr.7: 4682506-4682506 on Build GRCh38
rs952995240 Chr.7: 4682479-4682479 on Build GRCh38
rs947339952 Chr.7: 4682548-4682548 on Build GRCh38
rs984482474 Chr.7: 4682291-4682291 on Build GRCh38
rs13235043 Chr.7: 4682469-4682469 on Build GRCh38
rs924660756 Chr.7: 4682610-4682610 on Build GRCh38
rs958560661 Chr.7: 4682533-4682533 on Build GRCh38
rs928481061 Chr.7: 4682329-4682329 on Build GRCh38
rs560718579 Chr.7: 4682323-4682323 on Build GRCh38
rs1020820525 Chr.7: 4682247-4682247 on Build GRCh38
rs1042100046 Chr.7: 4682565-4682565 on Build GRCh38
rs997048138 Chr.7: 4682439-4682439 on Build GRCh38
rs112998729 Chr.7: 4682377-4682377 on Build GRCh38
rs895895899 Chr.7: 4682380-4682380 on Build GRCh38
rs975662447 Chr.7: 4682276-4682276 on Build GRCh38
rs1008790887 Chr.7: 4682264-4682264 on Build GRCh38
rs1019724196 Chr.7: 4682434-4682434 on Build GRCh38
rs767128784 Chr.7: 4682673-4682673 on Build GRCh38
rs775282631 Chr.7: 4682612-4682612 on Build GRCh38
rs763811279 Chr.7: 4682653-4682653 on Build GRCh38
rs893165046 Chr.7: 4682239-4682239 on Build GRCh38
rs760812037 Chr.7: 4682435-4682435 on Build GRCh38
rs964252043 Chr.7: 4682274-4682274 on Build GRCh38
rs937222554 Chr.7: 4682350-4682350 on Build GRCh38
rs769906305 Chr.7: 4682611-4682611 on Build GRCh38
rs962087329 Chr.7: 4682503-4682508 on Build GRCh38
rs984840406 Chr.7: 4682482-4682482 on Build GRCh38
rs751322767 Chr.7: 4682657-4682657 on Build GRCh38
rs1008341876 Chr.7: 4682430-4682430 on Build GRCh38
rs866354401 Chr.7: 4682240-4682240 on Build GRCh38
rs574086514 Chr.7: 4682510-4682510 on Build GRCh38
rs1028614709 Chr.7: 4682280-4682280 on Build GRCh38
rs903239836 Chr.7: 4682426-4682426 on Build GRCh38
rs531802798 Chr.7: 4682615-4682615 on Build GRCh38
rs947340031 Chr.7: 4682392-4682393 on Build GRCh38

View Full Product Details