Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6608281-6608796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf26
Entrez Gene ID
79034
Gene Name
chromosome 7 open reading frame 26
Gene Aliases
-
UniGene
Hs.487511
Gene Chromosome Location
Chr.7: 6589368-6608726 on Build GRCh38
Transcripts
NM_001303039.1 NM_024067.3 XM_005249845.4 XM_017012614.1 XM_017012615.1

SNP Detail

SNP ID Chromosome Location
rs898520129 Chr.7: 6608365-6608365 on Build GRCh38
rs936511776 Chr.7: 6608541-6608541 on Build GRCh38
rs1008736356 Chr.7: 6608435-6608435 on Build GRCh38
rs892327126 Chr.7: 6608690-6608690 on Build GRCh38
rs544182359 Chr.7: 6608436-6608436 on Build GRCh38
rs772322363 Chr.7: 6608527-6608527 on Build GRCh38
rs191615080 Chr.7: 6608327-6608327 on Build GRCh38
rs914959244 Chr.7: 6608460-6608460 on Build GRCh38
rs969810851 Chr.7: 6608768-6608768 on Build GRCh38
rs1008806819 Chr.7: 6608678-6608678 on Build GRCh38
rs953198200 Chr.7: 6608370-6608370 on Build GRCh38
rs527863460 Chr.7: 6608664-6608664 on Build GRCh38
rs150276869 Chr.7: 6608441-6608441 on Build GRCh38
rs992210003 Chr.7: 6608486-6608486 on Build GRCh38
rs529754090 Chr.7: 6608480-6608480 on Build GRCh38
rs1050198263 Chr.7: 6608644-6608644 on Build GRCh38
rs992827891 Chr.7: 6608736-6608736 on Build GRCh38
rs997267362 Chr.7: 6608389-6608389 on Build GRCh38
rs932711597 Chr.7: 6608642-6608642 on Build GRCh38
rs1052253051 Chr.7: 6608566-6608566 on Build GRCh38
rs1028302408 Chr.7: 6608390-6608390 on Build GRCh38
rs1021784433 Chr.7: 6608582-6608582 on Build GRCh38
rs540455251 Chr.7: 6608315-6608315 on Build GRCh38
rs370010311 Chr.7: 6608525-6608525 on Build GRCh38
rs968207593 Chr.7: 6608721-6608721 on Build GRCh38
rs775408556 Chr.7: 6608310-6608310 on Build GRCh38
rs981275546 Chr.7: 6608526-6608526 on Build GRCh38
rs1038858429 Chr.7: 6608359-6608359 on Build GRCh38
rs753728755 Chr.7: 6608640-6608640 on Build GRCh38
rs1007198748 Chr.7: 6608641-6608641 on Build GRCh38
rs940152172 Chr.7: 6608584-6608584 on Build GRCh38
rs980862585 Chr.7: 6608511-6608511 on Build GRCh38
rs1025193847 Chr.7: 6608752-6608752 on Build GRCh38
rs188846691 Chr.7: 6608614-6608614 on Build GRCh38
rs761478935 Chr.7: 6608705-6608705 on Build GRCh38
rs1015981717 Chr.7: 6608377-6608377 on Build GRCh38
rs1022107147 Chr.7: 6608718-6608718 on Build GRCh38
rs568878699 Chr.7: 6608715-6608715 on Build GRCh38
rs933223196 Chr.7: 6608589-6608589 on Build GRCh38
rs184219743 Chr.7: 6608342-6608342 on Build GRCh38
rs1025763978 Chr.7: 6608709-6608709 on Build GRCh38
rs371448855 Chr.7: 6608451-6608451 on Build GRCh38
rs951510357 Chr.7: 6608728-6608728 on Build GRCh38
rs956538450 Chr.7: 6608447-6608447 on Build GRCh38
rs939590035 Chr.7: 6608528-6608528 on Build GRCh38
rs548894652 Chr.7: 6608567-6608567 on Build GRCh38
rs746299506 Chr.7: 6608505-6608505 on Build GRCh38
rs1016446721 Chr.7: 6608652-6608652 on Build GRCh38
rs796876933 Chr.7: 6608496-6608496 on Build GRCh38
rs953300440 Chr.7: 6608393-6608393 on Build GRCh38
rs1007725935 Chr.7: 6608375-6608375 on Build GRCh38
rs1018797064 Chr.7: 6608682-6608682 on Build GRCh38
rs1056723370 Chr.7: 6608551-6608551 on Build GRCh38
rs892424391 Chr.7: 6608583-6608583 on Build GRCh38
rs925332255 Chr.7: 6608530-6608530 on Build GRCh38

View Full Product Details