Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 12230272-12230778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM106B
Entrez Gene ID
54664
Gene Name
transmembrane protein 106B
Gene Aliases
-
UniGene
Hs.396358
Gene Chromosome Location
Chr.7: 12211222-12237264 on Build GRCh38
Transcripts
NM_001134232.1 NM_018374.3 XM_005249789.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1447663 c.583-1G>A p.? Chr.7: 12230388-12230388 on Build GRCh38
6108297 c.590A>G p.Y197C Chr.7: 12230396-12230396 on Build GRCh38
5737870 c.604G>A p.V202I Chr.7: 12230410-12230410 on Build GRCh38
6803085 c.603C>T p.T201T Chr.7: 12230409-12230409 on Build GRCh38
6108296 c.627T>C p.Y209Y Chr.7: 12230433-12230433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370105760 Chr.7: 12230445-12230445 on Build GRCh38
rs944768367 Chr.7: 12230747-12230747 on Build GRCh38
rs757561026 Chr.7: 12230449-12230449 on Build GRCh38
rs202102292 Chr.7: 12230410-12230410 on Build GRCh38
rs771468165 Chr.7: 12230405-12230405 on Build GRCh38
rs185813432 Chr.7: 12230716-12230716 on Build GRCh38
rs188232679 Chr.7: 12230471-12230471 on Build GRCh38
rs1036106573 Chr.7: 12230311-12230311 on Build GRCh38
rs767438998 Chr.7: 12230454-12230454 on Build GRCh38
rs374151535 Chr.7: 12230375-12230375 on Build GRCh38
rs1043329505 Chr.7: 12230687-12230687 on Build GRCh38
rs144098183 Chr.7: 12230605-12230605 on Build GRCh38
rs189937008 Chr.7: 12230750-12230750 on Build GRCh38
rs749702450 Chr.7: 12230391-12230391 on Build GRCh38
rs770474045 Chr.7: 12230482-12230482 on Build GRCh38
rs959349759 Chr.7: 12230637-12230637 on Build GRCh38
rs767450918 Chr.7: 12230752-12230752 on Build GRCh38
rs556183053 Chr.7: 12230527-12230527 on Build GRCh38
rs769191545 Chr.7: 12230399-12230399 on Build GRCh38
rs574563818 Chr.7: 12230664-12230664 on Build GRCh38
rs961245130 Chr.7: 12230327-12230327 on Build GRCh38
rs200343375 Chr.7: 12230483-12230483 on Build GRCh38
rs757366288 Chr.7: 12230491-12230497 on Build GRCh38
rs923992795 Chr.7: 12230486-12230486 on Build GRCh38
rs1005190635 Chr.7: 12230589-12230589 on Build GRCh38
rs996824472 Chr.7: 12230528-12230528 on Build GRCh38
rs1040919316 Chr.7: 12230500-12230500 on Build GRCh38
rs998789349 Chr.7: 12230718-12230718 on Build GRCh38
rs968028815 Chr.7: 12230720-12230720 on Build GRCh38
rs1049462662 Chr.7: 12230575-12230575 on Build GRCh38
rs765061839 Chr.7: 12230434-12230434 on Build GRCh38
rs756539391 Chr.7: 12230459-12230459 on Build GRCh38
rs1011525044 Chr.7: 12230675-12230675 on Build GRCh38
rs540391943 Chr.7: 12230684-12230684 on Build GRCh38
rs34644075 Chr.7: 12230602-12230603 on Build GRCh38
rs888942410 Chr.7: 12230633-12230636 on Build GRCh38
rs780730635 Chr.7: 12230461-12230461 on Build GRCh38
rs1035250915 Chr.7: 12230620-12230620 on Build GRCh38
rs181999404 Chr.7: 12230498-12230498 on Build GRCh38
rs776358184 Chr.7: 12230420-12230420 on Build GRCh38
rs557263817 Chr.7: 12230458-12230460 on Build GRCh38
rs757256679 Chr.7: 12230366-12230366 on Build GRCh38
rs142177964 Chr.7: 12230433-12230433 on Build GRCh38
rs539448274 Chr.7: 12230526-12230526 on Build GRCh38
rs888116824 Chr.7: 12230588-12230588 on Build GRCh38
rs537783464 Chr.7: 12230617-12230617 on Build GRCh38
rs781234796 Chr.7: 12230491-12230492 on Build GRCh38
rs371037357 Chr.7: 12230629-12230629 on Build GRCh38
rs554822299 Chr.7: 12230628-12230628 on Build GRCh38
rs758967500 Chr.7: 12230436-12230438 on Build GRCh38
rs1050305389 Chr.7: 12230631-12230631 on Build GRCh38
rs374626555 Chr.7: 12230456-12230456 on Build GRCh38
rs370919981 Chr.7: 12230479-12230479 on Build GRCh38
rs961397326 Chr.7: 12230611-12230611 on Build GRCh38
rs765634930 Chr.7: 12230360-12230364 on Build GRCh38
rs751833626 Chr.7: 12230353-12230353 on Build GRCh38
rs751582621 Chr.7: 12230476-12230479 on Build GRCh38
rs991642361 Chr.7: 12230499-12230499 on Build GRCh38
rs984778245 Chr.7: 12230532-12230532 on Build GRCh38
rs367778603 Chr.7: 12230409-12230409 on Build GRCh38
rs371105661 Chr.7: 12230372-12230372 on Build GRCh38
rs753167226 Chr.7: 12230362-12230363 on Build GRCh38
rs568587185 Chr.7: 12230469-12230469 on Build GRCh38
rs747696215 Chr.7: 12230473-12230473 on Build GRCh38
rs780499974 Chr.7: 12230386-12230386 on Build GRCh38
rs772554486 Chr.7: 12230477-12230477 on Build GRCh38
rs924046628 Chr.7: 12230745-12230745 on Build GRCh38
rs933154580 Chr.7: 12230630-12230632 on Build GRCh38
rs1029344810 Chr.7: 12230429-12230429 on Build GRCh38
rs978032941 Chr.7: 12230735-12230735 on Build GRCh38
rs560294825 Chr.7: 12230685-12230685 on Build GRCh38
rs903192578 Chr.7: 12230689-12230689 on Build GRCh38
rs376650754 Chr.7: 12230403-12230403 on Build GRCh38
rs562465661 Chr.7: 12230300-12230300 on Build GRCh38
rs376443768 Chr.7: 12230382-12230382 on Build GRCh38

View Full Product Details