Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 12622672-12623171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCIN
Entrez Gene ID
85477
Gene Name
scinderin
Gene Aliases
-
UniGene
Hs.633359
Gene Chromosome Location
Chr.7: 12570349-12653603 on Build GRCh38
Transcripts
NM_001112706.2 NM_033128.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5006140 c.747T>G p.A249A Chr.7: 12622881-12622881 on Build GRCh38
7121612 c.698G>T p.G233V Chr.7: 12622832-12622832 on Build GRCh38
1447792 c.671T>C p.L224S Chr.7: 12622805-12622805 on Build GRCh38
6250133 c.724G>T p.D242Y Chr.7: 12622858-12622858 on Build GRCh38
1085073 c.756C>T p.Y252Y Chr.7: 12622890-12622890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973646195 Chr.7: 12622867-12622867 on Build GRCh38
rs936511454 Chr.7: 12622971-12622971 on Build GRCh38
rs1053670900 Chr.7: 12622972-12622972 on Build GRCh38
rs185545930 Chr.7: 12622831-12622831 on Build GRCh38
rs189367241 Chr.7: 12622772-12622772 on Build GRCh38
rs1017778624 Chr.7: 12623126-12623126 on Build GRCh38
rs1009241001 Chr.7: 12622787-12622787 on Build GRCh38
rs769527317 Chr.7: 12622903-12622903 on Build GRCh38
rs771043555 Chr.7: 12622936-12622936 on Build GRCh38
rs758097506 Chr.7: 12622853-12622853 on Build GRCh38
rs773134258 Chr.7: 12622817-12622817 on Build GRCh38
rs573778243 Chr.7: 12623038-12623038 on Build GRCh38
rs956368273 Chr.7: 12622778-12622778 on Build GRCh38
rs752360184 Chr.7: 12622835-12622835 on Build GRCh38
rs745647324 Chr.7: 12622796-12622796 on Build GRCh38
rs1007307675 Chr.7: 12623116-12623116 on Build GRCh38
rs113337605 Chr.7: 12622925-12622925 on Build GRCh38
rs756435665 Chr.7: 12622742-12622742 on Build GRCh38
rs778701098 Chr.7: 12622756-12622756 on Build GRCh38
rs778137393 Chr.7: 12622766-12622766 on Build GRCh38
rs890722680 Chr.7: 12622926-12622926 on Build GRCh38
rs535720137 Chr.7: 12622833-12622833 on Build GRCh38
rs980696928 Chr.7: 12623136-12623136 on Build GRCh38
rs777534555 Chr.7: 12622870-12622870 on Build GRCh38
rs200560336 Chr.7: 12622907-12622907 on Build GRCh38
rs556075801 Chr.7: 12622887-12622887 on Build GRCh38
rs1018211354 Chr.7: 12622960-12622960 on Build GRCh38
rs754433636 Chr.7: 12622829-12622829 on Build GRCh38
rs745820414 Chr.7: 12622895-12622895 on Build GRCh38
rs372300943 Chr.7: 12622812-12622812 on Build GRCh38
rs528391704 Chr.7: 12623141-12623141 on Build GRCh38
rs563663871 Chr.7: 12623077-12623077 on Build GRCh38
rs80237547 Chr.7: 12622945-12622945 on Build GRCh38
rs776907741 Chr.7: 12622915-12622917 on Build GRCh38
rs760855325 Chr.7: 12622825-12622825 on Build GRCh38
rs780927464 Chr.7: 12622893-12622893 on Build GRCh38
rs890165197 Chr.7: 12623078-12623078 on Build GRCh38
rs948785550 Chr.7: 12622954-12622954 on Build GRCh38
rs189150209 Chr.7: 12622962-12622962 on Build GRCh38
rs769583257 Chr.7: 12622801-12622801 on Build GRCh38
rs985907213 Chr.7: 12622815-12622815 on Build GRCh38
rs375855538 Chr.7: 12622854-12622854 on Build GRCh38
rs193133695 Chr.7: 12622779-12622779 on Build GRCh38
rs774498088 Chr.7: 12622802-12622802 on Build GRCh38
rs762043563 Chr.7: 12622809-12622809 on Build GRCh38
rs766779402 Chr.7: 12622826-12622826 on Build GRCh38
rs373124306 Chr.7: 12623081-12623081 on Build GRCh38
rs949283545 Chr.7: 12622816-12622816 on Build GRCh38
rs1009208432 Chr.7: 12622982-12622982 on Build GRCh38
rs559187113 Chr.7: 12623079-12623079 on Build GRCh38
rs75745885 Chr.7: 12622978-12622978 on Build GRCh38
rs773536193 Chr.7: 12622920-12622920 on Build GRCh38
rs756859043 Chr.7: 12622892-12622892 on Build GRCh38
rs755449537 Chr.7: 12622751-12622751 on Build GRCh38
rs963914235 Chr.7: 12622847-12622847 on Build GRCh38
rs770752583 Chr.7: 12622776-12622776 on Build GRCh38
rs112416510 Chr.7: 12622963-12622963 on Build GRCh38
rs777722788 Chr.7: 12623100-12623101 on Build GRCh38
rs570584400 Chr.7: 12622760-12622760 on Build GRCh38
rs747819371 Chr.7: 12622757-12622757 on Build GRCh38

View Full Product Details