Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 15685803-15686300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEOX2
Entrez Gene ID
4223
Gene Name
mesenchyme homeobox 2
Gene Aliases
GAX, MOX2
UniGene
Hs.170355
Gene Chromosome Location
Chr.7: 15611212-15686683 on Build GRCh38
Transcripts
NM_005924.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4564013 c.133_134TC>AT p.S45I Chr.7: 15686269-15686270 on Build GRCh38
4701749 c.482A>T p.K161M Chr.7: 15685921-15685921 on Build GRCh38
1449680 c.353delC p.P118fs*98 Chr.7: 15686050-15686050 on Build GRCh38
1449682 c.231T>C p.H77H Chr.7: 15686172-15686172 on Build GRCh38
3636762 c.440G>A p.G147E Chr.7: 15685963-15685963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767821407 Chr.7: 15685994-15685994 on Build GRCh38
rs376342964 Chr.7: 15685956-15685956 on Build GRCh38
rs756065472 Chr.7: 15686173-15686178 on Build GRCh38
rs370149136 Chr.7: 15686190-15686190 on Build GRCh38
rs893377581 Chr.7: 15686144-15686144 on Build GRCh38
rs940405249 Chr.7: 15686020-15686020 on Build GRCh38
rs766049663 Chr.7: 15686114-15686114 on Build GRCh38
rs373439702 Chr.7: 15685910-15685910 on Build GRCh38
rs769386764 Chr.7: 15685867-15685867 on Build GRCh38
rs771851933 Chr.7: 15686199-15686200 on Build GRCh38
rs745379359 Chr.7: 15685863-15685863 on Build GRCh38
rs773744356 Chr.7: 15686268-15686268 on Build GRCh38
rs372315372 Chr.7: 15686028-15686028 on Build GRCh38
rs777561137 Chr.7: 15686054-15686054 on Build GRCh38
rs774221299 Chr.7: 15685880-15685880 on Build GRCh38
rs201179928 Chr.7: 15686061-15686061 on Build GRCh38
rs746797445 Chr.7: 15686056-15686056 on Build GRCh38
rs780809317 Chr.7: 15686232-15686232 on Build GRCh38
rs141081461 Chr.7: 15686119-15686119 on Build GRCh38
rs755724547 Chr.7: 15686013-15686013 on Build GRCh38
rs778347553 Chr.7: 15686172-15686173 on Build GRCh38
rs932322308 Chr.7: 15686085-15686085 on Build GRCh38
rs577801614 Chr.7: 15685927-15685927 on Build GRCh38
rs146346021 Chr.7: 15686228-15686228 on Build GRCh38
rs924464418 Chr.7: 15686001-15686001 on Build GRCh38
rs746837361 Chr.7: 15686269-15686271 on Build GRCh38
rs199655872 Chr.7: 15686073-15686073 on Build GRCh38
rs780881826 Chr.7: 15685856-15685856 on Build GRCh38
rs775268162 Chr.7: 15685869-15685869 on Build GRCh38
rs747950584 Chr.7: 15685962-15685962 on Build GRCh38
rs750783408 Chr.7: 15685999-15685999 on Build GRCh38
rs528476147 Chr.7: 15686064-15686064 on Build GRCh38
rs534593518 Chr.7: 15685882-15685882 on Build GRCh38
rs895198118 Chr.7: 15686172-15686172 on Build GRCh38
rs751911345 Chr.7: 15685936-15685936 on Build GRCh38
rs576956999 Chr.7: 15685854-15685854 on Build GRCh38
rs753373068 Chr.7: 15686037-15686037 on Build GRCh38
rs773658129 Chr.7: 15686188-15686188 on Build GRCh38
rs772153901 Chr.7: 15686050-15686050 on Build GRCh38
rs759767365 Chr.7: 15685966-15685966 on Build GRCh38
rs772083188 Chr.7: 15686267-15686267 on Build GRCh38
rs1049360522 Chr.7: 15685988-15685988 on Build GRCh38
rs780233864 Chr.7: 15686198-15686198 on Build GRCh38
rs770045601 Chr.7: 15686057-15686057 on Build GRCh38
rs759113184 Chr.7: 15686164-15686175 on Build GRCh38
rs764063745 Chr.7: 15686226-15686226 on Build GRCh38
rs138296490 Chr.7: 15686231-15686231 on Build GRCh38
rs748219682 Chr.7: 15686266-15686266 on Build GRCh38
rs752755220 Chr.7: 15685919-15685919 on Build GRCh38
rs1003692562 Chr.7: 15686141-15686141 on Build GRCh38
rs775584752 Chr.7: 15685977-15685977 on Build GRCh38
rs765385637 Chr.7: 15685974-15685974 on Build GRCh38
rs757688209 Chr.7: 15685937-15685937 on Build GRCh38
rs747381134 Chr.7: 15685852-15685852 on Build GRCh38
rs758802174 Chr.7: 15686173-15686184 on Build GRCh38
rs764582682 Chr.7: 15685986-15685986 on Build GRCh38
rs544898693 Chr.7: 15686200-15686200 on Build GRCh38
rs373584566 Chr.7: 15686090-15686090 on Build GRCh38
rs777632205 Chr.7: 15686135-15686135 on Build GRCh38
rs1037275055 Chr.7: 15686125-15686125 on Build GRCh38
rs748140998 Chr.7: 15686046-15686046 on Build GRCh38
rs906709293 Chr.7: 15686132-15686132 on Build GRCh38
rs758050775 Chr.7: 15686221-15686221 on Build GRCh38
rs758998673 Chr.7: 15686066-15686066 on Build GRCh38
rs551036356 Chr.7: 15686195-15686195 on Build GRCh38
rs201879229 Chr.7: 15685931-15685931 on Build GRCh38
rs762144203 Chr.7: 15685990-15685990 on Build GRCh38
rs753283129 Chr.7: 15685847-15685847 on Build GRCh38
rs756848135 Chr.7: 15685853-15685853 on Build GRCh38
rs202079444 Chr.7: 15685952-15685952 on Build GRCh38
rs778960612 Chr.7: 15686045-15686045 on Build GRCh38
rs148170403 Chr.7: 15685982-15685982 on Build GRCh38
rs769939650 Chr.7: 15686195-15686198 on Build GRCh38
rs902106638 Chr.7: 15685842-15685842 on Build GRCh38
rs879008489 Chr.7: 15685984-15685984 on Build GRCh38
rs903273703 Chr.7: 15686178-15686178 on Build GRCh38
rs758357349 Chr.7: 15686131-15686131 on Build GRCh38
rs750338773 Chr.7: 15686176-15686178 on Build GRCh38
rs753233317 Chr.7: 15686148-15686149 on Build GRCh38
rs370719073 Chr.7: 15686150-15686150 on Build GRCh38
rs772029318 Chr.7: 15685963-15685963 on Build GRCh38
rs779584447 Chr.7: 15686016-15686016 on Build GRCh38
rs746818920 Chr.7: 15686149-15686149 on Build GRCh38
rs575381387 Chr.7: 15686041-15686041 on Build GRCh38
rs372066707 Chr.7: 15686200-15686202 on Build GRCh38
rs748751582 Chr.7: 15685951-15685951 on Build GRCh38
rs200609553 Chr.7: 15686249-15686249 on Build GRCh38
rs761067039 Chr.7: 15686189-15686189 on Build GRCh38
rs773053518 Chr.7: 15686201-15686202 on Build GRCh38
rs746685679 Chr.7: 15685846-15685846 on Build GRCh38
rs141784602 Chr.7: 15685913-15685913 on Build GRCh38
rs143756690 Chr.7: 15685944-15685944 on Build GRCh38
rs781724548 Chr.7: 15686192-15686193 on Build GRCh38
rs768507132 Chr.7: 15685877-15685877 on Build GRCh38
rs780793670 Chr.7: 15685938-15685938 on Build GRCh38
rs907684119 Chr.7: 15686276-15686276 on Build GRCh38
rs991895325 Chr.7: 15686222-15686222 on Build GRCh38
rs758856274 Chr.7: 15685850-15685850 on Build GRCh38
rs545808518 Chr.7: 15686055-15686055 on Build GRCh38
rs529311797 Chr.7: 15686191-15686191 on Build GRCh38
rs745896587 Chr.7: 15686195-15686196 on Build GRCh38
rs761000362 Chr.7: 15686113-15686113 on Build GRCh38
rs561867178 Chr.7: 15686105-15686105 on Build GRCh38
rs755133987 Chr.7: 15686242-15686242 on Build GRCh38
rs779577335 Chr.7: 15685945-15685945 on Build GRCh38
rs749200538 Chr.7: 15685875-15685875 on Build GRCh38
rs141947578 Chr.7: 15686214-15686214 on Build GRCh38
rs768897912 Chr.7: 15686174-15686174 on Build GRCh38
rs745725675 Chr.7: 15686241-15686241 on Build GRCh38
rs372077660 Chr.7: 15686259-15686259 on Build GRCh38
rs906697453 Chr.7: 15685901-15685901 on Build GRCh38
rs138679259 Chr.7: 15686198-15686198 on Build GRCh38
rs374499365 Chr.7: 15686202-15686202 on Build GRCh38
rs765404239 Chr.7: 15685915-15685915 on Build GRCh38
rs563305943 Chr.7: 15685965-15685965 on Build GRCh38
rs201748928 Chr.7: 15686130-15686130 on Build GRCh38
rs761894513 Chr.7: 15686176-15686176 on Build GRCh38
rs199751527 Chr.7: 15686181-15686181 on Build GRCh38
rs757023123 Chr.7: 15685881-15685881 on Build GRCh38
rs774437329 Chr.7: 15686080-15686080 on Build GRCh38
rs1048013657 Chr.7: 15686100-15686100 on Build GRCh38
rs369977466 Chr.7: 15685862-15685862 on Build GRCh38
rs147797539 Chr.7: 15685911-15685911 on Build GRCh38
rs113582077 Chr.7: 15686173-15686175 on Build GRCh38
rs752096687 Chr.7: 15686173-15686181 on Build GRCh38
rs977364770 Chr.7: 15686000-15686000 on Build GRCh38
rs965227479 Chr.7: 15686032-15686032 on Build GRCh38
rs774347485 Chr.7: 15686175-15686175 on Build GRCh38
rs753470533 Chr.7: 15686115-15686115 on Build GRCh38
rs373826169 Chr.7: 15686151-15686151 on Build GRCh38
rs35244570 Chr.7: 15685835-15685835 on Build GRCh38
rs780513538 Chr.7: 15686271-15686271 on Build GRCh38

View Full Product Details