Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 86838720-86839223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRM3
Entrez Gene ID
2913
Gene Name
glutamate metabotropic receptor 3
Gene Aliases
GLUR3, GPRC1C, MGLUR3, mGlu3
UniGene
Hs.590575
Gene Chromosome Location
Chr.7: 86643914-86864879 on Build GRCh38
Transcripts
NM_000840.2 XM_011516088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
141846 c.1682G>A p.G561E Chr.7: 86839196-86839196 on Build GRCh38
7223106 c.1010G>T p.R337L Chr.7: 86838908-86838908 on Build GRCh38
3924146 c.1598C>T p.P533L Chr.7: 86839112-86839112 on Build GRCh38
4526190 c.1379G>A p.G460E Chr.7: 86838893-86838893 on Build GRCh38
747783 c.1423G>A p.G475S Chr.7: 86838937-86838937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750691453 Chr.7: 86838916-86838916 on Build GRCh38
rs758853251 Chr.7: 86838923-86838923 on Build GRCh38
rs1012817006 Chr.7: 86838897-86838897 on Build GRCh38
rs781189896 Chr.7: 86838960-86838960 on Build GRCh38
rs745682319 Chr.7: 86838817-86838817 on Build GRCh38
rs755321060 Chr.7: 86839055-86839055 on Build GRCh38
rs757078535 Chr.7: 86838986-86838986 on Build GRCh38
rs1053369838 Chr.7: 86838876-86838876 on Build GRCh38
rs759963146 Chr.7: 86838809-86838810 on Build GRCh38
rs373668140 Chr.7: 86838787-86838787 on Build GRCh38
rs777825537 Chr.7: 86838795-86838795 on Build GRCh38
rs974918264 Chr.7: 86838794-86838794 on Build GRCh38
rs536332070 Chr.7: 86838980-86838980 on Build GRCh38
rs766121666 Chr.7: 86839008-86839008 on Build GRCh38
rs373913639 Chr.7: 86838904-86838904 on Build GRCh38
rs367616956 Chr.7: 86838801-86838801 on Build GRCh38
rs745602111 Chr.7: 86838998-86838998 on Build GRCh38
rs773795841 Chr.7: 86839042-86839042 on Build GRCh38
rs746808957 Chr.7: 86839143-86839143 on Build GRCh38
rs765608538 Chr.7: 86839031-86839031 on Build GRCh38
rs17856664 Chr.7: 86839048-86839048 on Build GRCh38
rs527768281 Chr.7: 86839044-86839044 on Build GRCh38
rs552553702 Chr.7: 86839141-86839141 on Build GRCh38
rs758174717 Chr.7: 86839125-86839125 on Build GRCh38
rs755143192 Chr.7: 86838791-86838791 on Build GRCh38
rs17852048 Chr.7: 86838850-86838850 on Build GRCh38
rs371949904 Chr.7: 86839068-86839068 on Build GRCh38
rs199660204 Chr.7: 86838881-86838881 on Build GRCh38
rs765776830 Chr.7: 86838915-86838915 on Build GRCh38
rs747567892 Chr.7: 86839164-86839164 on Build GRCh38
rs754080083 Chr.7: 86838950-86838950 on Build GRCh38
rs771892863 Chr.7: 86838999-86838999 on Build GRCh38
rs756240686 Chr.7: 86839075-86839075 on Build GRCh38
rs767785844 Chr.7: 86839056-86839056 on Build GRCh38
rs1019385107 Chr.7: 86839126-86839126 on Build GRCh38
rs267601604 Chr.7: 86839045-86839045 on Build GRCh38
rs763641866 Chr.7: 86838872-86838872 on Build GRCh38
rs17161026 Chr.7: 86838938-86838938 on Build GRCh38
rs761397983 Chr.7: 86839017-86839017 on Build GRCh38
rs750409205 Chr.7: 86839119-86839119 on Build GRCh38
rs772305009 Chr.7: 86838786-86838786 on Build GRCh38
rs921824097 Chr.7: 86838759-86838759 on Build GRCh38
rs911991699 Chr.7: 86838775-86838775 on Build GRCh38
rs140980216 Chr.7: 86839200-86839200 on Build GRCh38
rs374144916 Chr.7: 86839160-86839160 on Build GRCh38
rs776921442 Chr.7: 86838810-86838810 on Build GRCh38
rs952942353 Chr.7: 86838781-86838791 on Build GRCh38
rs267601605 Chr.7: 86839156-86839156 on Build GRCh38
rs936238554 Chr.7: 86838832-86838832 on Build GRCh38
rs200724318 Chr.7: 86838940-86838940 on Build GRCh38
rs768237070 Chr.7: 86839151-86839151 on Build GRCh38
rs762291770 Chr.7: 86838908-86838908 on Build GRCh38
rs974856347 Chr.7: 86839155-86839155 on Build GRCh38
rs760146774 Chr.7: 86838847-86838847 on Build GRCh38
rs760777918 Chr.7: 86838805-86838805 on Build GRCh38
rs891435292 Chr.7: 86838907-86838907 on Build GRCh38
rs778683864 Chr.7: 86839111-86839111 on Build GRCh38
rs774081479 Chr.7: 86838812-86838812 on Build GRCh38
rs1037891916 Chr.7: 86838803-86838803 on Build GRCh38
rs771655599 Chr.7: 86838818-86838818 on Build GRCh38
rs763382792 Chr.7: 86839043-86839043 on Build GRCh38
rs112722806 Chr.7: 86838987-86838987 on Build GRCh38
rs566982395 Chr.7: 86839015-86839015 on Build GRCh38
rs766672277 Chr.7: 86838937-86838937 on Build GRCh38
rs146588543 Chr.7: 86839016-86839016 on Build GRCh38
rs367599440 Chr.7: 86838828-86838828 on Build GRCh38
rs112997659 Chr.7: 86839187-86839187 on Build GRCh38
rs141295273 Chr.7: 86839061-86839061 on Build GRCh38
rs199787664 Chr.7: 86839182-86839182 on Build GRCh38
rs964809835 Chr.7: 86839152-86839152 on Build GRCh38
rs1025523332 Chr.7: 86839053-86839053 on Build GRCh38
rs752635710 Chr.7: 86838893-86838893 on Build GRCh38

View Full Product Details