Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 132126797-132127306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLXNA4
Entrez Gene ID
91584
Gene Name
plexin A4
Gene Aliases
FAYV2820, PLEXA4, PLXNA4A, PLXNA4B, PRO34003
UniGene
Hs.511454
Gene Chromosome Location
Chr.7: 132123332-132648688 on Build GRCh38
Transcripts
NM_020911.1 XM_005250686.4 XM_006716171.3 XM_017012779.1

SNP Detail

SNP ID Chromosome Location
rs138225241 Chr.7: 132127023-132127023 on Build GRCh38
rs980434370 Chr.7: 132126862-132126862 on Build GRCh38
rs184304249 Chr.7: 132126854-132126854 on Build GRCh38
rs947317500 Chr.7: 132127185-132127185 on Build GRCh38
rs1037916634 Chr.7: 132127120-132127120 on Build GRCh38
rs903008747 Chr.7: 132126986-132126986 on Build GRCh38
rs906110119 Chr.7: 132127122-132127122 on Build GRCh38
rs569051317 Chr.7: 132126913-132126913 on Build GRCh38
rs144060966 Chr.7: 132127261-132127261 on Build GRCh38
rs1054174690 Chr.7: 132127161-132127161 on Build GRCh38
rs933402131 Chr.7: 132127050-132127050 on Build GRCh38
rs979345428 Chr.7: 132126964-132126964 on Build GRCh38
rs888837810 Chr.7: 132127234-132127235 on Build GRCh38
rs11343554 Chr.7: 132127017-132127017 on Build GRCh38
rs768059353 Chr.7: 132127160-132127160 on Build GRCh38
rs770295881 Chr.7: 132127108-132127109 on Build GRCh38
rs766122943 Chr.7: 132126896-132126896 on Build GRCh38
rs1020154874 Chr.7: 132126983-132126983 on Build GRCh38
rs113091346 Chr.7: 132126873-132126873 on Build GRCh38
rs945223477 Chr.7: 132126932-132126932 on Build GRCh38
rs923308265 Chr.7: 132127041-132127041 on Build GRCh38
rs3734998 Chr.7: 132127028-132127028 on Build GRCh38
rs562880713 Chr.7: 132127013-132127013 on Build GRCh38
rs776433990 Chr.7: 132126951-132126951 on Build GRCh38
rs1002575718 Chr.7: 132126930-132126931 on Build GRCh38
rs1021138075 Chr.7: 132127199-132127199 on Build GRCh38
rs964591078 Chr.7: 132127234-132127234 on Build GRCh38
rs1016940008 Chr.7: 132126836-132126836 on Build GRCh38
rs997304167 Chr.7: 132127005-132127005 on Build GRCh38
rs905231947 Chr.7: 132126909-132126910 on Build GRCh38
rs73155241 Chr.7: 132127143-132127143 on Build GRCh38
rs925212277 Chr.7: 132126996-132126996 on Build GRCh38
rs184609411 Chr.7: 132127229-132127229 on Build GRCh38
rs35542849 Chr.7: 132127109-132127109 on Build GRCh38
rs117988713 Chr.7: 132127172-132127172 on Build GRCh38
rs543767933 Chr.7: 132126993-132126994 on Build GRCh38
rs754655183 Chr.7: 132127279-132127279 on Build GRCh38
rs926666527 Chr.7: 132127154-132127154 on Build GRCh38
rs559917011 Chr.7: 132127049-132127049 on Build GRCh38
rs150390064 Chr.7: 132127000-132127000 on Build GRCh38
rs893017441 Chr.7: 132126971-132126971 on Build GRCh38
rs189880714 Chr.7: 132127057-132127057 on Build GRCh38
rs1055725331 Chr.7: 132126944-132126944 on Build GRCh38
rs191622723 Chr.7: 132127093-132127093 on Build GRCh38
rs886580003 Chr.7: 132127099-132127099 on Build GRCh38
rs960434025 Chr.7: 132126893-132126893 on Build GRCh38
rs536655579 Chr.7: 132127100-132127100 on Build GRCh38
rs3734997 Chr.7: 132127227-132127227 on Build GRCh38
rs1016535403 Chr.7: 132127030-132127030 on Build GRCh38
rs751702902 Chr.7: 132127134-132127134 on Build GRCh38
rs1016371761 Chr.7: 132127266-132127266 on Build GRCh38
rs1011434990 Chr.7: 132126972-132126972 on Build GRCh38
rs903018519 Chr.7: 132127215-132127215 on Build GRCh38
rs34491432 Chr.7: 132127180-132127181 on Build GRCh38
rs553066403 Chr.7: 132127168-132127168 on Build GRCh38
rs557419702 Chr.7: 132127019-132127019 on Build GRCh38
rs1036464434 Chr.7: 132126828-132126828 on Build GRCh38
rs949501182 Chr.7: 132126890-132126890 on Build GRCh38
rs913719101 Chr.7: 132126909-132126909 on Build GRCh38
rs780296068 Chr.7: 132127207-132127207 on Build GRCh38
rs188924584 Chr.7: 132127282-132127282 on Build GRCh38
rs548754821 Chr.7: 132127070-132127070 on Build GRCh38

View Full Product Details