Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 135363762-135364272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT4
Entrez Gene ID
4850
Gene Name
CCR4-NOT transcription complex subunit 4
Gene Aliases
CLONE243, NOT4, NOT4H
UniGene
Hs.490224
Gene Chromosome Location
Chr.7: 135361795-135510127 on Build GRCh38
Transcripts
NM_001190847.1 NM_001190849.1 NM_001190850.1 NM_013316.3 XM_017012235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4673208 c.1748C>T p.A583V Chr.7: 135363946-135363946 on Build GRCh38
1622367 c.1663G>C p.E555Q Chr.7: 135364031-135364031 on Build GRCh38
1596755 c.1730C>A p.S577Y Chr.7: 135363964-135363964 on Build GRCh38
5930267 c.1739C>T p.P580L Chr.7: 135363955-135363955 on Build GRCh38
3633640 c.1738C>A p.P580T Chr.7: 135363956-135363956 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780568120 Chr.7: 135364099-135364099 on Build GRCh38
rs3812265 Chr.7: 135364052-135364052 on Build GRCh38
rs750895743 Chr.7: 135364103-135364103 on Build GRCh38
rs948307294 Chr.7: 135364139-135364139 on Build GRCh38
rs746893577 Chr.7: 135363928-135363928 on Build GRCh38
rs781105581 Chr.7: 135363889-135363889 on Build GRCh38
rs780181408 Chr.7: 135363908-135363908 on Build GRCh38
rs919500954 Chr.7: 135364034-135364034 on Build GRCh38
rs766510654 Chr.7: 135364231-135364231 on Build GRCh38
rs1055148978 Chr.7: 135364155-135364155 on Build GRCh38
rs966338808 Chr.7: 135363921-135363921 on Build GRCh38
rs1046573128 Chr.7: 135364152-135364152 on Build GRCh38
rs776248981 Chr.7: 135363819-135363819 on Build GRCh38
rs749528170 Chr.7: 135363909-135363909 on Build GRCh38
rs28452134 Chr.7: 135364098-135364098 on Build GRCh38
rs751022384 Chr.7: 135363958-135363960 on Build GRCh38
rs759089041 Chr.7: 135363827-135363827 on Build GRCh38
rs1052967812 Chr.7: 135364222-135364222 on Build GRCh38
rs764521860 Chr.7: 135364027-135364027 on Build GRCh38
rs79043969 Chr.7: 135363926-135363926 on Build GRCh38
rs370767888 Chr.7: 135364020-135364020 on Build GRCh38
rs747415149 Chr.7: 135364083-135364084 on Build GRCh38
rs28371048 Chr.7: 135364104-135364104 on Build GRCh38
rs77245177 Chr.7: 135364036-135364036 on Build GRCh38
rs1011358721 Chr.7: 135364248-135364248 on Build GRCh38
rs764609745 Chr.7: 135363864-135363864 on Build GRCh38
rs938062935 Chr.7: 135364150-135364150 on Build GRCh38
rs1055433352 Chr.7: 135364118-135364118 on Build GRCh38
rs756214971 Chr.7: 135363906-135363906 on Build GRCh38
rs775266666 Chr.7: 135363995-135363995 on Build GRCh38
rs999671137 Chr.7: 135364217-135364217 on Build GRCh38
rs773490102 Chr.7: 135364076-135364076 on Build GRCh38
rs768025295 Chr.7: 135364099-135364099 on Build GRCh38
rs745907506 Chr.7: 135363891-135363891 on Build GRCh38
rs757581004 Chr.7: 135364059-135364059 on Build GRCh38
rs770951118 Chr.7: 135363929-135363929 on Build GRCh38
rs1032804283 Chr.7: 135363790-135363790 on Build GRCh38
rs756127138 Chr.7: 135364108-135364108 on Build GRCh38
rs957310071 Chr.7: 135363821-135363821 on Build GRCh38
rs752004848 Chr.7: 135363879-135363879 on Build GRCh38
rs764845164 Chr.7: 135363978-135363978 on Build GRCh38
rs771158003 Chr.7: 135364109-135364109 on Build GRCh38
rs759047664 Chr.7: 135363967-135363967 on Build GRCh38
rs774529944 Chr.7: 135363927-135363927 on Build GRCh38
rs906697193 Chr.7: 135364163-135364163 on Build GRCh38
rs377392481 Chr.7: 135363915-135363915 on Build GRCh38
rs972235097 Chr.7: 135363989-135363989 on Build GRCh38
rs769092265 Chr.7: 135363828-135363828 on Build GRCh38
rs373363789 Chr.7: 135363881-135363881 on Build GRCh38
rs767602229 Chr.7: 135364065-135364065 on Build GRCh38
rs545287338 Chr.7: 135364179-135364179 on Build GRCh38
rs756736493 Chr.7: 135364098-135364099 on Build GRCh38
rs758771595 Chr.7: 135363837-135363837 on Build GRCh38
rs548071392 Chr.7: 135363947-135363947 on Build GRCh38
rs765806107 Chr.7: 135363816-135363816 on Build GRCh38
rs11976681 Chr.7: 135363971-135363971 on Build GRCh38
rs916865957 Chr.7: 135364128-135364128 on Build GRCh38
rs771843733 Chr.7: 135363919-135363919 on Build GRCh38

View Full Product Details