Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 138706351-138706847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0A4
Entrez Gene ID
50617
Gene Name
ATPase H+ transporting V0 subunit a4
Gene Aliases
A4, ATP6N1B, ATP6N2, RDRTA2, RTA1C, RTADR, STV1, VPH1, VPP2
UniGene
Hs.98967
Gene Chromosome Location
Chr.7: 138706294-138799839 on Build GRCh38
Transcripts
NM_020632.2 NM_130840.2 NM_130841.2 XM_005250393.1 XM_005250394.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6608057 c.2451C>A p.F817L Chr.7: 138706696-138706696 on Build GRCh38
342564 c.2479C>T p.P827S Chr.7: 138706668-138706668 on Build GRCh38
7079407 c.2458G>C p.G820R Chr.7: 138706689-138706689 on Build GRCh38
1086026 c.2472G>T p.K824N Chr.7: 138706675-138706675 on Build GRCh38
126562 c.2515G>T p.E839* Chr.7: 138706632-138706632 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752038920 Chr.7: 138706662-138706662 on Build GRCh38
rs768262906 Chr.7: 138706450-138706450 on Build GRCh38
rs1037724632 Chr.7: 138706482-138706482 on Build GRCh38
rs200496062 Chr.7: 138706757-138706757 on Build GRCh38
rs922397757 Chr.7: 138706728-138706728 on Build GRCh38
rs1031637872 Chr.7: 138706580-138706580 on Build GRCh38
rs374305455 Chr.7: 138706632-138706632 on Build GRCh38
rs78419825 Chr.7: 138706551-138706551 on Build GRCh38
rs190782161 Chr.7: 138706679-138706679 on Build GRCh38
rs267606671 Chr.7: 138706689-138706689 on Build GRCh38
rs563491903 Chr.7: 138706451-138706451 on Build GRCh38
rs890197772 Chr.7: 138706487-138706487 on Build GRCh38
rs572401906 Chr.7: 138706776-138706776 on Build GRCh38
rs934266733 Chr.7: 138706696-138706696 on Build GRCh38
rs745521611 Chr.7: 138706742-138706742 on Build GRCh38
rs773756702 Chr.7: 138706652-138706652 on Build GRCh38
rs73166921 Chr.7: 138706809-138706809 on Build GRCh38
rs1000481176 Chr.7: 138706514-138706514 on Build GRCh38
rs1038720440 Chr.7: 138706774-138706774 on Build GRCh38
rs765535735 Chr.7: 138706622-138706622 on Build GRCh38
rs775854630 Chr.7: 138706623-138706623 on Build GRCh38
rs556303754 Chr.7: 138706720-138706720 on Build GRCh38
rs948074385 Chr.7: 138706777-138706777 on Build GRCh38
rs541097291 Chr.7: 138706807-138706807 on Build GRCh38
rs758110248 Chr.7: 138706690-138706690 on Build GRCh38
rs770369261 Chr.7: 138706602-138706602 on Build GRCh38
rs184174235 Chr.7: 138706382-138706382 on Build GRCh38
rs933853259 Chr.7: 138706422-138706422 on Build GRCh38
rs368964932 Chr.7: 138706633-138706633 on Build GRCh38
rs1003748725 Chr.7: 138706570-138706570 on Build GRCh38
rs550036710 Chr.7: 138706405-138706405 on Build GRCh38
rs558827285 Chr.7: 138706768-138706768 on Build GRCh38
rs375615719 Chr.7: 138706708-138706708 on Build GRCh38
rs376502687 Chr.7: 138706772-138706772 on Build GRCh38
rs768870304 Chr.7: 138706764-138706764 on Build GRCh38
rs201317315 Chr.7: 138706737-138706737 on Build GRCh38
rs55832008 Chr.7: 138706462-138706462 on Build GRCh38
rs1047422453 Chr.7: 138706444-138706444 on Build GRCh38
rs770534833 Chr.7: 138706603-138706603 on Build GRCh38
rs75920897 Chr.7: 138706513-138706513 on Build GRCh38
rs1007355074 Chr.7: 138706490-138706490 on Build GRCh38
rs574753785 Chr.7: 138706825-138706825 on Build GRCh38
rs756268451 Chr.7: 138706683-138706683 on Build GRCh38
rs772009991 Chr.7: 138706577-138706577 on Build GRCh38
rs200276098 Chr.7: 138706627-138706627 on Build GRCh38
rs371818187 Chr.7: 138706726-138706726 on Build GRCh38
rs767396201 Chr.7: 138706812-138706812 on Build GRCh38
rs746571093 Chr.7: 138706600-138706600 on Build GRCh38
rs34783145 Chr.7: 138706771-138706772 on Build GRCh38
rs141194465 Chr.7: 138706676-138706676 on Build GRCh38
rs763400668 Chr.7: 138706631-138706631 on Build GRCh38
rs770624630 Chr.7: 138706736-138706736 on Build GRCh38
rs772623818 Chr.7: 138706591-138706591 on Build GRCh38
rs970115299 Chr.7: 138706640-138706640 on Build GRCh38
rs1004081484 Chr.7: 138706813-138706813 on Build GRCh38
rs201819879 Chr.7: 138706687-138706687 on Build GRCh38
rs753356162 Chr.7: 138706633-138706633 on Build GRCh38
rs763437707 Chr.7: 138706799-138706799 on Build GRCh38
rs8846 Chr.7: 138706385-138706385 on Build GRCh38
rs759250618 Chr.7: 138706620-138706620 on Build GRCh38
rs897446123 Chr.7: 138706414-138706414 on Build GRCh38
rs1037296429 Chr.7: 138706386-138706386 on Build GRCh38
rs375266833 Chr.7: 138706744-138706744 on Build GRCh38
rs776494245 Chr.7: 138706610-138706610 on Build GRCh38
rs377442551 Chr.7: 138706637-138706637 on Build GRCh38
rs1017440578 Chr.7: 138706505-138706505 on Build GRCh38
rs6956646 Chr.7: 138706666-138706666 on Build GRCh38

View Full Product Details