Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 138837241-138837740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1549
Entrez Gene ID
57670
Gene Name
KIAA1549
Gene Aliases
-
UniGene
Hs.605380
Gene Chromosome Location
Chr.7: 138831377-138981626 on Build GRCh38
Transcripts
NM_001164665.1 NM_020910.2 XM_011516442.2

SNP Detail

SNP ID Chromosome Location
rs1004198398 Chr.7: 138837307-138837307 on Build GRCh38
rs571709653 Chr.7: 138837655-138837655 on Build GRCh38
rs1020134870 Chr.7: 138837326-138837326 on Build GRCh38
rs555635640 Chr.7: 138837293-138837293 on Build GRCh38
rs751457734 Chr.7: 138837608-138837608 on Build GRCh38
rs550236632 Chr.7: 138837597-138837597 on Build GRCh38
rs752404662 Chr.7: 138837518-138837518 on Build GRCh38
rs1021639395 Chr.7: 138837707-138837707 on Build GRCh38
rs112392421 Chr.7: 138837633-138837633 on Build GRCh38
rs958188062 Chr.7: 138837413-138837413 on Build GRCh38
rs987462289 Chr.7: 138837535-138837535 on Build GRCh38
rs981342833 Chr.7: 138837681-138837681 on Build GRCh38
rs575429012 Chr.7: 138837298-138837298 on Build GRCh38
rs931136497 Chr.7: 138837522-138837522 on Build GRCh38
rs188187990 Chr.7: 138837630-138837630 on Build GRCh38
rs1052091323 Chr.7: 138837488-138837488 on Build GRCh38
rs1028453870 Chr.7: 138837382-138837382 on Build GRCh38
rs781458715 Chr.7: 138837698-138837698 on Build GRCh38
rs948209640 Chr.7: 138837639-138837639 on Build GRCh38
rs939568706 Chr.7: 138837710-138837710 on Build GRCh38
rs560409834 Chr.7: 138837525-138837525 on Build GRCh38
rs977088400 Chr.7: 138837479-138837479 on Build GRCh38
rs920751624 Chr.7: 138837415-138837416 on Build GRCh38
rs533182210 Chr.7: 138837486-138837486 on Build GRCh38
rs544339921 Chr.7: 138837460-138837460 on Build GRCh38
rs562716065 Chr.7: 138837607-138837607 on Build GRCh38
rs750847755 Chr.7: 138837504-138837504 on Build GRCh38
rs564225582 Chr.7: 138837474-138837474 on Build GRCh38
rs948319895 Chr.7: 138837426-138837426 on Build GRCh38
rs965462928 Chr.7: 138837335-138837335 on Build GRCh38
rs538248733 Chr.7: 138837561-138837561 on Build GRCh38
rs541295067 Chr.7: 138837590-138837590 on Build GRCh38
rs975584836 Chr.7: 138837379-138837379 on Build GRCh38
rs372041177 Chr.7: 138837483-138837483 on Build GRCh38
rs962121055 Chr.7: 138837555-138837555 on Build GRCh38
rs1052122039 Chr.7: 138837716-138837716 on Build GRCh38
rs540233340 Chr.7: 138837520-138837520 on Build GRCh38
rs928384563 Chr.7: 138837514-138837514 on Build GRCh38
rs914139897 Chr.7: 138837430-138837430 on Build GRCh38
rs193045454 Chr.7: 138837271-138837271 on Build GRCh38
rs894348093 Chr.7: 138837521-138837521 on Build GRCh38
rs958217488 Chr.7: 138837649-138837649 on Build GRCh38
rs928233069 Chr.7: 138837693-138837693 on Build GRCh38
rs139410722 Chr.7: 138837626-138837626 on Build GRCh38
rs1038301293 Chr.7: 138837385-138837388 on Build GRCh38
rs989763005 Chr.7: 138837420-138837420 on Build GRCh38
rs1022566929 Chr.7: 138837515-138837515 on Build GRCh38
rs185990437 Chr.7: 138837542-138837542 on Build GRCh38
rs892247886 Chr.7: 138837507-138837507 on Build GRCh38
rs906198606 Chr.7: 138837461-138837461 on Build GRCh38
rs765583971 Chr.7: 138837308-138837308 on Build GRCh38
rs184876140 Chr.7: 138837527-138837527 on Build GRCh38
rs868495209 Chr.7: 138837603-138837603 on Build GRCh38
rs191551821 Chr.7: 138837673-138837673 on Build GRCh38
rs1026852261 Chr.7: 138837272-138837272 on Build GRCh38

View Full Product Details