Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 141660260-141660763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1147
Entrez Gene ID
57189
Gene Name
KIAA1147
Gene Aliases
LCHN, PRO2561
UniGene
Hs.521240
Gene Chromosome Location
Chr.7: 141652381-141702188 on Build GRCh38
Transcripts
NM_001080392.1

SNP Detail

SNP ID Chromosome Location
rs755455621 Chr.7: 141660317-141660317 on Build GRCh38
rs752094448 Chr.7: 141660308-141660308 on Build GRCh38
rs968686520 Chr.7: 141660660-141660660 on Build GRCh38
rs978356507 Chr.7: 141660448-141660448 on Build GRCh38
rs968570233 Chr.7: 141660334-141660334 on Build GRCh38
rs1017516953 Chr.7: 141660641-141660641 on Build GRCh38
rs993664467 Chr.7: 141660625-141660625 on Build GRCh38
rs948157222 Chr.7: 141660618-141660618 on Build GRCh38
rs996278633 Chr.7: 141660411-141660411 on Build GRCh38
rs184806296 Chr.7: 141660630-141660630 on Build GRCh38
rs1044053071 Chr.7: 141660503-141660503 on Build GRCh38
rs934503383 Chr.7: 141660382-141660382 on Build GRCh38
rs964354511 Chr.7: 141660320-141660320 on Build GRCh38
rs916635830 Chr.7: 141660584-141660584 on Build GRCh38
rs968728315 Chr.7: 141660427-141660427 on Build GRCh38
rs924477303 Chr.7: 141660359-141660359 on Build GRCh38
rs565531204 Chr.7: 141660561-141660561 on Build GRCh38
rs774587861 Chr.7: 141660484-141660485 on Build GRCh38
rs948385772 Chr.7: 141660423-141660423 on Build GRCh38
rs898019272 Chr.7: 141660599-141660599 on Build GRCh38
rs776472711 Chr.7: 141660623-141660625 on Build GRCh38
rs143876077 Chr.7: 141660634-141660634 on Build GRCh38
rs993015866 Chr.7: 141660397-141660397 on Build GRCh38
rs916986337 Chr.7: 141660406-141660406 on Build GRCh38
rs886273737 Chr.7: 141660712-141660712 on Build GRCh38
rs189025728 Chr.7: 141660710-141660710 on Build GRCh38
rs752939305 Chr.7: 141660544-141660544 on Build GRCh38
rs764258410 Chr.7: 141660443-141660443 on Build GRCh38
rs138201419 Chr.7: 141660500-141660500 on Build GRCh38
rs181422859 Chr.7: 141660731-141660731 on Build GRCh38
rs528713287 Chr.7: 141660565-141660565 on Build GRCh38
rs1007521341 Chr.7: 141660636-141660636 on Build GRCh38
rs112134657 Chr.7: 141660724-141660724 on Build GRCh38
rs555014372 Chr.7: 141660726-141660726 on Build GRCh38
rs905447577 Chr.7: 141660528-141660528 on Build GRCh38
rs919921660 Chr.7: 141660631-141660631 on Build GRCh38
rs979118925 Chr.7: 141660692-141660692 on Build GRCh38
rs761151538 Chr.7: 141660476-141660477 on Build GRCh38
rs767992229 Chr.7: 141660346-141660346 on Build GRCh38
rs1021958458 Chr.7: 141660414-141660414 on Build GRCh38
rs1017485902 Chr.7: 141660309-141660309 on Build GRCh38

View Full Product Details