Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41655077-41655581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANK1
Entrez Gene ID
286
Gene Name
ankyrin 1
Gene Aliases
ANK, SPH1, SPH2
UniGene
Hs.654438
Gene Chromosome Location
Chr.8: 41653225-41896762 on Build GRCh38
Transcripts
NM_000037.3 NM_001142445.1 NM_001142446.1 NM_020475.2 NM_020476.2 NM_020477.2 NM_020478.4 NM_020480.4 XM_005273476.4 XM_011544490.2 XM_011544491.2 XM_011544494.2 XM_011544495.2 XM_011544496.2 XM_011544500.2 XM_011544501.2 XM_011544502.2 XM_011544503.2 XM_011544504.2 XM_011544505.2 XM_017013319.1 XM_017013320.1 XM_017013321.1 XM_017013322.1 XM_017013323.1 XM_017013324.1 XM_017013325.1 XM_017013326.1 XM_017013327.1 XM_017013328.1 XM_017013329.1

SNP Detail

SNP ID Chromosome Location
rs1052876715 Chr.8: 41655139-41655139 on Build GRCh38
rs547095997 Chr.8: 41655125-41655125 on Build GRCh38
rs1027183274 Chr.8: 41655496-41655496 on Build GRCh38
rs925874304 Chr.8: 41655423-41655423 on Build GRCh38
rs866619855 Chr.8: 41655371-41655371 on Build GRCh38
rs950377874 Chr.8: 41655185-41655185 on Build GRCh38
rs1015585619 Chr.8: 41655292-41655292 on Build GRCh38
rs5891167 Chr.8: 41655306-41655307 on Build GRCh38
rs1007422175 Chr.8: 41655530-41655530 on Build GRCh38
rs72638944 Chr.8: 41655552-41655552 on Build GRCh38
rs796345487 Chr.8: 41655130-41655133 on Build GRCh38
rs970184906 Chr.8: 41655498-41655498 on Build GRCh38
rs560680999 Chr.8: 41655167-41655167 on Build GRCh38
rs572272946 Chr.8: 41655219-41655219 on Build GRCh38
rs530516884 Chr.8: 41655520-41655520 on Build GRCh38
rs868688551 Chr.8: 41655461-41655461 on Build GRCh38
rs886062936 Chr.8: 41655139-41655139 on Build GRCh38
rs762228467 Chr.8: 41655136-41655137 on Build GRCh38
rs1042911297 Chr.8: 41655211-41655211 on Build GRCh38
rs913914193 Chr.8: 41655305-41655305 on Build GRCh38
rs775470921 Chr.8: 41655295-41655295 on Build GRCh38
rs988918946 Chr.8: 41655556-41655556 on Build GRCh38
rs570356781 Chr.8: 41655457-41655457 on Build GRCh38
rs552774855 Chr.8: 41655474-41655474 on Build GRCh38
rs566279026 Chr.8: 41655501-41655501 on Build GRCh38
rs962882956 Chr.8: 41655251-41655251 on Build GRCh38
rs3138830 Chr.8: 41655131-41655132 on Build GRCh38
rs147352953 Chr.8: 41655247-41655247 on Build GRCh38
rs947131412 Chr.8: 41655201-41655201 on Build GRCh38
rs771334078 Chr.8: 41655547-41655547 on Build GRCh38
rs910131455 Chr.8: 41655238-41655238 on Build GRCh38
rs1026412850 Chr.8: 41655172-41655172 on Build GRCh38
rs550204540 Chr.8: 41655395-41655395 on Build GRCh38
rs763206680 Chr.8: 41655138-41655139 on Build GRCh38
rs997243730 Chr.8: 41655156-41655156 on Build GRCh38
rs61311542 Chr.8: 41655548-41655548 on Build GRCh38
rs755916036 Chr.8: 41655305-41655305 on Build GRCh38
rs5891166 Chr.8: 41655153-41655154 on Build GRCh38
rs539354854 Chr.8: 41655460-41655460 on Build GRCh38
rs33981001 Chr.8: 41655130-41655131 on Build GRCh38
rs201295961 Chr.8: 41655320-41655320 on Build GRCh38
rs939453142 Chr.8: 41655456-41655456 on Build GRCh38
rs1007392735 Chr.8: 41655123-41655123 on Build GRCh38
rs112209933 Chr.8: 41655405-41655405 on Build GRCh38
rs980889226 Chr.8: 41655373-41655373 on Build GRCh38
rs1008609084 Chr.8: 41655287-41655287 on Build GRCh38
rs980368759 Chr.8: 41655522-41655522 on Build GRCh38
rs912940051 Chr.8: 41655165-41655165 on Build GRCh38
rs374082372 Chr.8: 41655408-41655408 on Build GRCh38
rs928842182 Chr.8: 41655524-41655524 on Build GRCh38
rs996080743 Chr.8: 41655239-41655239 on Build GRCh38
rs990360138 Chr.8: 41655492-41655493 on Build GRCh38
rs926771439 Chr.8: 41655400-41655400 on Build GRCh38
rs373938111 Chr.8: 41655129-41655130 on Build GRCh38
rs891128468 Chr.8: 41655253-41655253 on Build GRCh38
rs771828749 Chr.8: 41655232-41655232 on Build GRCh38
rs938324108 Chr.8: 41655133-41655133 on Build GRCh38
rs3063734 Chr.8: 41655152-41655153 on Build GRCh38
rs945394596 Chr.8: 41655475-41655475 on Build GRCh38
rs1042068456 Chr.8: 41655418-41655418 on Build GRCh38
rs59666244 Chr.8: 41655319-41655319 on Build GRCh38
rs540659486 Chr.8: 41655305-41655306 on Build GRCh38
rs144081612 Chr.8: 41655137-41655138 on Build GRCh38
rs764152737 Chr.8: 41655437-41655437 on Build GRCh38
rs527359681 Chr.8: 41655109-41655109 on Build GRCh38

View Full Product Details