Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 12990717-12991216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1456
Entrez Gene ID
57604
Gene Name
KIAA1456
Gene Aliases
C8orf79, TRM9L
UniGene
Hs.202521
Gene Chromosome Location
Chr.8: 12945674-13029777 on Build GRCh38
Transcripts
NM_020844.2 XM_005273584.4 XM_005273585.4 XM_005273586.4 XM_005273587.4 XM_005273588.4 XM_005273591.4 XM_011544596.2 XM_011544597.2 XM_011544598.2 XM_011544600.2 XM_011544601.2 XM_017013705.1 XM_017013706.1 XM_017013707.1 XM_017013708.1 XM_017013709.1 XM_017013710.1 XM_017013711.1 XM_017013712.1 XM_017013713.1 XM_017013714.1 XM_017013715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5368277 c.344G>A p.R115H Chr.8: 12990923-12990923 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs928962082 Chr.8: 12990803-12990803 on Build GRCh38
rs963123564 Chr.8: 12990943-12990943 on Build GRCh38
rs990244255 Chr.8: 12990845-12990845 on Build GRCh38
rs564359919 Chr.8: 12991123-12991123 on Build GRCh38
rs533230135 Chr.8: 12991173-12991173 on Build GRCh38
rs775606876 Chr.8: 12990745-12990745 on Build GRCh38
rs917171720 Chr.8: 12990807-12990807 on Build GRCh38
rs750809767 Chr.8: 12990999-12990999 on Build GRCh38
rs910358970 Chr.8: 12991039-12991039 on Build GRCh38
rs948678824 Chr.8: 12990990-12990990 on Build GRCh38
rs898633702 Chr.8: 12991074-12991074 on Build GRCh38
rs933066758 Chr.8: 12991029-12991029 on Build GRCh38
rs950338049 Chr.8: 12990957-12990957 on Build GRCh38
rs959232082 Chr.8: 12990961-12990961 on Build GRCh38
rs764782376 Chr.8: 12990839-12990839 on Build GRCh38
rs1025721399 Chr.8: 12990929-12990929 on Build GRCh38
rs1041452748 Chr.8: 12990875-12990875 on Build GRCh38
rs181398769 Chr.8: 12991104-12991104 on Build GRCh38
rs574805157 Chr.8: 12990885-12990885 on Build GRCh38
rs925750495 Chr.8: 12991001-12991001 on Build GRCh38
rs891901277 Chr.8: 12991027-12991027 on Build GRCh38
rs760038737 Chr.8: 12990806-12990806 on Build GRCh38
rs1007331877 Chr.8: 12990765-12990765 on Build GRCh38
rs569294225 Chr.8: 12990790-12990790 on Build GRCh38
rs992729147 Chr.8: 12990805-12990805 on Build GRCh38
rs1002933589 Chr.8: 12991060-12991060 on Build GRCh38
rs933990024 Chr.8: 12991044-12991044 on Build GRCh38
rs910382496 Chr.8: 12991177-12991177 on Build GRCh38
rs890038086 Chr.8: 12991055-12991055 on Build GRCh38
rs762027140 Chr.8: 12990922-12990922 on Build GRCh38
rs867738745 Chr.8: 12990889-12990889 on Build GRCh38
rs566901745 Chr.8: 12990836-12990836 on Build GRCh38
rs1029892219 Chr.8: 12990899-12990899 on Build GRCh38
rs1016918362 Chr.8: 12990770-12990770 on Build GRCh38
rs375494660 Chr.8: 12990811-12990811 on Build GRCh38
rs566985271 Chr.8: 12991174-12991174 on Build GRCh38
rs889031876 Chr.8: 12990751-12990751 on Build GRCh38
rs866772870 Chr.8: 12990917-12990917 on Build GRCh38
rs569397642 Chr.8: 12991113-12991113 on Build GRCh38
rs780998954 Chr.8: 12990890-12990890 on Build GRCh38
rs553269192 Chr.8: 12990881-12990881 on Build GRCh38
rs74593148 Chr.8: 12991057-12991057 on Build GRCh38
rs529570244 Chr.8: 12991079-12991079 on Build GRCh38
rs966995088 Chr.8: 12990995-12990995 on Build GRCh38
rs1021919992 Chr.8: 12991083-12991083 on Build GRCh38
rs530519912 Chr.8: 12990874-12990874 on Build GRCh38
rs866452104 Chr.8: 12991186-12991186 on Build GRCh38
rs936126899 Chr.8: 12990837-12990837 on Build GRCh38
rs563725715 Chr.8: 12991047-12991047 on Build GRCh38
rs990388571 Chr.8: 12990963-12990963 on Build GRCh38
rs1034943568 Chr.8: 12991068-12991068 on Build GRCh38
rs994354638 Chr.8: 12991119-12991119 on Build GRCh38
rs1037544094 Chr.8: 12991045-12991045 on Build GRCh38
rs992926046 Chr.8: 12990969-12990969 on Build GRCh38
rs77476499 Chr.8: 12990910-12990910 on Build GRCh38
rs1026079778 Chr.8: 12990956-12990956 on Build GRCh38
rs776138334 Chr.8: 12990821-12990821 on Build GRCh38
rs139710712 Chr.8: 12991048-12991048 on Build GRCh38
rs935844537 Chr.8: 12991004-12991004 on Build GRCh38
rs374331246 Chr.8: 12990886-12990886 on Build GRCh38
rs778038980 Chr.8: 12990931-12990931 on Build GRCh38
rs1050705195 Chr.8: 12991031-12991031 on Build GRCh38
rs79103201 Chr.8: 12990788-12990788 on Build GRCh38
rs568432239 Chr.8: 12990779-12990779 on Build GRCh38
rs1047037070 Chr.8: 12991056-12991056 on Build GRCh38
rs1003479446 Chr.8: 12990942-12990942 on Build GRCh38
rs1050257638 Chr.8: 12990896-12990896 on Build GRCh38
rs973113592 Chr.8: 12990955-12990955 on Build GRCh38
rs772467127 Chr.8: 12990814-12990814 on Build GRCh38
rs962789556 Chr.8: 12990777-12990777 on Build GRCh38
rs796543868 Chr.8: 12990904-12990904 on Build GRCh38
rs533445639 Chr.8: 12991122-12991122 on Build GRCh38
rs372402513 Chr.8: 12990789-12990789 on Build GRCh38
rs948561555 Chr.8: 12990808-12990808 on Build GRCh38
rs1034503368 Chr.8: 12990950-12990950 on Build GRCh38
rs2460340 Chr.8: 12991182-12991182 on Build GRCh38
rs180951877 Chr.8: 12990782-12990782 on Build GRCh38
rs904608513 Chr.8: 12990835-12990835 on Build GRCh38
rs971445634 Chr.8: 12990939-12990939 on Build GRCh38
rs1053105113 Chr.8: 12991012-12991012 on Build GRCh38
rs891764393 Chr.8: 12990869-12990869 on Build GRCh38
rs575286897 Chr.8: 12990953-12990953 on Build GRCh38
rs562000483 Chr.8: 12991073-12991073 on Build GRCh38
rs1026048729 Chr.8: 12991133-12991133 on Build GRCh38
rs545624937 Chr.8: 12991040-12991040 on Build GRCh38
rs762638089 Chr.8: 12990859-12990859 on Build GRCh38
rs77826707 Chr.8: 12990850-12990850 on Build GRCh38
rs558180743 Chr.8: 12990812-12990812 on Build GRCh38
rs1006115204 Chr.8: 12990907-12990907 on Build GRCh38
rs17179534 Chr.8: 12990911-12990911 on Build GRCh38
rs766124192 Chr.8: 12990867-12990867 on Build GRCh38
rs781312981 Chr.8: 12990908-12990909 on Build GRCh38
rs958865969 Chr.8: 12991070-12991070 on Build GRCh38
rs767863741 Chr.8: 12990980-12990980 on Build GRCh38
rs190184736 Chr.8: 12990884-12990884 on Build GRCh38
rs185572716 Chr.8: 12990791-12990791 on Build GRCh38
rs761240955 Chr.8: 12991065-12991065 on Build GRCh38
rs977110125 Chr.8: 12991121-12991121 on Build GRCh38
rs771394052 Chr.8: 12990870-12990870 on Build GRCh38

View Full Product Details