Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 23007020-23007513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHOBTB2
Entrez Gene ID
23221
Gene Name
Rho related BTB domain containing 2
Gene Aliases
DBC2
UniGene
Hs.372688
Gene Chromosome Location
Chr.8: 22987406-23020197 on Build GRCh38
Transcripts
NM_001160036.1 NM_001160037.1 NM_015178.2 XM_017013251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3647323 c.1068C>T p.G356G Chr.8: 23007313-23007313 on Build GRCh38
6766238 c.938G>T p.G313V Chr.8: 23007183-23007183 on Build GRCh38
5791198 c.1072G>A p.G358S Chr.8: 23007317-23007317 on Build GRCh38
3647321 c.927G>A p.S309S Chr.8: 23007172-23007172 on Build GRCh38
6766236 c.1151G>A p.R384H Chr.8: 23007396-23007396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531552869 Chr.8: 23007473-23007473 on Build GRCh38
rs371701582 Chr.8: 23007074-23007074 on Build GRCh38
rs753270855 Chr.8: 23007051-23007051 on Build GRCh38
rs779630400 Chr.8: 23007221-23007223 on Build GRCh38
rs750082460 Chr.8: 23007066-23007066 on Build GRCh38
rs559073741 Chr.8: 23007223-23007223 on Build GRCh38
rs777287975 Chr.8: 23007283-23007283 on Build GRCh38
rs757015873 Chr.8: 23007332-23007332 on Build GRCh38
rs760140573 Chr.8: 23007374-23007374 on Build GRCh38
rs754709051 Chr.8: 23007296-23007296 on Build GRCh38
rs751243247 Chr.8: 23007164-23007164 on Build GRCh38
rs373034424 Chr.8: 23007444-23007444 on Build GRCh38
rs140190863 Chr.8: 23007047-23007047 on Build GRCh38
rs746142449 Chr.8: 23007246-23007246 on Build GRCh38
rs547602967 Chr.8: 23007046-23007046 on Build GRCh38
rs778573219 Chr.8: 23007384-23007384 on Build GRCh38
rs61732771 Chr.8: 23007375-23007375 on Build GRCh38
rs923691001 Chr.8: 23007460-23007460 on Build GRCh38
rs762216794 Chr.8: 23007284-23007284 on Build GRCh38
rs747030867 Chr.8: 23007349-23007349 on Build GRCh38
rs773039376 Chr.8: 23007100-23007100 on Build GRCh38
rs748651823 Chr.8: 23007408-23007408 on Build GRCh38
rs146017675 Chr.8: 23007425-23007425 on Build GRCh38
rs974102807 Chr.8: 23007169-23007169 on Build GRCh38
rs939187701 Chr.8: 23007183-23007183 on Build GRCh38
rs551660992 Chr.8: 23007071-23007071 on Build GRCh38
rs991902634 Chr.8: 23007471-23007471 on Build GRCh38
rs879846552 Chr.8: 23007043-23007043 on Build GRCh38
rs749014521 Chr.8: 23007237-23007237 on Build GRCh38
rs757703143 Chr.8: 23007157-23007157 on Build GRCh38
rs144883643 Chr.8: 23007075-23007075 on Build GRCh38
rs146706431 Chr.8: 23007040-23007040 on Build GRCh38
rs753416885 Chr.8: 23007458-23007458 on Build GRCh38
rs772023790 Chr.8: 23007251-23007251 on Build GRCh38
rs557999062 Chr.8: 23007316-23007316 on Build GRCh38
rs150618811 Chr.8: 23007347-23007347 on Build GRCh38
rs768821582 Chr.8: 23007398-23007398 on Build GRCh38
rs765492852 Chr.8: 23007365-23007365 on Build GRCh38
rs766069178 Chr.8: 23007158-23007158 on Build GRCh38
rs553849595 Chr.8: 23007224-23007224 on Build GRCh38
rs141884602 Chr.8: 23007333-23007333 on Build GRCh38
rs759486308 Chr.8: 23007115-23007115 on Build GRCh38
rs777639993 Chr.8: 23007328-23007328 on Build GRCh38
rs148121402 Chr.8: 23007289-23007289 on Build GRCh38
rs947987293 Chr.8: 23007220-23007220 on Build GRCh38
rs201146480 Chr.8: 23007219-23007219 on Build GRCh38
rs761262371 Chr.8: 23007262-23007262 on Build GRCh38
rs775701039 Chr.8: 23007252-23007252 on Build GRCh38
rs2466178 Chr.8: 23007109-23007109 on Build GRCh38
rs146884841 Chr.8: 23007142-23007142 on Build GRCh38
rs766921853 Chr.8: 23007312-23007312 on Build GRCh38
rs139255771 Chr.8: 23007159-23007159 on Build GRCh38
rs754324313 Chr.8: 23007154-23007154 on Build GRCh38
rs1005521110 Chr.8: 23007267-23007267 on Build GRCh38
rs200206570 Chr.8: 23007149-23007149 on Build GRCh38
rs751694780 Chr.8: 23007042-23007042 on Build GRCh38
rs112771785 Chr.8: 23007171-23007171 on Build GRCh38
rs377368156 Chr.8: 23007474-23007474 on Build GRCh38
rs772408810 Chr.8: 23007346-23007346 on Build GRCh38
rs369567249 Chr.8: 23007385-23007385 on Build GRCh38
rs748013447 Chr.8: 23007199-23007199 on Build GRCh38
rs778327025 Chr.8: 23007063-23007063 on Build GRCh38
rs763582578 Chr.8: 23007455-23007455 on Build GRCh38
rs750353391 Chr.8: 23007486-23007486 on Build GRCh38
rs778224382 Chr.8: 23007233-23007235 on Build GRCh38
rs767667340 Chr.8: 23007134-23007134 on Build GRCh38
rs912492615 Chr.8: 23007393-23007393 on Build GRCh38
rs201349334 Chr.8: 23007430-23007430 on Build GRCh38
rs200943704 Chr.8: 23007259-23007259 on Build GRCh38
rs943893227 Chr.8: 23007232-23007232 on Build GRCh38
rs755623851 Chr.8: 23007325-23007325 on Build GRCh38
rs142645813 Chr.8: 23007360-23007360 on Build GRCh38
rs150304103 Chr.8: 23007069-23007069 on Build GRCh38
rs767129651 Chr.8: 23007369-23007369 on Build GRCh38
rs747298406 Chr.8: 23007338-23007339 on Build GRCh38
rs753391613 Chr.8: 23007160-23007160 on Build GRCh38
rs780789386 Chr.8: 23007190-23007190 on Build GRCh38
rs753753840 Chr.8: 23007380-23007380 on Build GRCh38
rs756394583 Chr.8: 23007061-23007061 on Build GRCh38
rs538212966 Chr.8: 23007103-23007103 on Build GRCh38
rs751137238 Chr.8: 23007295-23007295 on Build GRCh38
rs992302689 Chr.8: 23007125-23007125 on Build GRCh38
rs764632189 Chr.8: 23007275-23007275 on Build GRCh38
rs754479791 Chr.8: 23007221-23007226 on Build GRCh38
rs780271859 Chr.8: 23007395-23007395 on Build GRCh38
rs769547001 Chr.8: 23007094-23007094 on Build GRCh38
rs114883013 Chr.8: 23007244-23007244 on Build GRCh38
rs763318462 Chr.8: 23007443-23007443 on Build GRCh38
rs773683352 Chr.8: 23007367-23007367 on Build GRCh38
rs768876803 Chr.8: 23007350-23007350 on Build GRCh38
rs559635697 Chr.8: 23007068-23007068 on Build GRCh38
rs747262121 Chr.8: 23007396-23007396 on Build GRCh38
rs201976444 Chr.8: 23007151-23007151 on Build GRCh38
rs1029871876 Chr.8: 23007082-23007082 on Build GRCh38
rs758143293 Chr.8: 23007391-23007391 on Build GRCh38
rs759105470 Chr.8: 23007368-23007368 on Build GRCh38
rs775094999 Chr.8: 23007448-23007448 on Build GRCh38
rs753224986 Chr.8: 23007221-23007235 on Build GRCh38
rs960412645 Chr.8: 23007463-23007463 on Build GRCh38
rs758884291 Chr.8: 23007224-23007232 on Build GRCh38
rs749256694 Chr.8: 23007329-23007329 on Build GRCh38
rs760369594 Chr.8: 23007450-23007450 on Build GRCh38
rs200370658 Chr.8: 23007372-23007372 on Build GRCh38
rs978275158 Chr.8: 23007152-23007152 on Build GRCh38

View Full Product Details