Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 23007808-23008311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHOBTB2
Entrez Gene ID
23221
Gene Name
Rho related BTB domain containing 2
Gene Aliases
DBC2
UniGene
Hs.372688
Gene Chromosome Location
Chr.8: 22987406-23020197 on Build GRCh38
Transcripts
NM_001160036.1 NM_001160037.1 NM_015178.2 XM_017013251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3647324 c.1620+5G>T p.? Chr.8: 23008116-23008116 on Build GRCh38
5804123 c.1502-2A>T p.? Chr.8: 23007991-23007991 on Build GRCh38
1673805 c.1559C>T p.S520F Chr.8: 23008050-23008050 on Build GRCh38
6180338 c.1575G>A p.M525I Chr.8: 23008066-23008066 on Build GRCh38
1496842 c.1534A>G p.S512G Chr.8: 23008025-23008025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567037082 Chr.8: 23008133-23008133 on Build GRCh38
rs1016137302 Chr.8: 23008026-23008026 on Build GRCh38
rs975423011 Chr.8: 23008140-23008140 on Build GRCh38
rs758728268 Chr.8: 23007988-23007988 on Build GRCh38
rs904045865 Chr.8: 23008197-23008197 on Build GRCh38
rs368620278 Chr.8: 23007990-23007990 on Build GRCh38
rs1013263571 Chr.8: 23008276-23008276 on Build GRCh38
rs771981911 Chr.8: 23008042-23008042 on Build GRCh38
rs770532837 Chr.8: 23008138-23008138 on Build GRCh38
rs753795408 Chr.8: 23007974-23007974 on Build GRCh38
rs921181143 Chr.8: 23007864-23007864 on Build GRCh38
rs1012570576 Chr.8: 23007975-23007975 on Build GRCh38
rs748688208 Chr.8: 23008025-23008025 on Build GRCh38
rs529295783 Chr.8: 23007889-23007889 on Build GRCh38
rs140665864 Chr.8: 23008011-23008011 on Build GRCh38
rs772922097 Chr.8: 23008015-23008015 on Build GRCh38
rs367743899 Chr.8: 23007973-23007973 on Build GRCh38
rs896165019 Chr.8: 23007901-23007901 on Build GRCh38
rs904014284 Chr.8: 23007888-23007888 on Build GRCh38
rs549672154 Chr.8: 23007976-23007976 on Build GRCh38
rs539621782 Chr.8: 23008050-23008050 on Build GRCh38
rs935467386 Chr.8: 23008237-23008237 on Build GRCh38
rs779359057 Chr.8: 23008252-23008252 on Build GRCh38
rs367735889 Chr.8: 23008098-23008098 on Build GRCh38
rs1004295774 Chr.8: 23008128-23008128 on Build GRCh38
rs778625577 Chr.8: 23008139-23008139 on Build GRCh38
rs951469502 Chr.8: 23008126-23008126 on Build GRCh38
rs374511325 Chr.8: 23008028-23008028 on Build GRCh38
rs573517777 Chr.8: 23008157-23008157 on Build GRCh38
rs781218251 Chr.8: 23008130-23008131 on Build GRCh38
rs375571104 Chr.8: 23007981-23007981 on Build GRCh38
rs942866317 Chr.8: 23007874-23007874 on Build GRCh38
rs769395531 Chr.8: 23008005-23008005 on Build GRCh38
rs563379104 Chr.8: 23008281-23008281 on Build GRCh38
rs1043869704 Chr.8: 23007877-23007877 on Build GRCh38
rs767450118 Chr.8: 23007945-23007945 on Build GRCh38
rs565855383 Chr.8: 23007978-23007978 on Build GRCh38
rs758959310 Chr.8: 23008118-23008118 on Build GRCh38
rs1023390071 Chr.8: 23007919-23007919 on Build GRCh38
rs934471989 Chr.8: 23008223-23008223 on Build GRCh38
rs765344134 Chr.8: 23008107-23008107 on Build GRCh38
rs545210448 Chr.8: 23008264-23008264 on Build GRCh38
rs747819704 Chr.8: 23008004-23008004 on Build GRCh38
rs553077052 Chr.8: 23008208-23008208 on Build GRCh38
rs377499522 Chr.8: 23007948-23007948 on Build GRCh38
rs984940174 Chr.8: 23008022-23008022 on Build GRCh38
rs889845080 Chr.8: 23007865-23007865 on Build GRCh38
rs571326568 Chr.8: 23008106-23008106 on Build GRCh38
rs911317702 Chr.8: 23008154-23008154 on Build GRCh38
rs924476958 Chr.8: 23008217-23008217 on Build GRCh38
rs577843504 Chr.8: 23008227-23008227 on Build GRCh38
rs371059598 Chr.8: 23008027-23008027 on Build GRCh38
rs972033621 Chr.8: 23008051-23008051 on Build GRCh38
rs140851831 Chr.8: 23008175-23008175 on Build GRCh38
rs371034976 Chr.8: 23007950-23007950 on Build GRCh38
rs1051139456 Chr.8: 23007862-23007862 on Build GRCh38
rs756362680 Chr.8: 23007878-23007878 on Build GRCh38
rs1013232560 Chr.8: 23007907-23007907 on Build GRCh38
rs968627923 Chr.8: 23008189-23008189 on Build GRCh38
rs150986566 Chr.8: 23008097-23008097 on Build GRCh38
rs780458025 Chr.8: 23008125-23008125 on Build GRCh38
rs1056898437 Chr.8: 23007896-23007896 on Build GRCh38
rs547449878 Chr.8: 23007891-23007891 on Build GRCh38
rs371686715 Chr.8: 23008116-23008116 on Build GRCh38
rs868773574 Chr.8: 23008171-23008171 on Build GRCh38
rs934358019 Chr.8: 23007895-23007895 on Build GRCh38
rs764230246 Chr.8: 23007965-23007965 on Build GRCh38
rs979004344 Chr.8: 23008202-23008202 on Build GRCh38
rs73559727 Chr.8: 23008132-23008132 on Build GRCh38
rs746408441 Chr.8: 23007983-23007983 on Build GRCh38

View Full Product Details