Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 26407888-26408385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNIP3L
Entrez Gene ID
665
Gene Name
BCL2 interacting protein 3 like
Gene Aliases
BNIP3a, NIX
UniGene
Hs.131226
Gene Chromosome Location
Chr.8: 26383007-26413128 on Build GRCh38
Transcripts
NM_004331.2 XM_005273617.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6239056 c.471C>T p.H157H Chr.8: 26408236-26408236 on Build GRCh38
5998001 c.436A>G p.S146G Chr.8: 26408078-26408078 on Build GRCh38
2787447 c.453T>C p.I151I Chr.8: 26408095-26408095 on Build GRCh38
6206961 c.407A>G p.K136R Chr.8: 26408049-26408049 on Build GRCh38
2787449 c.488G>A p.R163H Chr.8: 26408253-26408253 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918195494 Chr.8: 26408270-26408270 on Build GRCh38
rs767130233 Chr.8: 26408056-26408056 on Build GRCh38
rs766469742 Chr.8: 26408002-26408004 on Build GRCh38
rs772220367 Chr.8: 26408029-26408029 on Build GRCh38
rs866392776 Chr.8: 26408063-26408063 on Build GRCh38
rs367948485 Chr.8: 26407956-26407956 on Build GRCh38
rs200534833 Chr.8: 26408086-26408086 on Build GRCh38
rs771290000 Chr.8: 26408298-26408298 on Build GRCh38
rs762423471 Chr.8: 26408045-26408045 on Build GRCh38
rs150321619 Chr.8: 26408116-26408116 on Build GRCh38
rs753890693 Chr.8: 26408017-26408024 on Build GRCh38
rs775762260 Chr.8: 26408030-26408030 on Build GRCh38
rs14994 Chr.8: 26408311-26408311 on Build GRCh38
rs758200966 Chr.8: 26408117-26408117 on Build GRCh38
rs369015422 Chr.8: 26408358-26408358 on Build GRCh38
rs777644458 Chr.8: 26408293-26408293 on Build GRCh38
rs760342858 Chr.8: 26408178-26408178 on Build GRCh38
rs767160183 Chr.8: 26407962-26407962 on Build GRCh38
rs774517642 Chr.8: 26408153-26408153 on Build GRCh38
rs770467255 Chr.8: 26408148-26408148 on Build GRCh38
rs774809502 Chr.8: 26408300-26408300 on Build GRCh38
rs968022295 Chr.8: 26408057-26408057 on Build GRCh38
rs369179592 Chr.8: 26408116-26408117 on Build GRCh38
rs773482295 Chr.8: 26408055-26408055 on Build GRCh38
rs747118549 Chr.8: 26408036-26408036 on Build GRCh38
rs201574024 Chr.8: 26408035-26408035 on Build GRCh38
rs751267986 Chr.8: 26408244-26408244 on Build GRCh38
rs564581632 Chr.8: 26408172-26408172 on Build GRCh38
rs778697827 Chr.8: 26408017-26408017 on Build GRCh38
rs113593460 Chr.8: 26408151-26408151 on Build GRCh38
rs373938692 Chr.8: 26408312-26408312 on Build GRCh38
rs757124295 Chr.8: 26408013-26408013 on Build GRCh38
rs575926839 Chr.8: 26408220-26408220 on Build GRCh38
rs777680555 Chr.8: 26408201-26408202 on Build GRCh38
rs556740718 Chr.8: 26407946-26407946 on Build GRCh38
rs753386018 Chr.8: 26408202-26408202 on Build GRCh38
rs747260723 Chr.8: 26408139-26408139 on Build GRCh38
rs757216151 Chr.8: 26408093-26408093 on Build GRCh38
rs570862981 Chr.8: 26407930-26407930 on Build GRCh38
rs746836011 Chr.8: 26408252-26408253 on Build GRCh38
rs368261648 Chr.8: 26408147-26408147 on Build GRCh38
rs138001749 Chr.8: 26408152-26408152 on Build GRCh38
rs532017041 Chr.8: 26408222-26408222 on Build GRCh38
rs867912249 Chr.8: 26408354-26408354 on Build GRCh38
rs761289218 Chr.8: 26408208-26408208 on Build GRCh38
rs140691550 Chr.8: 26408288-26408288 on Build GRCh38
rs901944191 Chr.8: 26407922-26407922 on Build GRCh38
rs752387592 Chr.8: 26408060-26408060 on Build GRCh38
rs776876975 Chr.8: 26408144-26408144 on Build GRCh38
rs755634876 Chr.8: 26407972-26407972 on Build GRCh38
rs764809129 Chr.8: 26408328-26408328 on Build GRCh38
rs932465094 Chr.8: 26408287-26408287 on Build GRCh38
rs778789585 Chr.8: 26408095-26408095 on Build GRCh38
rs763603270 Chr.8: 26408067-26408067 on Build GRCh38
rs372224652 Chr.8: 26408026-26408026 on Build GRCh38
rs749811787 Chr.8: 26408297-26408297 on Build GRCh38
rs1036247570 Chr.8: 26408100-26408100 on Build GRCh38
rs139026759 Chr.8: 26408236-26408236 on Build GRCh38
rs372154844 Chr.8: 26407961-26407961 on Build GRCh38
rs763695305 Chr.8: 26408189-26408189 on Build GRCh38
rs150078014 Chr.8: 26408065-26408065 on Build GRCh38
rs761664614 Chr.8: 26407979-26407979 on Build GRCh38
rs745554487 Chr.8: 26407987-26407987 on Build GRCh38
rs539804286 Chr.8: 26407938-26407938 on Build GRCh38
rs759141869 Chr.8: 26408303-26408303 on Build GRCh38
rs749845213 Chr.8: 26408347-26408347 on Build GRCh38
rs572517609 Chr.8: 26408135-26408135 on Build GRCh38
rs748289954 Chr.8: 26408289-26408289 on Build GRCh38
rs749228161 Chr.8: 26408000-26408000 on Build GRCh38
rs148067521 Chr.8: 26407927-26407927 on Build GRCh38
rs901751908 Chr.8: 26408113-26408114 on Build GRCh38
rs755286076 Chr.8: 26408253-26408253 on Build GRCh38
rs756249417 Chr.8: 26407987-26407989 on Build GRCh38
rs991166974 Chr.8: 26408133-26408133 on Build GRCh38
rs576584079 Chr.8: 26408040-26408040 on Build GRCh38
rs376715919 Chr.8: 26408241-26408241 on Build GRCh38
rs200081637 Chr.8: 26408314-26408314 on Build GRCh38
rs752195320 Chr.8: 26407970-26407970 on Build GRCh38
rs908467401 Chr.8: 26408166-26408166 on Build GRCh38
rs765790935 Chr.8: 26408052-26408052 on Build GRCh38
rs943926993 Chr.8: 26408182-26408182 on Build GRCh38

View Full Product Details