Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144500722-144501261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101928953
Entrez Gene ID
101928953
Gene Name
uncharacterized LOC101928953
Gene Aliases
-
UniGene
Hs.521937
Gene Chromosome Location
Chr.8: 144497357-144505474 on Build GRCh38
Transcripts
XM_005272350.2 XM_011517349.2 XM_017013912.1 XM_017013913.1 XM_017013914.1 XM_017013915.1

Gene Symbol
PPP1R16A
Entrez Gene ID
84988
Gene Name
protein phosphatase 1 regulatory subunit 16A
Gene Aliases
MYPT3
UniGene
Hs.521937
Gene Chromosome Location
Chr.8: 144477987-144502121 on Build GRCh38
Transcripts
XM_005272350.2 XM_011517349.2 XM_017013912.1 XM_017013913.1 XM_017013914.1 XM_017013915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4885269 c.1051C>T p.R351W Chr.8: 144501142-144501142 on Build GRCh38
1659776 c.1156_1158delAAC p.N386delN Chr.8: 144501247-144501249 on Build GRCh38
6374531 c.1052G>A p.R351Q Chr.8: 144501143-144501143 on Build GRCh38
3669806 c.985A>T p.S329C Chr.8: 144500919-144500919 on Build GRCh38
5976752 c.1163A>C p.D388A Chr.8: 144501254-144501254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868763978 Chr.8: 144501155-144501155 on Build GRCh38
rs992920418 Chr.8: 144500786-144500786 on Build GRCh38
rs548200227 Chr.8: 144501162-144501162 on Build GRCh38
rs763054355 Chr.8: 144500862-144500862 on Build GRCh38
rs375095062 Chr.8: 144501116-144501116 on Build GRCh38
rs112902507 Chr.8: 144500878-144500878 on Build GRCh38
rs767715606 Chr.8: 144500793-144500793 on Build GRCh38
rs1006585311 Chr.8: 144500889-144500889 on Build GRCh38
rs374889154 Chr.8: 144500835-144500835 on Build GRCh38
rs756403978 Chr.8: 144500801-144500801 on Build GRCh38
rs538807552 Chr.8: 144501008-144501008 on Build GRCh38
rs754271637 Chr.8: 144501118-144501120 on Build GRCh38
rs1038698903 Chr.8: 144500907-144500907 on Build GRCh38
rs890117662 Chr.8: 144500901-144500901 on Build GRCh38
rs925866151 Chr.8: 144501201-144501201 on Build GRCh38
rs775507923 Chr.8: 144500861-144500861 on Build GRCh38
rs771950529 Chr.8: 144501166-144501166 on Build GRCh38
rs753197453 Chr.8: 144500905-144500905 on Build GRCh38
rs758243896 Chr.8: 144501236-144501236 on Build GRCh38
rs776527086 Chr.8: 144500777-144500777 on Build GRCh38
rs925759582 Chr.8: 144500998-144500998 on Build GRCh38
rs911771887 Chr.8: 144501019-144501019 on Build GRCh38
rs765214755 Chr.8: 144500782-144500782 on Build GRCh38
rs372762824 Chr.8: 144501144-144501144 on Build GRCh38
rs915949348 Chr.8: 144500788-144500788 on Build GRCh38
rs577835978 Chr.8: 144501000-144501000 on Build GRCh38
rs775628935 Chr.8: 144501189-144501189 on Build GRCh38
rs777241995 Chr.8: 144500918-144500918 on Build GRCh38
rs371451014 Chr.8: 144500748-144500748 on Build GRCh38
rs766585665 Chr.8: 144500892-144500892 on Build GRCh38
rs756341516 Chr.8: 144501108-144501112 on Build GRCh38
rs532313058 Chr.8: 144500911-144500911 on Build GRCh38
rs765331413 Chr.8: 144501022-144501038 on Build GRCh38
rs938471863 Chr.8: 144500815-144500815 on Build GRCh38
rs765057110 Chr.8: 144501231-144501231 on Build GRCh38
rs777329738 Chr.8: 144500829-144500829 on Build GRCh38
rs368470587 Chr.8: 144501142-144501142 on Build GRCh38
rs529923184 Chr.8: 144501146-144501146 on Build GRCh38
rs761914169 Chr.8: 144500792-144500792 on Build GRCh38
rs888637150 Chr.8: 144500934-144500934 on Build GRCh38
rs763232780 Chr.8: 144501121-144501121 on Build GRCh38
rs868784403 Chr.8: 144500874-144500874 on Build GRCh38
rs754789116 Chr.8: 144501156-144501156 on Build GRCh38
rs527686200 Chr.8: 144500770-144500770 on Build GRCh38
rs542613881 Chr.8: 144500778-144500778 on Build GRCh38
rs749906793 Chr.8: 144501143-144501143 on Build GRCh38
rs374721145 Chr.8: 144500759-144500759 on Build GRCh38
rs749729638 Chr.8: 144500746-144500746 on Build GRCh38
rs760205268 Chr.8: 144501220-144501220 on Build GRCh38
rs930359114 Chr.8: 144501171-144501171 on Build GRCh38
rs745908236 Chr.8: 144501177-144501177 on Build GRCh38
rs779790078 Chr.8: 144501145-144501145 on Build GRCh38
rs780539557 Chr.8: 144500744-144500744 on Build GRCh38
rs201113193 Chr.8: 144501095-144501095 on Build GRCh38
rs554454561 Chr.8: 144501017-144501017 on Build GRCh38
rs771039469 Chr.8: 144501109-144501109 on Build GRCh38
rs749054711 Chr.8: 144501092-144501092 on Build GRCh38
rs767457007 Chr.8: 144501065-144501065 on Build GRCh38
rs750682428 Chr.8: 144501101-144501104 on Build GRCh38
rs776612392 Chr.8: 144501113-144501113 on Build GRCh38
rs934552099 Chr.8: 144500873-144500873 on Build GRCh38
rs963522755 Chr.8: 144500993-144500993 on Build GRCh38
rs774615837 Chr.8: 144501208-144501208 on Build GRCh38
rs1005531627 Chr.8: 144501087-144501087 on Build GRCh38
rs755326106 Chr.8: 144501194-144501195 on Build GRCh38
rs930368592 Chr.8: 144501031-144501031 on Build GRCh38
rs774189700 Chr.8: 144501042-144501042 on Build GRCh38
rs550030859 Chr.8: 144500880-144500880 on Build GRCh38
rs561474610 Chr.8: 144500810-144500810 on Build GRCh38
rs773863932 Chr.8: 144501218-144501218 on Build GRCh38
rs780622928 Chr.8: 144501077-144501077 on Build GRCh38
rs138377500 Chr.8: 144501227-144501227 on Build GRCh38
rs764787175 Chr.8: 144500818-144500818 on Build GRCh38
rs376968770 Chr.8: 144500767-144500767 on Build GRCh38
rs992622619 Chr.8: 144501023-144501023 on Build GRCh38
rs773394933 Chr.8: 144500869-144500869 on Build GRCh38
rs765553302 Chr.8: 144501059-144501059 on Build GRCh38
rs779327694 Chr.8: 144500813-144500813 on Build GRCh38
rs754628321 Chr.8: 144501230-144501230 on Build GRCh38
rs760741326 Chr.8: 144501045-144501045 on Build GRCh38
rs754011368 Chr.8: 144500894-144500894 on Build GRCh38
rs200051514 Chr.8: 144501210-144501210 on Build GRCh38
rs536732113 Chr.8: 144500981-144500981 on Build GRCh38
rs1015835201 Chr.8: 144500947-144500947 on Build GRCh38
rs764375709 Chr.8: 144501125-144501125 on Build GRCh38
rs994607240 Chr.8: 144500929-144500929 on Build GRCh38
rs547236150 Chr.8: 144501124-144501124 on Build GRCh38
rs779419372 Chr.8: 144500757-144500757 on Build GRCh38
rs755883296 Chr.8: 144501081-144501081 on Build GRCh38
rs768576455 Chr.8: 144501093-144501093 on Build GRCh38
rs921590468 Chr.8: 144501165-144501165 on Build GRCh38
rs779403069 Chr.8: 144501231-144501242 on Build GRCh38
rs777786200 Chr.8: 144501176-144501176 on Build GRCh38
rs1039080941 Chr.8: 144501028-144501028 on Build GRCh38
rs565790019 Chr.8: 144500933-144500933 on Build GRCh38
rs553528646 Chr.8: 144500903-144500903 on Build GRCh38
rs970518854 Chr.8: 144500970-144500970 on Build GRCh38
rs933735581 Chr.8: 144501241-144501241 on Build GRCh38
rs769713014 Chr.8: 144500860-144500860 on Build GRCh38
rs886727047 Chr.8: 144501083-144501083 on Build GRCh38
rs769057783 Chr.8: 144501202-144501202 on Build GRCh38
rs900192228 Chr.8: 144501029-144501029 on Build GRCh38
rs776169645 Chr.8: 144500837-144500837 on Build GRCh38
rs763330190 Chr.8: 144501066-144501066 on Build GRCh38
rs970108986 Chr.8: 144501096-144501096 on Build GRCh38
rs766632606 Chr.8: 144500803-144500803 on Build GRCh38
rs770779620 Chr.8: 144500805-144500806 on Build GRCh38
rs749566823 Chr.8: 144501199-144501199 on Build GRCh38
rs559645042 Chr.8: 144501126-144501126 on Build GRCh38
rs113246685 Chr.8: 144500999-144500999 on Build GRCh38
rs558195542 Chr.8: 144500997-144500997 on Build GRCh38
rs958143655 Chr.8: 144501004-144501004 on Build GRCh38
rs1033743562 Chr.8: 144500863-144500863 on Build GRCh38
rs569440310 Chr.8: 144500977-144500977 on Build GRCh38
rs748669302 Chr.8: 144500822-144500822 on Build GRCh38
rs779879908 Chr.8: 144501084-144501084 on Build GRCh38
rs142958733 Chr.8: 144501085-144501085 on Build GRCh38
rs753107681 Chr.8: 144501063-144501063 on Build GRCh38
rs767874822 Chr.8: 144501216-144501216 on Build GRCh38
rs979965984 Chr.8: 144500776-144500776 on Build GRCh38
rs367883975 Chr.8: 144500867-144500867 on Build GRCh38
rs1022987055 Chr.8: 144500961-144500961 on Build GRCh38
rs1016670428 Chr.8: 144500984-144500984 on Build GRCh38
rs370120097 Chr.8: 144500832-144500832 on Build GRCh38
rs376008663 Chr.8: 144501181-144501181 on Build GRCh38
rs978840009 Chr.8: 144501111-144501111 on Build GRCh38
rs754184671 Chr.8: 144500806-144500806 on Build GRCh38
rs770627758 Chr.8: 144500771-144500771 on Build GRCh38
rs865924992 Chr.8: 144500914-144500914 on Build GRCh38
rs770930250 Chr.8: 144501047-144501047 on Build GRCh38
rs746477697 Chr.8: 144500930-144500930 on Build GRCh38
rs1043114472 Chr.8: 144500859-144500859 on Build GRCh38
rs531841125 Chr.8: 144500864-144500864 on Build GRCh38
rs752668706 Chr.8: 144501041-144501042 on Build GRCh38
rs749218452 Chr.8: 144500983-144500983 on Build GRCh38
rs772794949 Chr.8: 144500768-144500768 on Build GRCh38
rs200594000 Chr.8: 144501080-144501080 on Build GRCh38
rs111341162 Chr.8: 144501039-144501039 on Build GRCh38
rs780826085 Chr.8: 144500945-144500945 on Build GRCh38
rs900225562 Chr.8: 144500922-144500922 on Build GRCh38
rs745631565 Chr.8: 144500849-144500849 on Build GRCh38
rs757892583 Chr.8: 144500828-144500828 on Build GRCh38
rs993200829 Chr.8: 144500960-144500960 on Build GRCh38
rs1871544 Chr.8: 144500807-144500807 on Build GRCh38
rs764620727 Chr.8: 144501068-144501068 on Build GRCh38
rs756169975 Chr.8: 144501240-144501240 on Build GRCh38
rs542669955 Chr.8: 144500833-144500835 on Build GRCh38
rs1016289572 Chr.8: 144501094-144501094 on Build GRCh38
rs780294492 Chr.8: 144501109-144501115 on Build GRCh38
rs762290090 Chr.8: 144501133-144501133 on Build GRCh38
rs572639218 Chr.8: 144501048-144501048 on Build GRCh38
rs746957997 Chr.8: 144501237-144501237 on Build GRCh38
rs750673373 Chr.8: 144500795-144500795 on Build GRCh38
rs1871543 Chr.8: 144500774-144500774 on Build GRCh38
rs775597747 Chr.8: 144500791-144500791 on Build GRCh38
rs866915441 Chr.8: 144500865-144500865 on Build GRCh38
rs761989809 Chr.8: 144501056-144501057 on Build GRCh38
rs960108240 Chr.8: 144501123-144501123 on Build GRCh38
rs547692332 Chr.8: 144500932-144500932 on Build GRCh38
rs755279250 Chr.8: 144500898-144500898 on Build GRCh38
rs201590136 Chr.8: 144501115-144501115 on Build GRCh38
rs752383291 Chr.8: 144501234-144501234 on Build GRCh38
rs776337762 Chr.8: 144500813-144500813 on Build GRCh38
rs1002265421 Chr.8: 144500819-144500819 on Build GRCh38
rs561106229 Chr.8: 144501069-144501069 on Build GRCh38
rs988619542 Chr.8: 144501129-144501129 on Build GRCh38
rs944658257 Chr.8: 144501020-144501020 on Build GRCh38
rs4378003 Chr.8: 144501163-144501163 on Build GRCh38
rs59798065 Chr.8: 144500951-144500951 on Build GRCh38
rs554263341 Chr.8: 144500966-144500966 on Build GRCh38
rs543057022 Chr.8: 144501053-144501053 on Build GRCh38
rs775719240 Chr.8: 144501120-144501120 on Build GRCh38

View Full Product Details