Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 69174355-69174863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TJP2
Entrez Gene ID
9414
Gene Name
tight junction protein 2
Gene Aliases
C9DUPq21.11, DFNA51, DUP9q21.11, PFIC4, X104, ZO2
UniGene
Hs.50382
Gene Chromosome Location
Chr.9: 69099907-69255208 on Build GRCh38
Transcripts
NM_001170414.2 NM_004817.3 NM_201629.3 XM_011519204.1 XM_011519206.2 XM_011519207.2 XM_011519208.2 XM_017015326.1 XM_017015327.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4828889 c.58C>T p.R20C Chr.9: 69174430-69174430 on Build GRCh38
6406116 c.37C>T p.R13W Chr.9: 69174409-69174409 on Build GRCh38
4628888 c.8T>A p.V3E Chr.9: 69174380-69174380 on Build GRCh38
4628889 c.8T>A p.V3E Chr.9: 69174380-69174380 on Build GRCh38
4828890 c.58C>T p.R20C Chr.9: 69174430-69174430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041744676 Chr.9: 69174753-69174753 on Build GRCh38
rs906872124 Chr.9: 69174758-69174758 on Build GRCh38
rs943396550 Chr.9: 69174565-69174565 on Build GRCh38
rs1018924290 Chr.9: 69174751-69174751 on Build GRCh38
rs1035149078 Chr.9: 69174604-69174604 on Build GRCh38
rs952257505 Chr.9: 69174827-69174827 on Build GRCh38
rs536084693 Chr.9: 69174694-69174694 on Build GRCh38
rs766935915 Chr.9: 69174467-69174467 on Build GRCh38
rs59568063 Chr.9: 69174486-69174487 on Build GRCh38
rs147512775 Chr.9: 69174470-69174470 on Build GRCh38
rs765192624 Chr.9: 69174779-69174782 on Build GRCh38
rs546915220 Chr.9: 69174640-69174640 on Build GRCh38
rs779063663 Chr.9: 69174376-69174376 on Build GRCh38
rs144607139 Chr.9: 69174470-69174471 on Build GRCh38
rs1054322307 Chr.9: 69174707-69174707 on Build GRCh38
rs185737699 Chr.9: 69174794-69174794 on Build GRCh38
rs1028141142 Chr.9: 69174649-69174649 on Build GRCh38
rs552174032 Chr.9: 69174493-69174493 on Build GRCh38
rs907902896 Chr.9: 69174685-69174685 on Build GRCh38
rs931826982 Chr.9: 69174702-69174702 on Build GRCh38
rs1039258282 Chr.9: 69174405-69174405 on Build GRCh38
rs1003777470 Chr.9: 69174773-69174773 on Build GRCh38
rs1014686796 Chr.9: 69174822-69174822 on Build GRCh38
rs377762494 Chr.9: 69174427-69174427 on Build GRCh38
rs796416981 Chr.9: 69174816-69174816 on Build GRCh38
rs117876260 Chr.9: 69174797-69174797 on Build GRCh38
rs1056032839 Chr.9: 69174635-69174635 on Build GRCh38
rs928154179 Chr.9: 69174382-69174382 on Build GRCh38
rs930653227 Chr.9: 69174406-69174406 on Build GRCh38
rs900351259 Chr.9: 69174490-69174490 on Build GRCh38
rs12378399 Chr.9: 69174647-69174647 on Build GRCh38
rs967360294 Chr.9: 69174596-69174596 on Build GRCh38
rs945854289 Chr.9: 69174748-69174748 on Build GRCh38
rs930523968 Chr.9: 69174410-69174410 on Build GRCh38
rs868216630 Chr.9: 69174487-69174487 on Build GRCh38
rs911915913 Chr.9: 69174544-69174544 on Build GRCh38
rs1010301662 Chr.9: 69174553-69174553 on Build GRCh38
rs967014248 Chr.9: 69174378-69174378 on Build GRCh38
rs760119569 Chr.9: 69174482-69174482 on Build GRCh38
rs66778410 Chr.9: 69174776-69174779 on Build GRCh38
rs202219041 Chr.9: 69174777-69174777 on Build GRCh38
rs764247013 Chr.9: 69174519-69174519 on Build GRCh38
rs1000347954 Chr.9: 69174698-69174698 on Build GRCh38
rs773854159 Chr.9: 69174440-69174440 on Build GRCh38
rs907766634 Chr.9: 69174462-69174462 on Build GRCh38
rs373291069 Chr.9: 69174395-69174395 on Build GRCh38
rs58801814 Chr.9: 69174777-69174780 on Build GRCh38
rs758381001 Chr.9: 69174379-69174379 on Build GRCh38
rs921744690 Chr.9: 69174701-69174701 on Build GRCh38
rs903638418 Chr.9: 69174670-69174670 on Build GRCh38
rs1031858926 Chr.9: 69174709-69174709 on Build GRCh38
rs920712043 Chr.9: 69174578-69174578 on Build GRCh38
rs747465781 Chr.9: 69174401-69174401 on Build GRCh38
rs987726986 Chr.9: 69174743-69174743 on Build GRCh38
rs573008982 Chr.9: 69174613-69174613 on Build GRCh38
rs766675583 Chr.9: 69174649-69174650 on Build GRCh38
rs770306033 Chr.9: 69174437-69174437 on Build GRCh38
rs1023513709 Chr.9: 69174500-69174500 on Build GRCh38
rs1011968042 Chr.9: 69174483-69174483 on Build GRCh38
rs373062178 Chr.9: 69174778-69174778 on Build GRCh38
rs920672695 Chr.9: 69174825-69174825 on Build GRCh38
rs369175124 Chr.9: 69174377-69174377 on Build GRCh38
rs886044200 Chr.9: 69174391-69174391 on Build GRCh38
rs557230975 Chr.9: 69174706-69174706 on Build GRCh38
rs1040175226 Chr.9: 69174486-69174486 on Build GRCh38
rs760767882 Chr.9: 69174508-69174508 on Build GRCh38
rs944590593 Chr.9: 69174690-69174690 on Build GRCh38
rs939284568 Chr.9: 69174629-69174629 on Build GRCh38
rs983855431 Chr.9: 69174675-69174675 on Build GRCh38
rs570478128 Chr.9: 69174598-69174598 on Build GRCh38
rs151034139 Chr.9: 69174476-69174477 on Build GRCh38
rs1022035164 Chr.9: 69174664-69174664 on Build GRCh38
rs968919908 Chr.9: 69174502-69174502 on Build GRCh38
rs1048045723 Chr.9: 69174599-69174599 on Build GRCh38
rs779935442 Chr.9: 69174775-69174776 on Build GRCh38
rs759277222 Chr.9: 69174456-69174456 on Build GRCh38
rs145066365 Chr.9: 69174404-69174404 on Build GRCh38
rs568654226 Chr.9: 69174687-69174687 on Build GRCh38
rs141127141 Chr.9: 69174409-69174409 on Build GRCh38
rs12378413 Chr.9: 69174717-69174717 on Build GRCh38
rs894815971 Chr.9: 69174639-69174639 on Build GRCh38
rs979371448 Chr.9: 69174507-69174507 on Build GRCh38
rs200373123 Chr.9: 69174779-69174779 on Build GRCh38
rs974864215 Chr.9: 69174569-69174569 on Build GRCh38
rs990874962 Chr.9: 69174625-69174625 on Build GRCh38
rs752479875 Chr.9: 69174648-69174648 on Build GRCh38
rs1003144509 Chr.9: 69174443-69174443 on Build GRCh38
rs193088063 Chr.9: 69174602-69174602 on Build GRCh38
rs987442284 Chr.9: 69174531-69174531 on Build GRCh38
rs557804054 Chr.9: 69174798-69174798 on Build GRCh38
rs959549036 Chr.9: 69174617-69174617 on Build GRCh38
rs1031918369 Chr.9: 69174509-69174509 on Build GRCh38
rs775823459 Chr.9: 69174468-69174469 on Build GRCh38
rs376941400 Chr.9: 69174738-69174738 on Build GRCh38

View Full Product Details