Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 71704505-71705012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM2
Entrez Gene ID
23670
Gene Name
transmembrane protein 2
Gene Aliases
-
UniGene
Hs.494146
Gene Chromosome Location
Chr.9: 71683366-71769532 on Build GRCh38
Transcripts
NM_001135820.1 NM_013390.2 XM_005251869.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3746036 c.3108T>G p.P1036P Chr.9: 71704681-71704681 on Build GRCh38
1756209 c.3004A>T p.S1002C Chr.9: 71704785-71704785 on Build GRCh38
6803751 c.3183C>T p.V1061V Chr.9: 71704606-71704606 on Build GRCh38
422349 c.3183C>G p.V1061V Chr.9: 71704606-71704606 on Build GRCh38
6882251 c.3047C>T p.P1016L Chr.9: 71704742-71704742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188489792 Chr.9: 71704883-71704883 on Build GRCh38
rs141686995 Chr.9: 71704741-71704741 on Build GRCh38
rs1034990097 Chr.9: 71704865-71704865 on Build GRCh38
rs370020649 Chr.9: 71704722-71704722 on Build GRCh38
rs754993785 Chr.9: 71704812-71704812 on Build GRCh38
rs542402079 Chr.9: 71704630-71704630 on Build GRCh38
rs755028253 Chr.9: 71704568-71704570 on Build GRCh38
rs753501146 Chr.9: 71704656-71704656 on Build GRCh38
rs143044230 Chr.9: 71704636-71704636 on Build GRCh38
rs755383332 Chr.9: 71704649-71704649 on Build GRCh38
rs571527768 Chr.9: 71704927-71704930 on Build GRCh38
rs25696 Chr.9: 71704660-71704660 on Build GRCh38
rs750170498 Chr.9: 71704790-71704790 on Build GRCh38
rs756892498 Chr.9: 71704735-71704735 on Build GRCh38
rs567130967 Chr.9: 71704830-71704830 on Build GRCh38
rs375145196 Chr.9: 71704742-71704742 on Build GRCh38
rs750023060 Chr.9: 71704739-71704739 on Build GRCh38
rs77085527 Chr.9: 71704981-71704981 on Build GRCh38
rs751940281 Chr.9: 71704646-71704646 on Build GRCh38
rs546956745 Chr.9: 71704622-71704622 on Build GRCh38
rs752099941 Chr.9: 71704721-71704721 on Build GRCh38
rs778580201 Chr.9: 71704736-71704736 on Build GRCh38
rs138339130 Chr.9: 71704608-71704608 on Build GRCh38
rs750963349 Chr.9: 71704577-71704577 on Build GRCh38
rs376716032 Chr.9: 71704620-71704620 on Build GRCh38
rs760984312 Chr.9: 71704564-71704564 on Build GRCh38
rs750455411 Chr.9: 71704637-71704637 on Build GRCh38
rs933516359 Chr.9: 71704597-71704597 on Build GRCh38
rs773193645 Chr.9: 71704693-71704693 on Build GRCh38
rs753907220 Chr.9: 71704565-71704565 on Build GRCh38
rs530194808 Chr.9: 71704633-71704633 on Build GRCh38
rs200932802 Chr.9: 71704698-71704698 on Build GRCh38
rs764372807 Chr.9: 71704568-71704568 on Build GRCh38
rs762969493 Chr.9: 71704703-71704703 on Build GRCh38
rs781504360 Chr.9: 71704655-71704655 on Build GRCh38
rs761950665 Chr.9: 71704687-71704687 on Build GRCh38
rs774400522 Chr.9: 71704766-71704766 on Build GRCh38
rs780386524 Chr.9: 71704787-71704787 on Build GRCh38
rs766005010 Chr.9: 71704799-71704799 on Build GRCh38
rs527409490 Chr.9: 71704767-71704767 on Build GRCh38
rs910772302 Chr.9: 71704930-71704930 on Build GRCh38
rs776761274 Chr.9: 71704631-71704631 on Build GRCh38
rs898880311 Chr.9: 71704650-71704650 on Build GRCh38
rs755791571 Chr.9: 71704599-71704599 on Build GRCh38
rs758830377 Chr.9: 71704642-71704642 on Build GRCh38
rs149448448 Chr.9: 71704613-71704613 on Build GRCh38
rs747305221 Chr.9: 71704592-71704592 on Build GRCh38
rs761152828 Chr.9: 71704783-71704783 on Build GRCh38
rs780849860 Chr.9: 71704755-71704755 on Build GRCh38
rs755976285 Chr.9: 71704836-71704836 on Build GRCh38
rs753997230 Chr.9: 71704572-71704572 on Build GRCh38
rs778291603 Chr.9: 71704661-71704661 on Build GRCh38
rs547044472 Chr.9: 71704802-71704802 on Build GRCh38
rs1041882969 Chr.9: 71704593-71704593 on Build GRCh38
rs768029292 Chr.9: 71704770-71704770 on Build GRCh38
rs763525494 Chr.9: 71704604-71704604 on Build GRCh38
rs943626175 Chr.9: 71704960-71704960 on Build GRCh38
rs767926473 Chr.9: 71704733-71704733 on Build GRCh38
rs747181594 Chr.9: 71704835-71704835 on Build GRCh38
rs765709720 Chr.9: 71704595-71704597 on Build GRCh38
rs767281430 Chr.9: 71704554-71704554 on Build GRCh38
rs150698902 Chr.9: 71704678-71704678 on Build GRCh38
rs978104691 Chr.9: 71704863-71704863 on Build GRCh38
rs17057133 Chr.9: 71704761-71704761 on Build GRCh38
rs763962057 Chr.9: 71704629-71704629 on Build GRCh38
rs775367096 Chr.9: 71704562-71704562 on Build GRCh38
rs776478152 Chr.9: 71704683-71704683 on Build GRCh38
rs775458927 Chr.9: 71704625-71704625 on Build GRCh38
rs753026946 Chr.9: 71704772-71704772 on Build GRCh38
rs769988634 Chr.9: 71704688-71704688 on Build GRCh38
rs916734208 Chr.9: 71704939-71704939 on Build GRCh38
rs892978799 Chr.9: 71704793-71704793 on Build GRCh38
rs373540675 Chr.9: 71704734-71704734 on Build GRCh38
rs772159983 Chr.9: 71704676-71704676 on Build GRCh38
rs766829965 Chr.9: 71704645-71704645 on Build GRCh38
rs746489559 Chr.9: 71704674-71704674 on Build GRCh38
rs780405519 Chr.9: 71704584-71704584 on Build GRCh38
rs773906205 Chr.9: 71704617-71704617 on Build GRCh38
rs12339216 Chr.9: 71704875-71704875 on Build GRCh38
rs559006853 Chr.9: 71704967-71704967 on Build GRCh38
rs780192646 Chr.9: 71704677-71704677 on Build GRCh38
rs372416639 Chr.9: 71704609-71704609 on Build GRCh38
rs569880205 Chr.9: 71704891-71704891 on Build GRCh38
rs760499289 Chr.9: 71704628-71704628 on Build GRCh38
rs758114022 Chr.9: 71704796-71704796 on Build GRCh38
rs763129325 Chr.9: 71704764-71704764 on Build GRCh38
rs770386937 Chr.9: 71704615-71704615 on Build GRCh38
rs781132489 Chr.9: 71704817-71704817 on Build GRCh38
rs977522428 Chr.9: 71704841-71704841 on Build GRCh38
rs193148555 Chr.9: 71704966-71704966 on Build GRCh38
rs757333064 Chr.9: 71704574-71704574 on Build GRCh38
rs748202087 Chr.9: 71704984-71704984 on Build GRCh38
rs768660270 Chr.9: 71704750-71704750 on Build GRCh38
rs759485734 Chr.9: 71704545-71704545 on Build GRCh38
rs766422778 Chr.9: 71704718-71704718 on Build GRCh38
rs757790702 Chr.9: 71704669-71704669 on Build GRCh38
rs747004409 Chr.9: 71704745-71704745 on Build GRCh38
rs1028798067 Chr.9: 71704782-71704782 on Build GRCh38
rs1040311179 Chr.9: 71704746-71704746 on Build GRCh38
rs924819356 Chr.9: 71704595-71704595 on Build GRCh38
rs758921939 Chr.9: 71704581-71704581 on Build GRCh38

View Full Product Details