Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 135484358-135484864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R26
Entrez Gene ID
9858
Gene Name
protein phosphatase 1 regulatory subunit 26
Gene Aliases
KIAA0649
UniGene
Hs.533260
Gene Chromosome Location
Chr.9: 135477650-135488894 on Build GRCh38
Transcripts
NM_014811.3 XM_005263411.2 XM_017015355.1 XM_017015356.1 XM_017015357.1 XM_017015358.1 XM_017015359.1 XM_017015360.1 XM_017015361.1 XM_017015362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5563082 c.282C>T p.P94P Chr.9: 135484792-135484792 on Build GRCh38
6754045 c.126G>A p.S42S Chr.9: 135484636-135484636 on Build GRCh38
7385256 c.316G>A p.D106N Chr.9: 135484826-135484826 on Build GRCh38
2152423 c.201C>T p.R67R Chr.9: 135484711-135484711 on Build GRCh38
3848007 c.47G>T p.W16L Chr.9: 135484557-135484557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375238310 Chr.9: 135484646-135484646 on Build GRCh38
rs61731943 Chr.9: 135484687-135484687 on Build GRCh38
rs893205769 Chr.9: 135484443-135484443 on Build GRCh38
rs368903595 Chr.9: 135484633-135484633 on Build GRCh38
rs139821365 Chr.9: 135484526-135484526 on Build GRCh38
rs891920917 Chr.9: 135484407-135484407 on Build GRCh38
rs767896126 Chr.9: 135484505-135484505 on Build GRCh38
rs151324944 Chr.9: 135484702-135484702 on Build GRCh38
rs766213987 Chr.9: 135484714-135484714 on Build GRCh38
rs371177420 Chr.9: 135484747-135484747 on Build GRCh38
rs573605570 Chr.9: 135484564-135484564 on Build GRCh38
rs762337402 Chr.9: 135484594-135484594 on Build GRCh38
rs201939322 Chr.9: 135484813-135484813 on Build GRCh38
rs370325909 Chr.9: 135484478-135484478 on Build GRCh38
rs745420408 Chr.9: 135484489-135484489 on Build GRCh38
rs771986591 Chr.9: 135484760-135484760 on Build GRCh38
rs370626672 Chr.9: 135484501-135484501 on Build GRCh38
rs142421416 Chr.9: 135484775-135484775 on Build GRCh38
rs1011683077 Chr.9: 135484434-135484434 on Build GRCh38
rs750530117 Chr.9: 135484508-135484508 on Build GRCh38
rs755736163 Chr.9: 135484676-135484676 on Build GRCh38
rs762925049 Chr.9: 135484620-135484620 on Build GRCh38
rs775569722 Chr.9: 135484703-135484703 on Build GRCh38
rs142346595 Chr.9: 135484692-135484692 on Build GRCh38
rs764789560 Chr.9: 135484734-135484734 on Build GRCh38
rs199673349 Chr.9: 135484820-135484820 on Build GRCh38
rs779063178 Chr.9: 135484462-135484462 on Build GRCh38
rs374673695 Chr.9: 135484484-135484484 on Build GRCh38
rs969618175 Chr.9: 135484681-135484681 on Build GRCh38
rs963098862 Chr.9: 135484649-135484649 on Build GRCh38
rs145020427 Chr.9: 135484647-135484647 on Build GRCh38
rs199947269 Chr.9: 135484631-135484631 on Build GRCh38
rs942848391 Chr.9: 135484439-135484439 on Build GRCh38
rs778999950 Chr.9: 135484828-135484828 on Build GRCh38
rs147210403 Chr.9: 135484544-135484544 on Build GRCh38
rs776097100 Chr.9: 135484488-135484488 on Build GRCh38
rs763376310 Chr.9: 135484492-135484492 on Build GRCh38
rs138434668 Chr.9: 135484770-135484770 on Build GRCh38
rs368057870 Chr.9: 135484491-135484491 on Build GRCh38
rs547817948 Chr.9: 135484384-135484384 on Build GRCh38
rs77110177 Chr.9: 135484711-135484711 on Build GRCh38
rs192524772 Chr.9: 135484543-135484543 on Build GRCh38
rs139951646 Chr.9: 135484678-135484678 on Build GRCh38
rs746113907 Chr.9: 135484698-135484698 on Build GRCh38
rs777430887 Chr.9: 135484480-135484480 on Build GRCh38
rs776503910 Chr.9: 135484788-135484788 on Build GRCh38
rs138505579 Chr.9: 135484804-135484804 on Build GRCh38
rs774750336 Chr.9: 135484592-135484592 on Build GRCh38
rs61731947 Chr.9: 135484642-135484642 on Build GRCh38
rs776739888 Chr.9: 135484643-135484643 on Build GRCh38
rs750038467 Chr.9: 135484629-135484629 on Build GRCh38
rs370197502 Chr.9: 135484814-135484814 on Build GRCh38
rs778257634 Chr.9: 135484689-135484689 on Build GRCh38
rs748170956 Chr.9: 135484557-135484557 on Build GRCh38
rs770105235 Chr.9: 135484753-135484753 on Build GRCh38
rs150799430 Chr.9: 135484572-135484572 on Build GRCh38
rs765018828 Chr.9: 135484792-135484792 on Build GRCh38
rs762608687 Chr.9: 135484802-135484802 on Build GRCh38
rs761718580 Chr.9: 135484709-135484709 on Build GRCh38
rs149244774 Chr.9: 135484835-135484835 on Build GRCh38
rs150450440 Chr.9: 135484751-135484751 on Build GRCh38
rs529637643 Chr.9: 135484758-135484758 on Build GRCh38
rs774076508 Chr.9: 135484712-135484712 on Build GRCh38
rs750268385 Chr.9: 135484668-135484668 on Build GRCh38
rs566152626 Chr.9: 135484403-135484403 on Build GRCh38
rs969188284 Chr.9: 135484796-135484796 on Build GRCh38
rs766583053 Chr.9: 135484774-135484774 on Build GRCh38
rs771061762 Chr.9: 135484622-135484622 on Build GRCh38
rs373599937 Chr.9: 135484606-135484606 on Build GRCh38
rs752608607 Chr.9: 135484519-135484519 on Build GRCh38
rs760891950 Chr.9: 135484513-135484513 on Build GRCh38
rs769096725 Chr.9: 135484573-135484573 on Build GRCh38
rs913202599 Chr.9: 135484836-135484836 on Build GRCh38
rs373929251 Chr.9: 135484521-135484521 on Build GRCh38
rs3748192 Chr.9: 135484803-135484803 on Build GRCh38
rs539940145 Chr.9: 135484436-135484436 on Build GRCh38
rs755674723 Chr.9: 135484811-135484811 on Build GRCh38
rs1002878278 Chr.9: 135484383-135484383 on Build GRCh38
rs145468835 Chr.9: 135484679-135484679 on Build GRCh38
rs375453673 Chr.9: 135484605-135484605 on Build GRCh38
rs201579051 Chr.9: 135484682-135484682 on Build GRCh38
rs766578068 Chr.9: 135484515-135484515 on Build GRCh38
rs139225850 Chr.9: 135484603-135484603 on Build GRCh38
rs746378037 Chr.9: 135484677-135484677 on Build GRCh38
rs1039822682 Chr.9: 135484442-135484442 on Build GRCh38
rs201226416 Chr.9: 135484661-135484661 on Build GRCh38
rs778488719 Chr.9: 135484744-135484744 on Build GRCh38
rs749142844 Chr.9: 135484563-135484563 on Build GRCh38
rs769025041 Chr.9: 135484496-135484496 on Build GRCh38
rs368677537 Chr.9: 135484525-135484525 on Build GRCh38
rs373319471 Chr.9: 135484748-135484748 on Build GRCh38
rs140596223 Chr.9: 135484695-135484695 on Build GRCh38
rs377730970 Chr.9: 135484738-135484738 on Build GRCh38
rs780971585 Chr.9: 135484628-135484628 on Build GRCh38
rs754938428 Chr.9: 135484460-135484460 on Build GRCh38
rs773741384 Chr.9: 135484635-135484635 on Build GRCh38
rs770279912 Chr.9: 135484490-135484490 on Build GRCh38
rs781644488 Chr.9: 135484750-135484750 on Build GRCh38
rs754686666 Chr.9: 135484715-135484715 on Build GRCh38
rs533250185 Chr.9: 135484793-135484793 on Build GRCh38
rs759086841 Chr.9: 135484790-135484790 on Build GRCh38
rs377452619 Chr.9: 135484759-135484759 on Build GRCh38
rs577812668 Chr.9: 135484467-135484470 on Build GRCh38
rs983152198 Chr.9: 135484481-135484481 on Build GRCh38
rs1023801657 Chr.9: 135484794-135484794 on Build GRCh38
rs753883281 Chr.9: 135484616-135484616 on Build GRCh38
rs780887855 Chr.9: 135484535-135484535 on Build GRCh38
rs371967332 Chr.9: 135484527-135484527 on Build GRCh38
rs763978698 Chr.9: 135484621-135484621 on Build GRCh38
rs767484243 Chr.9: 135484710-135484710 on Build GRCh38
rs755787492 Chr.9: 135484630-135484630 on Build GRCh38
rs61731944 Chr.9: 135484651-135484651 on Build GRCh38
rs527406827 Chr.9: 135484625-135484625 on Build GRCh38
rs758151978 Chr.9: 135484479-135484479 on Build GRCh38
rs746628916 Chr.9: 135484482-135484482 on Build GRCh38
rs757806357 Chr.9: 135484808-135484808 on Build GRCh38
rs774601664 Chr.9: 135484497-135484497 on Build GRCh38
rs373419165 Chr.9: 135484636-135484636 on Build GRCh38
rs367729948 Chr.9: 135484632-135484632 on Build GRCh38
rs770588814 Chr.9: 135484777-135484777 on Build GRCh38

View Full Product Details