Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137050395-137050885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD2
Entrez Gene ID
954
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Gene Aliases
CD39L1, NTPDase-2
UniGene
Hs.123036
Gene Chromosome Location
Chr.9: 137048098-137054051 on Build GRCh38
Transcripts
NM_001246.3 NM_203468.2 XM_011519212.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6866980 c.829G>T p.G277W Chr.9: 137050484-137050484 on Build GRCh38
4994054 c.829G>A p.G277R Chr.9: 137050484-137050484 on Build GRCh38
3926476 c.807C>T p.G269G Chr.9: 137050506-137050506 on Build GRCh38
3926475 c.812C>T p.S271F Chr.9: 137050501-137050501 on Build GRCh38
1107024 c.828C>T p.L276L Chr.9: 137050485-137050485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370555871 Chr.9: 137050513-137050513 on Build GRCh38
rs549413347 Chr.9: 137050753-137050753 on Build GRCh38
rs910484356 Chr.9: 137050647-137050647 on Build GRCh38
rs370968571 Chr.9: 137050573-137050573 on Build GRCh38
rs566324002 Chr.9: 137050766-137050766 on Build GRCh38
rs1037086545 Chr.9: 137050477-137050477 on Build GRCh38
rs747460688 Chr.9: 137050496-137050498 on Build GRCh38
rs748902935 Chr.9: 137050475-137050475 on Build GRCh38
rs931370734 Chr.9: 137050771-137050771 on Build GRCh38
rs778135985 Chr.9: 137050463-137050468 on Build GRCh38
rs368559678 Chr.9: 137050863-137050863 on Build GRCh38
rs866672782 Chr.9: 137050516-137050516 on Build GRCh38
rs774134020 Chr.9: 137050426-137050426 on Build GRCh38
rs753979087 Chr.9: 137050441-137050441 on Build GRCh38
rs778033366 Chr.9: 137050755-137050755 on Build GRCh38
rs1002379884 Chr.9: 137050694-137050694 on Build GRCh38
rs371567979 Chr.9: 137050512-137050512 on Build GRCh38
rs760544909 Chr.9: 137050855-137050855 on Build GRCh38
rs1043872286 Chr.9: 137050687-137050687 on Build GRCh38
rs993142652 Chr.9: 137050847-137050847 on Build GRCh38
rs142156660 Chr.9: 137050448-137050448 on Build GRCh38
rs1015839243 Chr.9: 137050746-137050746 on Build GRCh38
rs770836776 Chr.9: 137050861-137050861 on Build GRCh38
rs564022734 Chr.9: 137050636-137050636 on Build GRCh38
rs912478743 Chr.9: 137050715-137050715 on Build GRCh38
rs775013850 Chr.9: 137050474-137050474 on Build GRCh38
rs758600277 Chr.9: 137050455-137050455 on Build GRCh38
rs745407606 Chr.9: 137050470-137050470 on Build GRCh38
rs961723907 Chr.9: 137050598-137050598 on Build GRCh38
rs148619460 Chr.9: 137050533-137050533 on Build GRCh38
rs752765897 Chr.9: 137050515-137050515 on Build GRCh38
rs761747255 Chr.9: 137050427-137050427 on Build GRCh38
rs761587692 Chr.9: 137050496-137050496 on Build GRCh38
rs116205444 Chr.9: 137050670-137050670 on Build GRCh38
rs764362875 Chr.9: 137050537-137050537 on Build GRCh38
rs113129077 Chr.9: 137050556-137050556 on Build GRCh38
rs757054994 Chr.9: 137050794-137050794 on Build GRCh38
rs202014172 Chr.9: 137050549-137050549 on Build GRCh38
rs533388981 Chr.9: 137050668-137050668 on Build GRCh38
rs903899320 Chr.9: 137050688-137050688 on Build GRCh38
rs201314893 Chr.9: 137050570-137050570 on Build GRCh38
rs752018909 Chr.9: 137050544-137050544 on Build GRCh38
rs757895216 Chr.9: 137050461-137050461 on Build GRCh38
rs913918218 Chr.9: 137050675-137050675 on Build GRCh38
rs767496302 Chr.9: 137050507-137050507 on Build GRCh38
rs750985089 Chr.9: 137050553-137050553 on Build GRCh38
rs541869731 Chr.9: 137050592-137050592 on Build GRCh38
rs374694972 Chr.9: 137050555-137050555 on Build GRCh38
rs1033933600 Chr.9: 137050539-137050539 on Build GRCh38
rs2292924 Chr.9: 137050740-137050740 on Build GRCh38
rs757664312 Chr.9: 137050545-137050545 on Build GRCh38
rs771173872 Chr.9: 137050589-137050589 on Build GRCh38
rs368515415 Chr.9: 137050458-137050458 on Build GRCh38
rs558487969 Chr.9: 137050485-137050485 on Build GRCh38
rs767443128 Chr.9: 137050438-137050438 on Build GRCh38
rs763813336 Chr.9: 137050425-137050425 on Build GRCh38
rs781758869 Chr.9: 137050463-137050463 on Build GRCh38
rs771886410 Chr.9: 137050577-137050577 on Build GRCh38
rs570001881 Chr.9: 137050697-137050697 on Build GRCh38
rs927313534 Chr.9: 137050822-137050822 on Build GRCh38
rs139202937 Chr.9: 137050484-137050484 on Build GRCh38
rs985894380 Chr.9: 137050614-137050614 on Build GRCh38
rs7869520 Chr.9: 137050599-137050599 on Build GRCh38
rs146204639 Chr.9: 137050452-137050452 on Build GRCh38
rs967785019 Chr.9: 137050632-137050632 on Build GRCh38
rs778013442 Chr.9: 137050456-137050456 on Build GRCh38
rs533858427 Chr.9: 137050775-137050775 on Build GRCh38
rs891955534 Chr.9: 137050603-137050603 on Build GRCh38
rs773102878 Chr.9: 137050508-137050508 on Build GRCh38
rs191479203 Chr.9: 137050612-137050612 on Build GRCh38
rs535342828 Chr.9: 137050447-137050447 on Build GRCh38
rs896635898 Chr.9: 137050737-137050737 on Build GRCh38
rs945386505 Chr.9: 137050676-137050676 on Build GRCh38
rs143867433 Chr.9: 137050532-137050532 on Build GRCh38
rs267602176 Chr.9: 137050459-137050459 on Build GRCh38
rs146231324 Chr.9: 137050613-137050613 on Build GRCh38
rs376308559 Chr.9: 137050578-137050578 on Build GRCh38
rs866931253 Chr.9: 137050684-137050684 on Build GRCh38
rs769215368 Chr.9: 137050473-137050473 on Build GRCh38
rs1024416026 Chr.9: 137050590-137050590 on Build GRCh38
rs143961573 Chr.9: 137050514-137050514 on Build GRCh38

View Full Product Details