Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137188298-137188788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC2
Entrez Gene ID
29882
Gene Name
anaphase promoting complex subunit 2
Gene Aliases
APC2
UniGene
Hs.533262
Gene Chromosome Location
Chr.9: 137174784-137188605 on Build GRCh38
Transcripts
NM_013366.3 NM_003731.2

Gene Symbol
SSNA1
Entrez Gene ID
8636
Gene Name
SS nuclear autoantigen 1
Gene Aliases
N14, NA-14, NA14
UniGene
Hs.530314
Gene Chromosome Location
Chr.9: 137188602-137190370 on Build GRCh38
Transcripts
NM_013366.3 NM_003731.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3926486 c.22G>T p.A8S Chr.9: 137188511-137188511 on Build GRCh38
6869071 c.51G>A p.K17K Chr.9: 137188777-137188777 on Build GRCh38
3395691 c.57G>T p.E19D Chr.9: 137188476-137188476 on Build GRCh38
1166652 c.20T>C p.V7A Chr.9: 137188513-137188513 on Build GRCh38
7319030 c.11C>T p.A4V Chr.9: 137188522-137188522 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753402373 Chr.9: 137188561-137188561 on Build GRCh38
rs140578056 Chr.9: 137188692-137188693 on Build GRCh38
rs768862405 Chr.9: 137188677-137188677 on Build GRCh38
rs772717022 Chr.9: 137188578-137188578 on Build GRCh38
rs377078426 Chr.9: 137188714-137188714 on Build GRCh38
rs568351324 Chr.9: 137188687-137188687 on Build GRCh38
rs147593956 Chr.9: 137188468-137188468 on Build GRCh38
rs763298617 Chr.9: 137188452-137188452 on Build GRCh38
rs931520178 Chr.9: 137188615-137188615 on Build GRCh38
rs937732433 Chr.9: 137188649-137188649 on Build GRCh38
rs950397175 Chr.9: 137188602-137188625 on Build GRCh38
rs991704075 Chr.9: 137188351-137188351 on Build GRCh38
rs149020460 Chr.9: 137188508-137188508 on Build GRCh38
rs775590656 Chr.9: 137188447-137188447 on Build GRCh38
rs552229574 Chr.9: 137188667-137188667 on Build GRCh38
rs757881118 Chr.9: 137188374-137188374 on Build GRCh38
rs774710497 Chr.9: 137188544-137188544 on Build GRCh38
rs111630032 Chr.9: 137188670-137188670 on Build GRCh38
rs533462404 Chr.9: 137188561-137188562 on Build GRCh38
rs748210377 Chr.9: 137188713-137188713 on Build GRCh38
rs757072290 Chr.9: 137188710-137188710 on Build GRCh38
rs751370399 Chr.9: 137188709-137188709 on Build GRCh38
rs746404561 Chr.9: 137188743-137188743 on Build GRCh38
rs921093042 Chr.9: 137188651-137188651 on Build GRCh38
rs770087721 Chr.9: 137188591-137188591 on Build GRCh38
rs554228076 Chr.9: 137188712-137188712 on Build GRCh38
rs774060070 Chr.9: 137188761-137188761 on Build GRCh38
rs941305907 Chr.9: 137188646-137188646 on Build GRCh38
rs977656247 Chr.9: 137188332-137188332 on Build GRCh38
rs921220053 Chr.9: 137188748-137188748 on Build GRCh38
rs201295555 Chr.9: 137188733-137188733 on Build GRCh38
rs139478852 Chr.9: 137188619-137188619 on Build GRCh38
rs904952388 Chr.9: 137188622-137188622 on Build GRCh38
rs764745728 Chr.9: 137188698-137188698 on Build GRCh38
rs564799824 Chr.9: 137188392-137188392 on Build GRCh38
rs762257507 Chr.9: 137188472-137188472 on Build GRCh38
rs761120734 Chr.9: 137188556-137188556 on Build GRCh38
rs773856619 Chr.9: 137188387-137188387 on Build GRCh38
rs762551561 Chr.9: 137188708-137188708 on Build GRCh38
rs748910799 Chr.9: 137188398-137188398 on Build GRCh38
rs763176176 Chr.9: 137188367-137188367 on Build GRCh38
rs750899636 Chr.9: 137188498-137188498 on Build GRCh38
rs200327224 Chr.9: 137188454-137188454 on Build GRCh38
rs368162981 Chr.9: 137188557-137188557 on Build GRCh38
rs927759046 Chr.9: 137188608-137188608 on Build GRCh38
rs533337955 Chr.9: 137188528-137188528 on Build GRCh38
rs767354119 Chr.9: 137188711-137188711 on Build GRCh38
rs750207161 Chr.9: 137188766-137188766 on Build GRCh38
rs1027771864 Chr.9: 137188577-137188601 on Build GRCh38
rs1051176867 Chr.9: 137188664-137188664 on Build GRCh38
rs535804481 Chr.9: 137188593-137188593 on Build GRCh38
rs777125127 Chr.9: 137188693-137188693 on Build GRCh38
rs770255175 Chr.9: 137188745-137188745 on Build GRCh38
rs538038045 Chr.9: 137188671-137188671 on Build GRCh38
rs768116951 Chr.9: 137188381-137188381 on Build GRCh38
rs1039913809 Chr.9: 137188650-137188650 on Build GRCh38
rs1002272841 Chr.9: 137188340-137188340 on Build GRCh38
rs751337063 Chr.9: 137188581-137188581 on Build GRCh38
rs755973684 Chr.9: 137188720-137188720 on Build GRCh38
rs562403249 Chr.9: 137188550-137188550 on Build GRCh38
rs773015622 Chr.9: 137188431-137188431 on Build GRCh38
rs774122737 Chr.9: 137188655-137188655 on Build GRCh38
rs1033284610 Chr.9: 137188339-137188339 on Build GRCh38
rs779079125 Chr.9: 137188678-137188678 on Build GRCh38
rs960598390 Chr.9: 137188363-137188363 on Build GRCh38
rs764573307 Chr.9: 137188371-137188371 on Build GRCh38
rs369033443 Chr.9: 137188410-137188410 on Build GRCh38
rs957615380 Chr.9: 137188587-137188587 on Build GRCh38
rs898636611 Chr.9: 137188488-137188488 on Build GRCh38
rs774941574 Chr.9: 137188701-137188701 on Build GRCh38
rs968127071 Chr.9: 137188384-137188384 on Build GRCh38
rs772340832 Chr.9: 137188553-137188553 on Build GRCh38
rs142065515 Chr.9: 137188519-137188519 on Build GRCh38
rs548238192 Chr.9: 137188673-137188673 on Build GRCh38
rs1023149257 Chr.9: 137188602-137188602 on Build GRCh38
rs202152762 Chr.9: 137188507-137188507 on Build GRCh38
rs746257396 Chr.9: 137188496-137188496 on Build GRCh38
rs972688325 Chr.9: 137188727-137188727 on Build GRCh38
rs1033253844 Chr.9: 137188586-137188586 on Build GRCh38
rs144698305 Chr.9: 137188327-137188327 on Build GRCh38
rs534184401 Chr.9: 137188361-137188361 on Build GRCh38
rs764656597 Chr.9: 137188565-137188565 on Build GRCh38
rs779554873 Chr.9: 137188394-137188394 on Build GRCh38
rs771404729 Chr.9: 137188688-137188688 on Build GRCh38
rs750614319 Chr.9: 137188719-137188719 on Build GRCh38
rs534042066 Chr.9: 137188697-137188697 on Build GRCh38
rs371712073 Chr.9: 137188566-137188566 on Build GRCh38
rs375516831 Chr.9: 137188349-137188349 on Build GRCh38
rs778048764 Chr.9: 137188715-137188715 on Build GRCh38
rs780036272 Chr.9: 137188534-137188534 on Build GRCh38
rs752355611 Chr.9: 137188571-137188571 on Build GRCh38
rs916281386 Chr.9: 137188352-137188352 on Build GRCh38
rs768206761 Chr.9: 137188478-137188478 on Build GRCh38
rs374208930 Chr.9: 137188426-137188426 on Build GRCh38
rs760691084 Chr.9: 137188708-137188709 on Build GRCh38
rs992026311 Chr.9: 137188598-137188598 on Build GRCh38
rs746894266 Chr.9: 137188432-137188432 on Build GRCh38
rs190485152 Chr.9: 137188513-137188513 on Build GRCh38
rs1009447112 Chr.9: 137188559-137188559 on Build GRCh38
rs747736306 Chr.9: 137188522-137188524 on Build GRCh38
rs769776763 Chr.9: 137188433-137188433 on Build GRCh38
rs1051522894 Chr.9: 137188438-137188438 on Build GRCh38
rs754579062 Chr.9: 137188403-137188403 on Build GRCh38
rs773492650 Chr.9: 137188684-137188684 on Build GRCh38
rs929638504 Chr.9: 137188614-137188614 on Build GRCh38
rs752551945 Chr.9: 137188527-137188527 on Build GRCh38
rs750007776 Chr.9: 137188387-137188387 on Build GRCh38
rs775027744 Chr.9: 137188747-137188747 on Build GRCh38
rs749358216 Chr.9: 137188538-137188538 on Build GRCh38
rs377619980 Chr.9: 137188533-137188533 on Build GRCh38
rs769058672 Chr.9: 137188539-137188539 on Build GRCh38
rs577285892 Chr.9: 137188718-137188718 on Build GRCh38
rs981867711 Chr.9: 137188606-137188606 on Build GRCh38
rs546290485 Chr.9: 137188458-137188458 on Build GRCh38
rs772302205 Chr.9: 137188680-137188680 on Build GRCh38
rs747130835 Chr.9: 137188716-137188716 on Build GRCh38
rs373415628 Chr.9: 137188703-137188703 on Build GRCh38
rs753352370 Chr.9: 137188514-137188514 on Build GRCh38
rs913712965 Chr.9: 137188375-137188375 on Build GRCh38
rs751930913 Chr.9: 137188373-137188373 on Build GRCh38
rs183026742 Chr.9: 137188605-137188605 on Build GRCh38
rs909764985 Chr.9: 137188643-137188643 on Build GRCh38
rs762509728 Chr.9: 137188758-137188758 on Build GRCh38
rs370229437 Chr.9: 137188762-137188762 on Build GRCh38
rs759039069 Chr.9: 137188564-137188564 on Build GRCh38
rs765832676 Chr.9: 137188504-137188504 on Build GRCh38
rs757189099 Chr.9: 137188582-137188582 on Build GRCh38
rs757996975 Chr.9: 137188573-137188573 on Build GRCh38
rs763722577 Chr.9: 137188759-137188759 on Build GRCh38
rs546197339 Chr.9: 137188576-137188576 on Build GRCh38
rs767443753 Chr.9: 137188765-137188765 on Build GRCh38
rs932693389 Chr.9: 137188429-137188429 on Build GRCh38
rs962721498 Chr.9: 137188722-137188722 on Build GRCh38
rs775980857 Chr.9: 137188595-137188595 on Build GRCh38
rs761191548 Chr.9: 137188500-137188500 on Build GRCh38
rs778713960 Chr.9: 137188404-137188404 on Build GRCh38
rs375988347 Chr.9: 137188584-137188584 on Build GRCh38
rs879165221 Chr.9: 137188572-137188572 on Build GRCh38

View Full Product Details