Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127802783-127803280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FPGS
Entrez Gene ID
2356
Gene Name
folylpolyglutamate synthase
Gene Aliases
-
UniGene
Hs.335084
Gene Chromosome Location
Chr.9: 127802858-127814520 on Build GRCh38
Transcripts
NM_001018078.2 NM_001288803.1 NM_004957.5 NR_110170.1 XM_005251864.3 XM_017014565.1 XM_017014566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3750153 c.64A>G p.I22V Chr.9: 127802988-127802988 on Build GRCh38
4406593 c.133T>G p.Y45D Chr.9: 127803057-127803057 on Build GRCh38
6211559 c.15G>A p.R5R Chr.9: 127802939-127802939 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs904247682 Chr.9: 127802893-127802893 on Build GRCh38
rs1034067436 Chr.9: 127802996-127802996 on Build GRCh38
rs935418118 Chr.9: 127802836-127802836 on Build GRCh38
rs983928288 Chr.9: 127802958-127802958 on Build GRCh38
rs780426708 Chr.9: 127803088-127803088 on Build GRCh38
rs1014032496 Chr.9: 127803182-127803182 on Build GRCh38
rs746485228 Chr.9: 127803094-127803094 on Build GRCh38
rs1034425998 Chr.9: 127803100-127803100 on Build GRCh38
rs772896112 Chr.9: 127803092-127803092 on Build GRCh38
rs974845044 Chr.9: 127802908-127802908 on Build GRCh38
rs566648760 Chr.9: 127802919-127802919 on Build GRCh38
rs895462892 Chr.9: 127803175-127803176 on Build GRCh38
rs941994071 Chr.9: 127802889-127802889 on Build GRCh38
rs753615973 Chr.9: 127802954-127802954 on Build GRCh38
rs992650940 Chr.9: 127803123-127803123 on Build GRCh38
rs1055503292 Chr.9: 127803175-127803175 on Build GRCh38
rs771638675 Chr.9: 127802909-127802909 on Build GRCh38
rs1008651151 Chr.9: 127802937-127802937 on Build GRCh38
rs986316496 Chr.9: 127803237-127803237 on Build GRCh38
rs11554716 Chr.9: 127803047-127803047 on Build GRCh38
rs952769410 Chr.9: 127802951-127802951 on Build GRCh38
rs1052564290 Chr.9: 127802841-127802841 on Build GRCh38
rs759336102 Chr.9: 127802961-127802961 on Build GRCh38
rs890049327 Chr.9: 127802912-127802912 on Build GRCh38
rs1005027355 Chr.9: 127803192-127803192 on Build GRCh38
rs922011451 Chr.9: 127802910-127802910 on Build GRCh38
rs765099606 Chr.9: 127802966-127802966 on Build GRCh38
rs974990425 Chr.9: 127803241-127803241 on Build GRCh38
rs901411365 Chr.9: 127803084-127803084 on Build GRCh38
rs1050182157 Chr.9: 127803043-127803043 on Build GRCh38
rs1051567652 Chr.9: 127802923-127802923 on Build GRCh38
rs893783425 Chr.9: 127802871-127802871 on Build GRCh38
rs994111687 Chr.9: 127802930-127802930 on Build GRCh38
rs963584482 Chr.9: 127803226-127803226 on Build GRCh38
rs1050278582 Chr.9: 127802885-127802885 on Build GRCh38
rs760460465 Chr.9: 127802943-127802943 on Build GRCh38
rs774062519 Chr.9: 127803076-127803076 on Build GRCh38
rs766106139 Chr.9: 127802952-127802952 on Build GRCh38
rs904215517 Chr.9: 127802967-127802967 on Build GRCh38
rs866970380 Chr.9: 127803009-127803009 on Build GRCh38
rs965789915 Chr.9: 127802926-127802926 on Build GRCh38
rs1001045385 Chr.9: 127802903-127802903 on Build GRCh38
rs1041719551 Chr.9: 127803077-127803077 on Build GRCh38
rs1042935574 Chr.9: 127802964-127802964 on Build GRCh38
rs528236662 Chr.9: 127802945-127802945 on Build GRCh38
rs1016534186 Chr.9: 127803213-127803213 on Build GRCh38
rs935695766 Chr.9: 127803017-127803017 on Build GRCh38
rs1029030365 Chr.9: 127803249-127803249 on Build GRCh38
rs371032601 Chr.9: 127803101-127803101 on Build GRCh38
rs764218816 Chr.9: 127802986-127802986 on Build GRCh38
rs10760502 Chr.9: 127802988-127802988 on Build GRCh38
rs540121654 Chr.9: 127803186-127803186 on Build GRCh38
rs138322374 Chr.9: 127803090-127803090 on Build GRCh38
rs1034635821 Chr.9: 127802963-127802963 on Build GRCh38
rs758265682 Chr.9: 127802980-127802980 on Build GRCh38
rs977085543 Chr.9: 127803161-127803161 on Build GRCh38
rs545631206 Chr.9: 127802824-127802824 on Build GRCh38
rs1007595326 Chr.9: 127802918-127802918 on Build GRCh38
rs945710832 Chr.9: 127802984-127802984 on Build GRCh38
rs370828833 Chr.9: 127803147-127803147 on Build GRCh38
rs889044334 Chr.9: 127802892-127802892 on Build GRCh38
rs534841452 Chr.9: 127803103-127803103 on Build GRCh38
rs1027434816 Chr.9: 127803214-127803214 on Build GRCh38
rs752614531 Chr.9: 127802968-127802968 on Build GRCh38
rs781355383 Chr.9: 127803038-127803038 on Build GRCh38
rs945994284 Chr.9: 127802938-127802938 on Build GRCh38
rs1010928619 Chr.9: 127802876-127802876 on Build GRCh38
rs866663999 Chr.9: 127803030-127803030 on Build GRCh38
rs1024653618 Chr.9: 127803132-127803132 on Build GRCh38
rs550566089 Chr.9: 127803113-127803113 on Build GRCh38
rs769220901 Chr.9: 127803070-127803077 on Build GRCh38
rs1025799335 Chr.9: 127803184-127803184 on Build GRCh38
rs895525821 Chr.9: 127803000-127803000 on Build GRCh38
rs757514010 Chr.9: 127803012-127803012 on Build GRCh38
rs993181344 Chr.9: 127802977-127802977 on Build GRCh38
rs115992881 Chr.9: 127803168-127803168 on Build GRCh38
rs1014416938 Chr.9: 127803001-127803001 on Build GRCh38
rs945858843 Chr.9: 127803145-127803145 on Build GRCh38
rs913087646 Chr.9: 127803108-127803108 on Build GRCh38
rs933158045 Chr.9: 127802913-127802913 on Build GRCh38
rs1025566033 Chr.9: 127803091-127803091 on Build GRCh38

View Full Product Details