Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128742451-128742895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZER1
Entrez Gene ID
10444
Gene Name
zyg-11 related cell cycle regulator
Gene Aliases
C9orf60, ZYG, ZYG11BL
UniGene
Hs.147950
Gene Chromosome Location
Chr.9: 128729786-128772507 on Build GRCh38
Transcripts
NM_006336.3 XM_005251645.2 XM_005251646.2 XM_011518122.2 XM_017014185.1 XM_017014186.1 XM_017014187.1 XM_017014188.1 XM_017014189.1 XM_017014190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6852833 c.1523G>T p.C508F Chr.9: 128742582-128742582 on Build GRCh38
6485555 c.1541A>T p.H514L Chr.9: 128742564-128742564 on Build GRCh38
4761421 c.1498G>A p.V500M Chr.9: 128742607-128742607 on Build GRCh38
1233324 c.1417T>G p.F473V Chr.9: 128742688-128742688 on Build GRCh38
5583114 c.1567T>G p.F523V Chr.9: 128742538-128742538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768832521 Chr.9: 128742607-128742607 on Build GRCh38
rs781047932 Chr.9: 128742787-128742787 on Build GRCh38
rs762675056 Chr.9: 128742769-128742769 on Build GRCh38
rs908626349 Chr.9: 128742639-128742639 on Build GRCh38
rs904908253 Chr.9: 128742833-128742833 on Build GRCh38
rs763729484 Chr.9: 128742641-128742641 on Build GRCh38
rs10739731 Chr.9: 128742813-128742813 on Build GRCh38
rs532786216 Chr.9: 128742779-128742779 on Build GRCh38
rs762243667 Chr.9: 128742500-128742500 on Build GRCh38
rs867731338 Chr.9: 128742564-128742564 on Build GRCh38
rs151193282 Chr.9: 128742521-128742521 on Build GRCh38
rs772724504 Chr.9: 128742707-128742707 on Build GRCh38
rs752166640 Chr.9: 128742520-128742520 on Build GRCh38
rs199888547 Chr.9: 128742514-128742514 on Build GRCh38
rs1003345640 Chr.9: 128742473-128742473 on Build GRCh38
rs762476242 Chr.9: 128742610-128742610 on Build GRCh38
rs753536763 Chr.9: 128742509-128742509 on Build GRCh38
rs751434921 Chr.9: 128742659-128742659 on Build GRCh38
rs956178829 Chr.9: 128742488-128742488 on Build GRCh38
rs775004024 Chr.9: 128742755-128742755 on Build GRCh38
rs773725126 Chr.9: 128742626-128742626 on Build GRCh38
rs201549935 Chr.9: 128742608-128742608 on Build GRCh38
rs936344305 Chr.9: 128742863-128742863 on Build GRCh38
rs376716226 Chr.9: 128742753-128742753 on Build GRCh38
rs201770268 Chr.9: 128742679-128742679 on Build GRCh38
rs747302235 Chr.9: 128742752-128742752 on Build GRCh38
rs746036225 Chr.9: 128742572-128742572 on Build GRCh38
rs767412601 Chr.9: 128742790-128742790 on Build GRCh38
rs774964774 Chr.9: 128742493-128742493 on Build GRCh38
rs757203967 Chr.9: 128742669-128742669 on Build GRCh38
rs1044419715 Chr.9: 128742856-128742856 on Build GRCh38
rs779438418 Chr.9: 128742569-128742569 on Build GRCh38
rs750249910 Chr.9: 128742678-128742678 on Build GRCh38
rs780119077 Chr.9: 128742680-128742680 on Build GRCh38
rs552493910 Chr.9: 128742532-128742532 on Build GRCh38
rs749579949 Chr.9: 128742684-128742684 on Build GRCh38
rs535056384 Chr.9: 128742605-128742605 on Build GRCh38
rs769932301 Chr.9: 128742601-128742601 on Build GRCh38
rs769076517 Chr.9: 128742484-128742484 on Build GRCh38
rs780973576 Chr.9: 128742553-128742553 on Build GRCh38
rs764825206 Chr.9: 128742636-128742636 on Build GRCh38
rs755007423 Chr.9: 128742699-128742699 on Build GRCh38
rs540873746 Chr.9: 128742611-128742611 on Build GRCh38
rs775446180 Chr.9: 128742482-128742482 on Build GRCh38
rs371028406 Chr.9: 128742789-128742789 on Build GRCh38
rs201468706 Chr.9: 128742847-128742847 on Build GRCh38
rs377315216 Chr.9: 128742526-128742526 on Build GRCh38
rs771939300 Chr.9: 128742629-128742629 on Build GRCh38
rs746522166 Chr.9: 128742638-128742638 on Build GRCh38
rs775246598 Chr.9: 128742637-128742637 on Build GRCh38
rs767251280 Chr.9: 128742675-128742675 on Build GRCh38
rs575010117 Chr.9: 128742515-128742515 on Build GRCh38
rs183927008 Chr.9: 128742533-128742533 on Build GRCh38
rs188207744 Chr.9: 128742844-128742844 on Build GRCh38
rs149251289 Chr.9: 128742701-128742701 on Build GRCh38
rs763469728 Chr.9: 128742483-128742483 on Build GRCh38
rs1028384558 Chr.9: 128742676-128742676 on Build GRCh38
rs773667866 Chr.9: 128742719-128742719 on Build GRCh38
rs796313337 Chr.9: 128742808-128742808 on Build GRCh38

View Full Product Details