Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153794461-153794966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDH3G
Entrez Gene ID
3421
Gene Name
isocitrate dehydrogenase 3 (NAD(+)) gamma
Gene Aliases
H-IDHG
UniGene
Hs.410197
Gene Chromosome Location
Chr.X: 153785766-153794523 on Build GRCh38
Transcripts
NM_004135.3 NM_174869.2 NM_001204526.1 NM_001204527.1 NM_006280.2 NR_037927.1 XM_011531186.1 XM_011531187.1 XM_017029756.1 XM_017029757.1

Gene Symbol
SSR4
Entrez Gene ID
6748
Gene Name
signal sequence receptor subunit 4
Gene Aliases
CDG1Y, TRAPD
UniGene
Hs.409223
Gene Chromosome Location
Chr.X: 153794175-153798512 on Build GRCh38
Transcripts
NM_004135.3 NM_174869.2 NM_001204526.1 NM_001204527.1 NM_006280.2 NR_037927.1 XM_011531186.1 XM_011531187.1 XM_017029756.1 XM_017029757.1

SNP Detail

SNP ID Chromosome Location
rs781966565 Chr.X: 153794695-153794695 on Build GRCh38
rs782443260 Chr.X: 153794727-153794727 on Build GRCh38
rs782683933 Chr.X: 153794561-153794561 on Build GRCh38
rs782457591 Chr.X: 153794649-153794649 on Build GRCh38
rs782662159 Chr.X: 153794643-153794643 on Build GRCh38
rs782353229 Chr.X: 153794909-153794909 on Build GRCh38
rs782011695 Chr.X: 153794699-153794699 on Build GRCh38
rs372799541 Chr.X: 153794837-153794837 on Build GRCh38
rs782284661 Chr.X: 153794654-153794654 on Build GRCh38
rs782162337 Chr.X: 153794601-153794601 on Build GRCh38
rs782307452 Chr.X: 153794766-153794766 on Build GRCh38
rs782282094 Chr.X: 153794830-153794830 on Build GRCh38
rs781972434 Chr.X: 153794492-153794492 on Build GRCh38
rs782343090 Chr.X: 153794550-153794550 on Build GRCh38
rs782224678 Chr.X: 153794758-153794758 on Build GRCh38
rs957141930 Chr.X: 153794873-153794873 on Build GRCh38
rs782582757 Chr.X: 153794788-153794788 on Build GRCh38
rs781797447 Chr.X: 153794619-153794619 on Build GRCh38
rs977554126 Chr.X: 153794502-153794502 on Build GRCh38
rs370819935 Chr.X: 153794607-153794607 on Build GRCh38
rs919220847 Chr.X: 153794635-153794635 on Build GRCh38
rs998518531 Chr.X: 153794810-153794810 on Build GRCh38
rs782054111 Chr.X: 153794496-153794497 on Build GRCh38
rs782695514 Chr.X: 153794606-153794606 on Build GRCh38
rs782631593 Chr.X: 153794524-153794524 on Build GRCh38
rs781989811 Chr.X: 153794902-153794902 on Build GRCh38
rs782222222 Chr.X: 153794796-153794796 on Build GRCh38
rs915709748 Chr.X: 153794899-153794899 on Build GRCh38
rs782786164 Chr.X: 153794571-153794571 on Build GRCh38
rs782679700 Chr.X: 153794787-153794787 on Build GRCh38
rs782111229 Chr.X: 153794696-153794696 on Build GRCh38
rs372335182 Chr.X: 153794637-153794637 on Build GRCh38
rs1008163661 Chr.X: 153794663-153794663 on Build GRCh38
rs374141854 Chr.X: 153794653-153794653 on Build GRCh38
rs11538940 Chr.X: 153794726-153794726 on Build GRCh38
rs782678482 Chr.X: 153794656-153794656 on Build GRCh38
rs782234497 Chr.X: 153794651-153794651 on Build GRCh38
rs782121089 Chr.X: 153794558-153794558 on Build GRCh38
rs375831510 Chr.X: 153794647-153794647 on Build GRCh38
rs781975256 Chr.X: 153794553-153794553 on Build GRCh38
rs782552919 Chr.X: 153794784-153794784 on Build GRCh38
rs782500246 Chr.X: 153794681-153794682 on Build GRCh38
rs1029354847 Chr.X: 153794765-153794765 on Build GRCh38
rs782321163 Chr.X: 153794793-153794793 on Build GRCh38
rs925822637 Chr.X: 153794509-153794509 on Build GRCh38
rs782079597 Chr.X: 153794905-153794905 on Build GRCh38
rs782061175 Chr.X: 153794667-153794667 on Build GRCh38
rs782716135 Chr.X: 153794738-153794738 on Build GRCh38
rs782022260 Chr.X: 153794521-153794521 on Build GRCh38
rs781941547 Chr.X: 153794795-153794795 on Build GRCh38
rs991321163 Chr.X: 153794531-153794531 on Build GRCh38
rs782265237 Chr.X: 153794773-153794773 on Build GRCh38
rs375165935 Chr.X: 153794701-153794702 on Build GRCh38
rs781896845 Chr.X: 153794574-153794574 on Build GRCh38
rs112969179 Chr.X: 153794541-153794541 on Build GRCh38
rs1033199173 Chr.X: 153794832-153794832 on Build GRCh38
rs782489766 Chr.X: 153794546-153794546 on Build GRCh38
rs781921159 Chr.X: 153794535-153794535 on Build GRCh38
rs782711765 Chr.X: 153794612-153794613 on Build GRCh38
rs782420694 Chr.X: 153794844-153794844 on Build GRCh38
rs919271834 Chr.X: 153794918-153794918 on Build GRCh38
rs782246006 Chr.X: 153794683-153794684 on Build GRCh38
rs782523022 Chr.X: 153794693-153794693 on Build GRCh38
rs4908 Chr.X: 153794750-153794750 on Build GRCh38
rs981301306 Chr.X: 153794903-153794903 on Build GRCh38
rs782774093 Chr.X: 153794528-153794528 on Build GRCh38
rs991766613 Chr.X: 153794880-153794880 on Build GRCh38
rs781824791 Chr.X: 153794742-153794742 on Build GRCh38
rs782604567 Chr.X: 153794719-153794719 on Build GRCh38
rs929324226 Chr.X: 153794934-153794934 on Build GRCh38
rs782625708 Chr.X: 153794630-153794630 on Build GRCh38
rs967124453 Chr.X: 153794498-153794498 on Build GRCh38
rs781802779 Chr.X: 153794626-153794626 on Build GRCh38
rs781916038 Chr.X: 153794661-153794661 on Build GRCh38
rs782736375 Chr.X: 153794497-153794497 on Build GRCh38
rs781852813 Chr.X: 153794634-153794634 on Build GRCh38
rs537293003 Chr.X: 153794760-153794760 on Build GRCh38

View Full Product Details