Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 110964023-110964524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHOC2
Entrez Gene ID
8036
Gene Name
SHOC2, leucine rich repeat scaffold protein
Gene Aliases
SIAA0862, SOC2, SUR8
UniGene
Hs.104315
Gene Chromosome Location
Chr.10: 110919370-111013667 on Build GRCh38
Transcripts
NM_001269039.2 NM_001324336.1 NM_001324337.1 NM_007373.3 NR_136749.1 XM_017016702.1 XM_017016703.1 XM_017016704.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6567757 c.59C>T p.S20L Chr.10: 110964417-110964417 on Build GRCh38
6863474 c.91T>C p.S31P Chr.10: 110964449-110964449 on Build GRCh38
3434108 c.92C>T p.S31F Chr.10: 110964450-110964450 on Build GRCh38
914776 c.22G>T p.E8* Chr.10: 110964380-110964380 on Build GRCh38
3665494 c.123A>G p.K41K Chr.10: 110964481-110964481 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs267607048 Chr.10: 110964362-110964362 on Build GRCh38
rs730881019 Chr.10: 110964432-110964432 on Build GRCh38
rs937822393 Chr.10: 110964255-110964255 on Build GRCh38
rs902146493 Chr.10: 110964323-110964323 on Build GRCh38
rs751634012 Chr.10: 110964461-110964461 on Build GRCh38
rs560817115 Chr.10: 110964172-110964172 on Build GRCh38
rs376966948 Chr.10: 110964131-110964131 on Build GRCh38
rs730881018 Chr.10: 110964396-110964396 on Build GRCh38
rs757303783 Chr.10: 110964465-110964465 on Build GRCh38
rs1020324522 Chr.10: 110964306-110964306 on Build GRCh38
rs904505084 Chr.10: 110964316-110964316 on Build GRCh38
rs560036999 Chr.10: 110964133-110964133 on Build GRCh38
rs397517231 Chr.10: 110964368-110964368 on Build GRCh38
rs765714933 Chr.10: 110964436-110964436 on Build GRCh38
rs905061787 Chr.10: 110964210-110964210 on Build GRCh38
rs764545601 Chr.10: 110964460-110964460 on Build GRCh38
rs1054997592 Chr.10: 110964260-110964260 on Build GRCh38
rs138055318 Chr.10: 110964435-110964435 on Build GRCh38
rs757468449 Chr.10: 110964348-110964348 on Build GRCh38
rs758965011 Chr.10: 110964450-110964450 on Build GRCh38
rs566047015 Chr.10: 110964334-110964334 on Build GRCh38
rs774208246 Chr.10: 110964372-110964372 on Build GRCh38
rs747122393 Chr.10: 110964335-110964335 on Build GRCh38
rs766010467 Chr.10: 110964416-110964416 on Build GRCh38
rs547869981 Chr.10: 110964317-110964317 on Build GRCh38
rs1054545590 Chr.10: 110964389-110964390 on Build GRCh38
rs542226017 Chr.10: 110964171-110964171 on Build GRCh38
rs946634032 Chr.10: 110964294-110964294 on Build GRCh38
rs939150628 Chr.10: 110964225-110964225 on Build GRCh38
rs915074052 Chr.10: 110964274-110964274 on Build GRCh38
rs398124252 Chr.10: 110964358-110964358 on Build GRCh38
rs150364514 Chr.10: 110964385-110964385 on Build GRCh38
rs188765124 Chr.10: 110964234-110964234 on Build GRCh38
rs760154441 Chr.10: 110964405-110964405 on Build GRCh38
rs929676640 Chr.10: 110964146-110964146 on Build GRCh38
rs143180451 Chr.10: 110964180-110964180 on Build GRCh38
rs1050830219 Chr.10: 110964383-110964383 on Build GRCh38
rs895863051 Chr.10: 110964253-110964253 on Build GRCh38
rs397517230 Chr.10: 110964293-110964293 on Build GRCh38
rs982384703 Chr.10: 110964154-110964154 on Build GRCh38
rs749032903 Chr.10: 110964365-110964365 on Build GRCh38
rs753165612 Chr.10: 110964442-110964442 on Build GRCh38
rs1010252920 Chr.10: 110964273-110964273 on Build GRCh38
rs951163627 Chr.10: 110964449-110964449 on Build GRCh38
rs781434461 Chr.10: 110964471-110964471 on Build GRCh38
rs549693975 Chr.10: 110964196-110964196 on Build GRCh38
rs1057217316 Chr.10: 110964245-110964245 on Build GRCh38
rs981482001 Chr.10: 110964224-110964224 on Build GRCh38
rs527240159 Chr.10: 110964157-110964157 on Build GRCh38
rs976561291 Chr.10: 110964099-110964099 on Build GRCh38
rs72819758 Chr.10: 110964200-110964200 on Build GRCh38
rs372210369 Chr.10: 110964326-110964326 on Build GRCh38
rs761608447 Chr.10: 110964176-110964176 on Build GRCh38
rs1029427711 Chr.10: 110964447-110964447 on Build GRCh38

View Full Product Details