Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 113577939-113578434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HABP2
Entrez Gene ID
3026
Gene Name
hyaluronan binding protein 2
Gene Aliases
FSAP, HABP, HGFAL, NMTC5, PHBP
UniGene
Hs.422542
Gene Chromosome Location
Chr.10: 113550831-113589602 on Build GRCh38
Transcripts
NM_001177660.2 NM_004132.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
914893 c.507G>T p.K169N Chr.10: 113578084-113578084 on Build GRCh38
7112520 c.459T>A p.P153P Chr.10: 113578070-113578070 on Build GRCh38
6892012 c.558C>T p.F186F Chr.10: 113578135-113578135 on Build GRCh38
6892013 c.524C>T p.S175F Chr.10: 113578135-113578135 on Build GRCh38
3434197 c.454C>T p.P152S Chr.10: 113578031-113578031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1012442730 Chr.10: 113578051-113578051 on Build GRCh38
rs944021837 Chr.10: 113577966-113577966 on Build GRCh38
rs371091192 Chr.10: 113578127-113578127 on Build GRCh38
rs754504728 Chr.10: 113578115-113578115 on Build GRCh38
rs762276904 Chr.10: 113577998-113577998 on Build GRCh38
rs767415703 Chr.10: 113578102-113578102 on Build GRCh38
rs767195288 Chr.10: 113578179-113578179 on Build GRCh38
rs201266591 Chr.10: 113578148-113578148 on Build GRCh38
rs559972478 Chr.10: 113578119-113578119 on Build GRCh38
rs199727283 Chr.10: 113578152-113578152 on Build GRCh38
rs368835886 Chr.10: 113578085-113578085 on Build GRCh38
rs538405893 Chr.10: 113578059-113578059 on Build GRCh38
rs1034555808 Chr.10: 113578162-113578162 on Build GRCh38
rs771431897 Chr.10: 113578151-113578151 on Build GRCh38
rs769362048 Chr.10: 113577985-113577985 on Build GRCh38
rs779506767 Chr.10: 113577991-113577991 on Build GRCh38
rs888567816 Chr.10: 113578203-113578203 on Build GRCh38
rs745314699 Chr.10: 113577979-113577979 on Build GRCh38
rs757143238 Chr.10: 113578034-113578034 on Build GRCh38
rs971690485 Chr.10: 113578149-113578149 on Build GRCh38
rs753998839 Chr.10: 113578010-113578010 on Build GRCh38
rs748918390 Chr.10: 113578063-113578063 on Build GRCh38
rs1041057465 Chr.10: 113577969-113577969 on Build GRCh38
rs527563321 Chr.10: 113578316-113578316 on Build GRCh38
rs569429050 Chr.10: 113578069-113578069 on Build GRCh38
rs766481694 Chr.10: 113578006-113578006 on Build GRCh38
rs780935895 Chr.10: 113578046-113578046 on Build GRCh38
rs573698414 Chr.10: 113578201-113578201 on Build GRCh38
rs376762540 Chr.10: 113578297-113578297 on Build GRCh38
rs543856067 Chr.10: 113578212-113578212 on Build GRCh38
rs11575756 Chr.10: 113578288-113578288 on Build GRCh38
rs374554126 Chr.10: 113578064-113578064 on Build GRCh38
rs113801070 Chr.10: 113578250-113578250 on Build GRCh38
rs778566957 Chr.10: 113578065-113578065 on Build GRCh38
rs956367134 Chr.10: 113578093-113578093 on Build GRCh38
rs546987507 Chr.10: 113578326-113578326 on Build GRCh38
rs199906097 Chr.10: 113578146-113578146 on Build GRCh38
rs774080665 Chr.10: 113578283-113578283 on Build GRCh38
rs923305396 Chr.10: 113578242-113578242 on Build GRCh38
rs760418017 Chr.10: 113578190-113578190 on Build GRCh38
rs761636169 Chr.10: 113578095-113578095 on Build GRCh38
rs371724336 Chr.10: 113578375-113578375 on Build GRCh38
rs932187436 Chr.10: 113578244-113578244 on Build GRCh38
rs753305812 Chr.10: 113578114-113578114 on Build GRCh38
rs774745762 Chr.10: 113577997-113577997 on Build GRCh38
rs773789415 Chr.10: 113578005-113578005 on Build GRCh38
rs532998009 Chr.10: 113578232-113578232 on Build GRCh38
rs573445354 Chr.10: 113578110-113578110 on Build GRCh38
rs765775244 Chr.10: 113578111-113578111 on Build GRCh38
rs775851480 Chr.10: 113578173-113578173 on Build GRCh38
rs150203631 Chr.10: 113578392-113578392 on Build GRCh38
rs758975611 Chr.10: 113578345-113578345 on Build GRCh38
rs766291330 Chr.10: 113578197-113578197 on Build GRCh38
rs145980446 Chr.10: 113578076-113578076 on Build GRCh38
rs747980965 Chr.10: 113577961-113577961 on Build GRCh38
rs148113667 Chr.10: 113578224-113578224 on Build GRCh38
rs747800162 Chr.10: 113578136-113578136 on Build GRCh38
rs745887427 Chr.10: 113578161-113578161 on Build GRCh38
rs897135095 Chr.10: 113578163-113578163 on Build GRCh38
rs142936446 Chr.10: 113578074-113578074 on Build GRCh38
rs768995711 Chr.10: 113577995-113577995 on Build GRCh38
rs896888072 Chr.10: 113578312-113578313 on Build GRCh38
rs755140777 Chr.10: 113578072-113578072 on Build GRCh38
rs141890505 Chr.10: 113578287-113578287 on Build GRCh38
rs748460774 Chr.10: 113578067-113578067 on Build GRCh38
rs202053426 Chr.10: 113578132-113578132 on Build GRCh38
rs906697965 Chr.10: 113578239-113578239 on Build GRCh38
rs772527165 Chr.10: 113577999-113577999 on Build GRCh38
rs763955525 Chr.10: 113578011-113578011 on Build GRCh38
rs143527420 Chr.10: 113577994-113577994 on Build GRCh38
rs145891602 Chr.10: 113578048-113578048 on Build GRCh38
rs779983821 Chr.10: 113578058-113578058 on Build GRCh38
rs1050739708 Chr.10: 113578249-113578249 on Build GRCh38
rs11575757 Chr.10: 113578347-113578347 on Build GRCh38
rs779673888 Chr.10: 113578056-113578056 on Build GRCh38
rs956336184 Chr.10: 113578285-113578285 on Build GRCh38
rs148642824 Chr.10: 113578140-113578140 on Build GRCh38
rs773100199 Chr.10: 113578181-113578181 on Build GRCh38
rs750314701 Chr.10: 113578107-113578107 on Build GRCh38
rs558470144 Chr.10: 113578077-113578077 on Build GRCh38
rs375076367 Chr.10: 113578167-113578167 on Build GRCh38
rs775871794 Chr.10: 113578086-113578086 on Build GRCh38
rs755895173 Chr.10: 113577983-113577985 on Build GRCh38
rs778254844 Chr.10: 113578145-113578145 on Build GRCh38
rs946011729 Chr.10: 113578225-113578225 on Build GRCh38
rs375742994 Chr.10: 113577996-113577996 on Build GRCh38
rs551142296 Chr.10: 113578390-113578390 on Build GRCh38
rs11575755 Chr.10: 113578154-113578154 on Build GRCh38
rs370893980 Chr.10: 113578015-113578015 on Build GRCh38
rs368377443 Chr.10: 113578174-113578174 on Build GRCh38

View Full Product Details