Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 7620846-7621351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPFIBP2
Entrez Gene ID
8495
Gene Name
PPFIA binding protein 2
Gene Aliases
Cclp1
UniGene
Hs.655714
Gene Chromosome Location
Chr.11: 7513765-7667329 on Build GRCh38
Transcripts
NM_001256568.1 NM_001256569.1 NM_003621.3 XM_005253177.2 XM_006718349.2 XM_006718350.2 XM_006718351.2 XM_006718352.2 XM_006718353.2 XM_011520413.2 XM_011520414.2 XM_011520415.2 XM_011520416.2 XM_011520417.2 XM_011520418.2 XM_011520419.2 XM_011520420.2 XM_017018443.1 XM_017018444.1 XM_017018445.1 XM_017018446.1 XM_017018447.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4505060 c.682C>T p.Q228* Chr.11: 7620998-7620998 on Build GRCh38
7426909 c.706A>G p.T236A Chr.11: 7621022-7621022 on Build GRCh38
2018484 c.619G>T p.E207* Chr.11: 7620935-7620935 on Build GRCh38
1605245 c.619-10C>T p.? Chr.11: 7620925-7620925 on Build GRCh38
4716922 c.667_668insA p.N224fs*2 Chr.11: 7620983-7620984 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772645735 Chr.11: 7620872-7620872 on Build GRCh38
rs1026188587 Chr.11: 7620873-7620873 on Build GRCh38
rs1013071391 Chr.11: 7621108-7621108 on Build GRCh38
rs975905986 Chr.11: 7621192-7621192 on Build GRCh38
rs577711302 Chr.11: 7621202-7621202 on Build GRCh38
rs917417779 Chr.11: 7621296-7621296 on Build GRCh38
rs12272404 Chr.11: 7621200-7621200 on Build GRCh38
rs768965314 Chr.11: 7620886-7620886 on Build GRCh38
rs898886216 Chr.11: 7621319-7621319 on Build GRCh38
rs986802800 Chr.11: 7621282-7621282 on Build GRCh38
rs140084492 Chr.11: 7621067-7621067 on Build GRCh38
rs1044105571 Chr.11: 7621116-7621116 on Build GRCh38
rs908510979 Chr.11: 7621299-7621299 on Build GRCh38
rs975475068 Chr.11: 7620979-7620979 on Build GRCh38
rs759073996 Chr.11: 7620926-7620926 on Build GRCh38
rs1044542876 Chr.11: 7621307-7621307 on Build GRCh38
rs765467279 Chr.11: 7621137-7621137 on Build GRCh38
rs530980592 Chr.11: 7621283-7621283 on Build GRCh38
rs80061616 Chr.11: 7621278-7621278 on Build GRCh38
rs898826964 Chr.11: 7621135-7621135 on Build GRCh38
rs138013370 Chr.11: 7620983-7620983 on Build GRCh38
rs781557445 Chr.11: 7620894-7620894 on Build GRCh38
rs895598694 Chr.11: 7621097-7621097 on Build GRCh38
rs368014210 Chr.11: 7621038-7621038 on Build GRCh38
rs368814947 Chr.11: 7621272-7621272 on Build GRCh38
rs376799754 Chr.11: 7621033-7621033 on Build GRCh38
rs950561029 Chr.11: 7620889-7620889 on Build GRCh38
rs921262588 Chr.11: 7620980-7620980 on Build GRCh38
rs1036251450 Chr.11: 7621313-7621313 on Build GRCh38
rs556692938 Chr.11: 7621106-7621106 on Build GRCh38
rs773086258 Chr.11: 7621026-7621027 on Build GRCh38
rs111940239 Chr.11: 7621076-7621076 on Build GRCh38
rs766084894 Chr.11: 7620949-7620949 on Build GRCh38
rs760573759 Chr.11: 7621048-7621048 on Build GRCh38
rs770589693 Chr.11: 7620923-7620923 on Build GRCh38
rs965474007 Chr.11: 7621191-7621191 on Build GRCh38
rs781695831 Chr.11: 7621000-7621000 on Build GRCh38
rs371984331 Chr.11: 7620956-7620956 on Build GRCh38
rs763103869 Chr.11: 7621073-7621073 on Build GRCh38
rs957532947 Chr.11: 7621233-7621233 on Build GRCh38
rs540180925 Chr.11: 7621217-7621217 on Build GRCh38
rs770901487 Chr.11: 7621073-7621074 on Build GRCh38
rs755458671 Chr.11: 7620999-7620999 on Build GRCh38
rs950611873 Chr.11: 7621173-7621173 on Build GRCh38
rs1047366598 Chr.11: 7621327-7621327 on Build GRCh38
rs375627103 Chr.11: 7621061-7621061 on Build GRCh38
rs576741503 Chr.11: 7621125-7621125 on Build GRCh38
rs994929019 Chr.11: 7621326-7621326 on Build GRCh38
rs934888470 Chr.11: 7621238-7621238 on Build GRCh38
rs924882446 Chr.11: 7621222-7621222 on Build GRCh38
rs771496649 Chr.11: 7620931-7620931 on Build GRCh38
rs771572698 Chr.11: 7621034-7621034 on Build GRCh38
rs536550610 Chr.11: 7621100-7621100 on Build GRCh38
rs934836275 Chr.11: 7621065-7621065 on Build GRCh38
rs750618918 Chr.11: 7620958-7620958 on Build GRCh38
rs1018727640 Chr.11: 7621188-7621188 on Build GRCh38
rs773946272 Chr.11: 7620925-7620925 on Build GRCh38
rs968076743 Chr.11: 7621205-7621205 on Build GRCh38
rs758432552 Chr.11: 7620968-7620968 on Build GRCh38
rs948873859 Chr.11: 7621298-7621298 on Build GRCh38
rs761610902 Chr.11: 7621056-7621056 on Build GRCh38
rs10839822 Chr.11: 7621176-7621176 on Build GRCh38
rs1046607380 Chr.11: 7621150-7621150 on Build GRCh38
rs529152367 Chr.11: 7621295-7621295 on Build GRCh38
rs533969622 Chr.11: 7621019-7621019 on Build GRCh38
rs115874491 Chr.11: 7621055-7621055 on Build GRCh38
rs150433585 Chr.11: 7620952-7620952 on Build GRCh38
rs369413255 Chr.11: 7620933-7620933 on Build GRCh38
rs921376796 Chr.11: 7621255-7621255 on Build GRCh38
rs761765318 Chr.11: 7620955-7620955 on Build GRCh38
rs764174368 Chr.11: 7620934-7620934 on Build GRCh38
rs371452929 Chr.11: 7621051-7621051 on Build GRCh38
rs780026081 Chr.11: 7620969-7620969 on Build GRCh38
rs61888791 Chr.11: 7620876-7620876 on Build GRCh38
rs770118942 Chr.11: 7621027-7621027 on Build GRCh38
rs182769375 Chr.11: 7620916-7620916 on Build GRCh38

View Full Product Details