Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 7991793-7992298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3F
Entrez Gene ID
8665
Gene Name
eukaryotic translation initiation factor 3 subunit F
Gene Aliases
EIF3S5, eIF3-p47
UniGene
Hs.516023
Gene Chromosome Location
Chr.11: 7987320-7996171 on Build GRCh38
Transcripts
NM_003754.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3453843 c.489T>A p.V163V Chr.11: 7992137-7992137 on Build GRCh38
3935647 c.526A>T p.K176* Chr.11: 7992129-7992129 on Build GRCh38
6404124 c.460A>G p.K154E Chr.11: 7992108-7992108 on Build GRCh38
4390962 c.451G>T p.E151* Chr.11: 7992099-7992099 on Build GRCh38
6420772 c.523C>A p.H175N Chr.11: 7992126-7992126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34841314 Chr.11: 7991956-7991957 on Build GRCh38
rs771696683 Chr.11: 7992038-7992038 on Build GRCh38
rs748388254 Chr.11: 7992188-7992188 on Build GRCh38
rs778397241 Chr.11: 7992176-7992176 on Build GRCh38
rs934856271 Chr.11: 7992246-7992246 on Build GRCh38
rs768651293 Chr.11: 7992232-7992232 on Build GRCh38
rs996943849 Chr.11: 7992007-7992007 on Build GRCh38
rs1018859340 Chr.11: 7992089-7992089 on Build GRCh38
rs765882254 Chr.11: 7991875-7991875 on Build GRCh38
rs77896992 Chr.11: 7991932-7991935 on Build GRCh38
rs764021694 Chr.11: 7991867-7991867 on Build GRCh38
rs752237803 Chr.11: 7991882-7991882 on Build GRCh38
rs77396747 Chr.11: 7992156-7992156 on Build GRCh38
rs919816657 Chr.11: 7991889-7991889 on Build GRCh38
rs758652319 Chr.11: 7991880-7991880 on Build GRCh38
rs559726356 Chr.11: 7991982-7991982 on Build GRCh38
rs937931364 Chr.11: 7992195-7992195 on Build GRCh38
rs76503657 Chr.11: 7992186-7992186 on Build GRCh38
rs779748781 Chr.11: 7992225-7992225 on Build GRCh38
rs374664670 Chr.11: 7992031-7992031 on Build GRCh38
rs377659463 Chr.11: 7992183-7992183 on Build GRCh38
rs746245283 Chr.11: 7992051-7992051 on Build GRCh38
rs781580386 Chr.11: 7992112-7992112 on Build GRCh38
rs1045853015 Chr.11: 7992222-7992222 on Build GRCh38
rs763590401 Chr.11: 7991839-7991839 on Build GRCh38
rs115511435 Chr.11: 7991898-7991898 on Build GRCh38
rs762401046 Chr.11: 7991874-7991874 on Build GRCh38
rs777260376 Chr.11: 7991893-7991893 on Build GRCh38
rs756914335 Chr.11: 7992012-7992012 on Build GRCh38
rs1037589015 Chr.11: 7991921-7991921 on Build GRCh38
rs375040597 Chr.11: 7991987-7991987 on Build GRCh38
rs771509339 Chr.11: 7992204-7992204 on Build GRCh38
rs915869017 Chr.11: 7991910-7991910 on Build GRCh38
rs773790677 Chr.11: 7992135-7992135 on Build GRCh38
rs749738368 Chr.11: 7992209-7992209 on Build GRCh38
rs7941782 Chr.11: 7991903-7991903 on Build GRCh38
rs897510740 Chr.11: 7991928-7991928 on Build GRCh38
rs776817370 Chr.11: 7992095-7992095 on Build GRCh38
rs530604639 Chr.11: 7991873-7991873 on Build GRCh38
rs759897844 Chr.11: 7992208-7992208 on Build GRCh38
rs576942875 Chr.11: 7991915-7991915 on Build GRCh38
rs998994212 Chr.11: 7991916-7991916 on Build GRCh38
rs775532203 Chr.11: 7991841-7991841 on Build GRCh38
rs752489638 Chr.11: 7991895-7991895 on Build GRCh38
rs747185553 Chr.11: 7992067-7992067 on Build GRCh38
rs192330846 Chr.11: 7992033-7992033 on Build GRCh38
rs760955467 Chr.11: 7991858-7991858 on Build GRCh38
rs977378925 Chr.11: 7992242-7992242 on Build GRCh38
rs771592632 Chr.11: 7991931-7991932 on Build GRCh38
rs768974792 Chr.11: 7992185-7992185 on Build GRCh38
rs769939260 Chr.11: 7992189-7992189 on Build GRCh38
rs146954555 Chr.11: 7992161-7992161 on Build GRCh38
rs1044058 Chr.11: 7992163-7992163 on Build GRCh38
rs370850293 Chr.11: 7992144-7992144 on Build GRCh38
rs941946272 Chr.11: 7991912-7991912 on Build GRCh38
rs769062706 Chr.11: 7992094-7992094 on Build GRCh38
rs530699496 Chr.11: 7992122-7992122 on Build GRCh38
rs1028523774 Chr.11: 7991847-7991847 on Build GRCh38
rs139512698 Chr.11: 7992217-7992217 on Build GRCh38
rs879612019 Chr.11: 7991939-7991939 on Build GRCh38
rs762030714 Chr.11: 7992118-7992118 on Build GRCh38
rs763369944 Chr.11: 7992201-7992201 on Build GRCh38
rs573609056 Chr.11: 7992207-7992207 on Build GRCh38
rs557065077 Chr.11: 7991909-7991909 on Build GRCh38
rs993141468 Chr.11: 7991970-7991970 on Build GRCh38
rs61734577 Chr.11: 7992141-7992141 on Build GRCh38
rs755858263 Chr.11: 7991883-7991883 on Build GRCh38
rs189315686 Chr.11: 7992010-7992010 on Build GRCh38
rs543167950 Chr.11: 7991907-7991907 on Build GRCh38
rs930678013 Chr.11: 7991890-7991890 on Build GRCh38
rs545799816 Chr.11: 7991976-7991976 on Build GRCh38
rs750792058 Chr.11: 7991878-7991878 on Build GRCh38
rs777403718 Chr.11: 7992178-7992179 on Build GRCh38
rs563853145 Chr.11: 7991881-7991881 on Build GRCh38
rs955111065 Chr.11: 7991864-7991864 on Build GRCh38
rs928757982 Chr.11: 7992262-7992262 on Build GRCh38
rs1008433277 Chr.11: 7992079-7992079 on Build GRCh38
rs757165270 Chr.11: 7991901-7991901 on Build GRCh38
rs756718044 Chr.11: 7992172-7992172 on Build GRCh38
rs773403049 Chr.11: 7992197-7992197 on Build GRCh38
rs758061652 Chr.11: 7992074-7992075 on Build GRCh38
rs202109709 Chr.11: 7992152-7992152 on Build GRCh38
rs550436916 Chr.11: 7991887-7991887 on Build GRCh38
rs745733190 Chr.11: 7992182-7992182 on Build GRCh38
rs368962454 Chr.11: 7991830-7991830 on Build GRCh38
rs753879698 Chr.11: 7992238-7992238 on Build GRCh38
rs1055791369 Chr.11: 7991895-7991895 on Build GRCh38
rs915926859 Chr.11: 7992216-7992216 on Build GRCh38

View Full Product Details